Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 14522521-14523030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLGN4Y
Entrez Gene ID
22829
Gene Name
neuroligin 4, Y-linked
Gene Aliases
-
UniGene
Hs.439199
Gene Chromosome Location
Chr.Y: 14522578-14844945 on Build GRCh38
Transcripts
NM_001206850.1 XM_011531426.2 XM_017030035.1

SNP Detail

SNP ID Chromosome Location
rs563831392 Chr.Y: 14522818-14522818 on Build GRCh38
rs983213163 Chr.Y: 14522890-14522890 on Build GRCh38
rs979783457 Chr.Y: 14522593-14522593 on Build GRCh38
rs762642454 Chr.Y: 14522984-14522984 on Build GRCh38
rs959793816 Chr.Y: 14522715-14522715 on Build GRCh38
rs1035792302 Chr.Y: 14522673-14522673 on Build GRCh38
rs908677638 Chr.Y: 14522976-14522976 on Build GRCh38

View Full Product Details