Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 14733273-14733672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLGN4Y
Entrez Gene ID
22829
Gene Name
neuroligin 4, Y-linked
Gene Aliases
-
UniGene
Hs.439199
Gene Chromosome Location
Chr.Y: 14522578-14844945 on Build GRCh38
Transcripts
NM_001164238.1 NM_001206850.1 NM_014893.4 NR_028319.1 NR_046355.1 XM_006724874.2 XM_011531424.2 XM_011531425.2 XM_011531426.2 XM_011531427.1 XM_011531428.1 XM_011531429.2 XM_011531430.2 XM_017030034.1 XM_017030035.1 XM_017030036.1 XM_017030037.1 XM_017030038.1 XM_017030039.1 XM_017030040.1 XM_017030041.1

SNP Detail

SNP ID Chromosome Location
rs775645548 Chr.Y: 14733529-14733529 on Build GRCh38
rs776557550 Chr.Y: 14733527-14733528 on Build GRCh38
rs776833545 Chr.Y: 14733528-14733528 on Build GRCh38
rs1056438717 Chr.Y: 14733649-14733649 on Build GRCh38
rs926859623 Chr.Y: 14733543-14733543 on Build GRCh38
rs113775683 Chr.Y: 14733359-14733359 on Build GRCh38
rs775834801 Chr.Y: 14733451-14733451 on Build GRCh38
rs770025800 Chr.Y: 14733494-14733494 on Build GRCh38
rs56375424 Chr.Y: 14733618-14733618 on Build GRCh38
rs759565848 Chr.Y: 14733650-14733650 on Build GRCh38
rs747154396 Chr.Y: 14733549-14733549 on Build GRCh38
rs763325931 Chr.Y: 14733428-14733428 on Build GRCh38

View Full Product Details