Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 14793550-14794055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLGN4Y
Entrez Gene ID
22829
Gene Name
neuroligin 4, Y-linked
Gene Aliases
-
UniGene
Hs.439199
Gene Chromosome Location
Chr.Y: 14522578-14844945 on Build GRCh38
Transcripts
NM_001206850.1 NM_014893.4 NR_028319.1 NR_046355.1 XM_006724874.2 XM_011531424.2 XM_011531425.2 XM_011531426.2 XM_011531427.1 XM_011531428.1 XM_011531429.2 XM_011531430.2 XM_017030034.1 XM_017030035.1 XM_017030036.1 XM_017030037.1 XM_017030038.1 XM_017030039.1 XM_017030040.1 NR_046504.1

Gene Symbol
NLGN4Y-AS1
Entrez Gene ID
100874056
Gene Name
NLGN4Y antisense RNA 1
Gene Aliases
NLGN4Y-AS, NLGN4YAS
UniGene
Hs.743197
Gene Chromosome Location
Chr.Y: 14793642-14804033 on Build GRCh38
Transcripts
NM_001206850.1 NM_014893.4 NR_028319.1 NR_046355.1 XM_006724874.2 XM_011531424.2 XM_011531425.2 XM_011531426.2 XM_011531427.1 XM_011531428.1 XM_011531429.2 XM_011531430.2 XM_017030034.1 XM_017030035.1 XM_017030036.1 XM_017030037.1 XM_017030038.1 XM_017030039.1 XM_017030040.1 NR_046504.1

SNP Detail

SNP ID Chromosome Location
rs200776528 Chr.Y: 14793802-14793802 on Build GRCh38
rs772371153 Chr.Y: 14793741-14793741 on Build GRCh38
rs539097730 Chr.Y: 14793681-14793681 on Build GRCh38

View Full Product Details