Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63328281-63328785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDSR
Entrez Gene ID
2531
Gene Name
3-ketodihydrosphingosine reductase
Gene Aliases
DHSR, FVT1, SDR35C1
UniGene
Hs.74050
Gene Chromosome Location
Chr.18: 63327736-63367273 on Build GRCh38
Transcripts
NM_002035.2 XM_005266677.2 XM_017025690.1

SNP Detail

SNP ID Chromosome Location
rs550594132 Chr.18: 63328644-63328644 on Build GRCh38
rs939752657 Chr.18: 63328402-63328402 on Build GRCh38
rs79761558 Chr.18: 63328370-63328370 on Build GRCh38
rs964686642 Chr.18: 63328652-63328652 on Build GRCh38
rs928564664 Chr.18: 63328571-63328571 on Build GRCh38
rs896323033 Chr.18: 63328391-63328391 on Build GRCh38
rs940380533 Chr.18: 63328509-63328509 on Build GRCh38
rs368851860 Chr.18: 63328414-63328414 on Build GRCh38
rs754726556 Chr.18: 63328500-63328500 on Build GRCh38
rs988888653 Chr.18: 63328503-63328503 on Build GRCh38
rs1027595537 Chr.18: 63328691-63328691 on Build GRCh38
rs530983440 Chr.18: 63328629-63328629 on Build GRCh38
rs557603523 Chr.18: 63328515-63328515 on Build GRCh38
rs75987306 Chr.18: 63328369-63328369 on Build GRCh38
rs528524717 Chr.18: 63328424-63328424 on Build GRCh38
rs1036763063 Chr.18: 63328650-63328650 on Build GRCh38
rs1019840335 Chr.18: 63328646-63328646 on Build GRCh38
rs926523659 Chr.18: 63328566-63328566 on Build GRCh38
rs78620491 Chr.18: 63328371-63328371 on Build GRCh38
rs926556485 Chr.18: 63328354-63328354 on Build GRCh38
rs1019889684 Chr.18: 63328421-63328421 on Build GRCh38
rs572776978 Chr.18: 63328726-63328729 on Build GRCh38
rs573449301 Chr.18: 63328616-63328616 on Build GRCh38
rs551466548 Chr.18: 63328489-63328489 on Build GRCh38
rs372218813 Chr.18: 63328428-63328428 on Build GRCh38
rs144569535 Chr.18: 63328592-63328592 on Build GRCh38
rs374604103 Chr.18: 63328355-63328355 on Build GRCh38
rs1020326118 Chr.18: 63328347-63328347 on Build GRCh38
rs1045595974 Chr.18: 63328546-63328546 on Build GRCh38
rs956419242 Chr.18: 63328467-63328467 on Build GRCh38
rs930979446 Chr.18: 63328728-63328728 on Build GRCh38
rs368551129 Chr.18: 63328516-63328516 on Build GRCh38
rs398033183 Chr.18: 63328370-63328370 on Build GRCh38
rs1037200131 Chr.18: 63328411-63328411 on Build GRCh38
rs979324948 Chr.18: 63328346-63328346 on Build GRCh38
rs967602685 Chr.18: 63328389-63328389 on Build GRCh38
rs536950461 Chr.18: 63328561-63328561 on Build GRCh38
rs1049346553 Chr.18: 63328735-63328735 on Build GRCh38
rs951924505 Chr.18: 63328488-63328488 on Build GRCh38
rs796125693 Chr.18: 63328423-63328423 on Build GRCh38
rs188183839 Chr.18: 63328656-63328656 on Build GRCh38
rs1056349794 Chr.18: 63328384-63328384 on Build GRCh38
rs566424834 Chr.18: 63328325-63328325 on Build GRCh38
rs890077951 Chr.18: 63328431-63328431 on Build GRCh38
rs1019709376 Chr.18: 63328456-63328456 on Build GRCh38
rs11349419 Chr.18: 63328356-63328356 on Build GRCh38
rs893204914 Chr.18: 63328603-63328603 on Build GRCh38
rs2551407 Chr.18: 63328505-63328505 on Build GRCh38
rs980132172 Chr.18: 63328563-63328563 on Build GRCh38
rs751412795 Chr.18: 63328455-63328455 on Build GRCh38
rs937185681 Chr.18: 63328581-63328581 on Build GRCh38
rs372609863 Chr.18: 63328614-63328614 on Build GRCh38
rs1028516547 Chr.18: 63328429-63328429 on Build GRCh38
rs921701605 Chr.18: 63328443-63328443 on Build GRCh38
rs185315969 Chr.18: 63328492-63328492 on Build GRCh38
rs1008238367 Chr.18: 63328645-63328645 on Build GRCh38
rs1041841743 Chr.18: 63328639-63328639 on Build GRCh38
rs766204707 Chr.18: 63328447-63328447 on Build GRCh38

View Full Product Details