Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 123216560-123217047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRDN
Entrez Gene ID
10345
Gene Name
triadin
Gene Aliases
CPVT5, TDN, TRISK
UniGene
Hs.144744
Gene Chromosome Location
Chr.6: 123216339-123637093 on Build GRCh38
Transcripts
NM_006073.3

SNP Detail

SNP ID Chromosome Location
rs909738306 Chr.6: 123216911-123216911 on Build GRCh38
rs1000096105 Chr.6: 123216624-123216624 on Build GRCh38
rs575898010 Chr.6: 123216714-123216714 on Build GRCh38
rs997333346 Chr.6: 123216859-123216859 on Build GRCh38
rs754557914 Chr.6: 123216641-123216641 on Build GRCh38
rs1045968877 Chr.6: 123216654-123216654 on Build GRCh38
rs901968154 Chr.6: 123216843-123216843 on Build GRCh38
rs886061024 Chr.6: 123216598-123216598 on Build GRCh38
rs573243898 Chr.6: 123216672-123216672 on Build GRCh38
rs984324883 Chr.6: 123216740-123216740 on Build GRCh38
rs987856914 Chr.6: 123216970-123216970 on Build GRCh38
rs564360695 Chr.6: 123216756-123216756 on Build GRCh38
rs753394140 Chr.6: 123216626-123216626 on Build GRCh38
rs771705742 Chr.6: 123216809-123216809 on Build GRCh38
rs935071018 Chr.6: 123216742-123216742 on Build GRCh38
rs202088134 Chr.6: 123216723-123216724 on Build GRCh38
rs953299377 Chr.6: 123216716-123216716 on Build GRCh38
rs979108177 Chr.6: 123216727-123216727 on Build GRCh38
rs886061026 Chr.6: 123216735-123216735 on Build GRCh38
rs937621608 Chr.6: 123216822-123216822 on Build GRCh38
rs950262900 Chr.6: 123216646-123216646 on Build GRCh38
rs933654454 Chr.6: 123216956-123216956 on Build GRCh38
rs956581520 Chr.6: 123216955-123216955 on Build GRCh38
rs138251788 Chr.6: 123216886-123216886 on Build GRCh38
rs886061025 Chr.6: 123216655-123216655 on Build GRCh38
rs560234682 Chr.6: 123216870-123216870 on Build GRCh38
rs975947903 Chr.6: 123216713-123216713 on Build GRCh38
rs772911322 Chr.6: 123216826-123216826 on Build GRCh38
rs141561616 Chr.6: 123216771-123216771 on Build GRCh38
rs887841296 Chr.6: 123216674-123216674 on Build GRCh38
rs148371661 Chr.6: 123216698-123216698 on Build GRCh38
rs951524858 Chr.6: 123216897-123216897 on Build GRCh38
rs913365681 Chr.6: 123216769-123216769 on Build GRCh38
rs192301634 Chr.6: 123216733-123216733 on Build GRCh38
rs927529794 Chr.6: 123216814-123216814 on Build GRCh38
rs1049035485 Chr.6: 123216825-123216825 on Build GRCh38
rs150406434 Chr.6: 123216818-123216818 on Build GRCh38
rs181917537 Chr.6: 123216627-123216627 on Build GRCh38
rs1001485745 Chr.6: 123216667-123216667 on Build GRCh38
rs1023563669 Chr.6: 123216683-123216683 on Build GRCh38
rs796776192 Chr.6: 123216912-123216912 on Build GRCh38
rs1036847699 Chr.6: 123216837-123216837 on Build GRCh38
rs1001209019 Chr.6: 123216909-123216909 on Build GRCh38
rs187387580 Chr.6: 123216635-123216635 on Build GRCh38
rs199894760 Chr.6: 123216724-123216724 on Build GRCh38
rs766194808 Chr.6: 123216616-123216616 on Build GRCh38
rs62418707 Chr.6: 123216631-123216631 on Build GRCh38
rs981560467 Chr.6: 123216990-123216990 on Build GRCh38
rs74448738 Chr.6: 123216890-123216890 on Build GRCh38
rs867143824 Chr.6: 123216946-123216946 on Build GRCh38
rs906156714 Chr.6: 123216660-123216660 on Build GRCh38
rs1039632116 Chr.6: 123216757-123216757 on Build GRCh38
rs1005397278 Chr.6: 123216679-123216679 on Build GRCh38
rs1004938202 Chr.6: 123216835-123216835 on Build GRCh38
rs970473510 Chr.6: 123216985-123216985 on Build GRCh38
rs551675124 Chr.6: 123216977-123216977 on Build GRCh38
rs7743126 Chr.6: 123216692-123216692 on Build GRCh38
rs143914184 Chr.6: 123216929-123216929 on Build GRCh38
rs1003488658 Chr.6: 123216889-123216889 on Build GRCh38

View Full Product Details