Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42685060-42685551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP1
Entrez Gene ID
8506
Gene Name
contactin associated protein 1
Gene Aliases
CASPR, CNTNAP, NRXN4, P190
UniGene
Hs.408730
Gene Chromosome Location
Chr.17: 42679963-42699993 on Build GRCh38
Transcripts
NM_003632.2 XM_005257748.4 XM_017025238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4753495 c.478G>A p.G160S Chr.17: 42685105-42685105 on Build GRCh38
3819656 c.533A>G p.D178G Chr.17: 42685238-42685238 on Build GRCh38
5825252 c.652G>A p.A218T Chr.17: 42685357-42685357 on Build GRCh38
417511 c.660C>T p.G220G Chr.17: 42685365-42685365 on Build GRCh38
1679840 c.589G>C p.D197H Chr.17: 42685294-42685294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768387220 Chr.17: 42685347-42685347 on Build GRCh38
rs758476843 Chr.17: 42685203-42685203 on Build GRCh38
rs954192105 Chr.17: 42685242-42685242 on Build GRCh38
rs971683099 Chr.17: 42685441-42685441 on Build GRCh38
rs948394739 Chr.17: 42685449-42685449 on Build GRCh38
rs938868566 Chr.17: 42685141-42685141 on Build GRCh38
rs566767746 Chr.17: 42685220-42685220 on Build GRCh38
rs753597178 Chr.17: 42685290-42685291 on Build GRCh38
rs750249811 Chr.17: 42685186-42685186 on Build GRCh38
rs960306831 Chr.17: 42685477-42685477 on Build GRCh38
rs751453812 Chr.17: 42685107-42685107 on Build GRCh38
rs60104774 Chr.17: 42685501-42685501 on Build GRCh38
rs778544132 Chr.17: 42685461-42685461 on Build GRCh38
rs771483542 Chr.17: 42685152-42685152 on Build GRCh38
rs754704710 Chr.17: 42685377-42685377 on Build GRCh38
rs781415725 Chr.17: 42685440-42685440 on Build GRCh38
rs768847403 Chr.17: 42685457-42685457 on Build GRCh38
rs779018831 Chr.17: 42685192-42685192 on Build GRCh38
rs770121100 Chr.17: 42685335-42685335 on Build GRCh38
rs760660292 Chr.17: 42685138-42685139 on Build GRCh38
rs1052794932 Chr.17: 42685336-42685336 on Build GRCh38
rs374560939 Chr.17: 42685134-42685134 on Build GRCh38
rs144554326 Chr.17: 42685269-42685269 on Build GRCh38
rs114045118 Chr.17: 42685513-42685513 on Build GRCh38
rs773203084 Chr.17: 42685380-42685380 on Build GRCh38
rs757882183 Chr.17: 42685079-42685079 on Build GRCh38
rs547038534 Chr.17: 42685390-42685390 on Build GRCh38
rs938630556 Chr.17: 42685099-42685099 on Build GRCh38
rs368581393 Chr.17: 42685371-42685371 on Build GRCh38
rs752342751 Chr.17: 42685166-42685166 on Build GRCh38
rs1028565981 Chr.17: 42685240-42685240 on Build GRCh38
rs777899535 Chr.17: 42685204-42685204 on Build GRCh38
rs762381959 Chr.17: 42685179-42685179 on Build GRCh38
rs747797746 Chr.17: 42685463-42685463 on Build GRCh38
rs780385946 Chr.17: 42685455-42685455 on Build GRCh38
rs748337819 Chr.17: 42685198-42685198 on Build GRCh38
rs763570245 Chr.17: 42685236-42685236 on Build GRCh38
rs568282289 Chr.17: 42685523-42685524 on Build GRCh38
rs777750878 Chr.17: 42685300-42685300 on Build GRCh38
rs762662790 Chr.17: 42685228-42685228 on Build GRCh38
rs767466032 Chr.17: 42685267-42685267 on Build GRCh38
rs373521234 Chr.17: 42685450-42685450 on Build GRCh38
rs757416273 Chr.17: 42685438-42685438 on Build GRCh38
rs745943872 Chr.17: 42685085-42685085 on Build GRCh38
rs372590028 Chr.17: 42685181-42685181 on Build GRCh38
rs371727880 Chr.17: 42685436-42685436 on Build GRCh38
rs777110060 Chr.17: 42685156-42685156 on Build GRCh38
rs746309332 Chr.17: 42685333-42685333 on Build GRCh38
rs551276110 Chr.17: 42685130-42685130 on Build GRCh38
rs775909719 Chr.17: 42685339-42685339 on Build GRCh38
rs561659603 Chr.17: 42685302-42685302 on Build GRCh38
rs775008173 Chr.17: 42685225-42685225 on Build GRCh38
rs546838489 Chr.17: 42685464-42685465 on Build GRCh38
rs767341222 Chr.17: 42685183-42685183 on Build GRCh38
rs771530268 Chr.17: 42685211-42685211 on Build GRCh38
rs781627902 Chr.17: 42685287-42685287 on Build GRCh38
rs761624632 Chr.17: 42685449-42685450 on Build GRCh38
rs753641708 Chr.17: 42685398-42685398 on Build GRCh38
rs779157700 Chr.17: 42685128-42685128 on Build GRCh38
rs751029282 Chr.17: 42685447-42685447 on Build GRCh38
rs749562770 Chr.17: 42685122-42685122 on Build GRCh38
rs768836175 Chr.17: 42685125-42685125 on Build GRCh38
rs992994009 Chr.17: 42685206-42685206 on Build GRCh38
rs199605282 Chr.17: 42685432-42685432 on Build GRCh38
rs752924771 Chr.17: 42685282-42685282 on Build GRCh38
rs780961124 Chr.17: 42685119-42685119 on Build GRCh38
rs761328167 Chr.17: 42685366-42685366 on Build GRCh38
rs777593216 Chr.17: 42685090-42685090 on Build GRCh38
rs529351386 Chr.17: 42685460-42685460 on Build GRCh38
rs755910689 Chr.17: 42685275-42685275 on Build GRCh38
rs770136038 Chr.17: 42685224-42685224 on Build GRCh38
rs972299600 Chr.17: 42685132-42685132 on Build GRCh38
rs948775492 Chr.17: 42685229-42685229 on Build GRCh38
rs748941568 Chr.17: 42685345-42685345 on Build GRCh38
rs188166956 Chr.17: 42685182-42685182 on Build GRCh38
rs761273010 Chr.17: 42685251-42685251 on Build GRCh38
rs759297244 Chr.17: 42685163-42685163 on Build GRCh38
rs747572457 Chr.17: 42685205-42685205 on Build GRCh38
rs778521443 Chr.17: 42685428-42685428 on Build GRCh38
rs140590172 Chr.17: 42685239-42685239 on Build GRCh38
rs749687032 Chr.17: 42685456-42685456 on Build GRCh38
rs371406732 Chr.17: 42685372-42685372 on Build GRCh38
rs764826957 Chr.17: 42685165-42685165 on Build GRCh38
rs759207925 Chr.17: 42685401-42685401 on Build GRCh38
rs755855645 Chr.17: 42685189-42685189 on Build GRCh38
rs763999034 Chr.17: 42685188-42685189 on Build GRCh38
rs563501193 Chr.17: 42685429-42685429 on Build GRCh38
rs892358714 Chr.17: 42685348-42685348 on Build GRCh38
rs1052851077 Chr.17: 42685516-42685516 on Build GRCh38
rs761264256 Chr.17: 42685147-42685147 on Build GRCh38
rs773599082 Chr.17: 42685145-42685145 on Build GRCh38
rs781767249 Chr.17: 42685311-42685311 on Build GRCh38
rs984099759 Chr.17: 42685263-42685263 on Build GRCh38
rs760650248 Chr.17: 42685382-42685382 on Build GRCh38
rs757050594 Chr.17: 42685114-42685114 on Build GRCh38
rs766195304 Chr.17: 42685276-42685276 on Build GRCh38

View Full Product Details