Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28775666-28776202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS9
Entrez Gene ID
55756
Gene Name
integrator complex subunit 9
Gene Aliases
CPSF2L, INT9, RC74
UniGene
Hs.162397
Gene Chromosome Location
Chr.8: 28767658-28890226 on Build GRCh38
Transcripts
NM_001145159.2 NM_001172562.1 NM_018250.3 NR_026826.2 XM_006716356.2 XM_006716357.3 XM_011544573.1 XM_011544574.1 XM_011544575.2 XM_011544576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5744349 c.1421A>T p.Q474L Chr.8: 28775901-28775901 on Build GRCh38
422159 c.1417G>A p.E473K Chr.8: 28775905-28775905 on Build GRCh38
5409383 c.1534C>T p.R512W Chr.8: 28775788-28775788 on Build GRCh38
4592910 c.1511T>G p.V504G Chr.8: 28775811-28775811 on Build GRCh38
85110 c.1561G>A p.E521K Chr.8: 28775761-28775761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760564353 Chr.8: 28775742-28775742 on Build GRCh38
rs911620982 Chr.8: 28775999-28775999 on Build GRCh38
rs148560235 Chr.8: 28775837-28775837 on Build GRCh38
rs1016234055 Chr.8: 28775700-28775700 on Build GRCh38
rs993238521 Chr.8: 28775716-28775716 on Build GRCh38
rs775094295 Chr.8: 28775732-28775732 on Build GRCh38
rs894629758 Chr.8: 28776067-28776067 on Build GRCh38
rs774670609 Chr.8: 28775798-28775798 on Build GRCh38
rs762550139 Chr.8: 28775873-28775873 on Build GRCh38
rs548180024 Chr.8: 28775823-28775823 on Build GRCh38
rs766078408 Chr.8: 28775767-28775767 on Build GRCh38
rs954153367 Chr.8: 28776018-28776019 on Build GRCh38
rs754398392 Chr.8: 28775918-28775918 on Build GRCh38
rs778992467 Chr.8: 28775843-28775843 on Build GRCh38
rs540547131 Chr.8: 28776049-28776049 on Build GRCh38
rs373683663 Chr.8: 28775793-28775793 on Build GRCh38
rs142907634 Chr.8: 28775870-28775870 on Build GRCh38
rs78086231 Chr.8: 28775783-28775783 on Build GRCh38
rs879215328 Chr.8: 28775998-28775998 on Build GRCh38
rs770415705 Chr.8: 28775875-28775875 on Build GRCh38
rs1051505820 Chr.8: 28775993-28775993 on Build GRCh38
rs748146699 Chr.8: 28775711-28775711 on Build GRCh38
rs781005594 Chr.8: 28775852-28775852 on Build GRCh38
rs767097653 Chr.8: 28775895-28775895 on Build GRCh38
rs772800096 Chr.8: 28775807-28775807 on Build GRCh38
rs547979899 Chr.8: 28776136-28776136 on Build GRCh38
rs777789213 Chr.8: 28775790-28775790 on Build GRCh38
rs773737752 Chr.8: 28775876-28775876 on Build GRCh38
rs562153395 Chr.8: 28776029-28776029 on Build GRCh38
rs1000325657 Chr.8: 28775867-28775867 on Build GRCh38
rs747921642 Chr.8: 28775970-28775970 on Build GRCh38
rs753034116 Chr.8: 28775914-28775914 on Build GRCh38
rs369630069 Chr.8: 28775893-28775893 on Build GRCh38
rs534558238 Chr.8: 28775842-28775842 on Build GRCh38
rs754306495 Chr.8: 28775882-28775883 on Build GRCh38
rs764578456 Chr.8: 28775917-28775917 on Build GRCh38
rs974007241 Chr.8: 28775718-28775718 on Build GRCh38
rs973684724 Chr.8: 28775987-28775987 on Build GRCh38
rs776701034 Chr.8: 28775751-28775751 on Build GRCh38
rs926930603 Chr.8: 28776096-28776096 on Build GRCh38
rs755361005 Chr.8: 28775909-28775910 on Build GRCh38
rs917252640 Chr.8: 28775736-28775736 on Build GRCh38
rs963251614 Chr.8: 28775986-28775986 on Build GRCh38
rs763903311 Chr.8: 28775745-28775745 on Build GRCh38
rs779231471 Chr.8: 28775931-28775931 on Build GRCh38
rs904784155 Chr.8: 28776028-28776028 on Build GRCh38
rs1004219213 Chr.8: 28776078-28776078 on Build GRCh38
rs749446245 Chr.8: 28775729-28775729 on Build GRCh38
rs201254959 Chr.8: 28775946-28775946 on Build GRCh38
rs200675183 Chr.8: 28775801-28775801 on Build GRCh38
rs765917957 Chr.8: 28775820-28775820 on Build GRCh38
rs531180984 Chr.8: 28775713-28775713 on Build GRCh38
rs776307091 Chr.8: 28775802-28775802 on Build GRCh38
rs1010471162 Chr.8: 28775833-28775833 on Build GRCh38
rs73669441 Chr.8: 28775695-28775695 on Build GRCh38
rs372201911 Chr.8: 28775806-28775806 on Build GRCh38
rs747979330 Chr.8: 28775864-28775864 on Build GRCh38
rs11540279 Chr.8: 28775886-28775886 on Build GRCh38
rs753091252 Chr.8: 28775829-28775829 on Build GRCh38
rs761064546 Chr.8: 28775915-28775915 on Build GRCh38
rs758085228 Chr.8: 28775765-28775765 on Build GRCh38
rs760907819 Chr.8: 28776163-28776163 on Build GRCh38
rs199679008 Chr.8: 28775942-28775942 on Build GRCh38
rs756564539 Chr.8: 28775836-28775836 on Build GRCh38
rs181870094 Chr.8: 28775717-28775717 on Build GRCh38
rs562632463 Chr.8: 28776095-28776095 on Build GRCh38
rs139056710 Chr.8: 28776069-28776069 on Build GRCh38
rs896752822 Chr.8: 28775694-28775694 on Build GRCh38
rs749961902 Chr.8: 28775761-28775761 on Build GRCh38
rs761397539 Chr.8: 28775754-28775754 on Build GRCh38
rs241202 Chr.8: 28776168-28776168 on Build GRCh38
rs553124923 Chr.8: 28775988-28775988 on Build GRCh38
rs912632710 Chr.8: 28776010-28776010 on Build GRCh38
rs572345853 Chr.8: 28776048-28776048 on Build GRCh38
rs945731532 Chr.8: 28776002-28776002 on Build GRCh38
rs767853863 Chr.8: 28775826-28775826 on Build GRCh38
rs771173147 Chr.8: 28775730-28775730 on Build GRCh38
rs368126287 Chr.8: 28775788-28775788 on Build GRCh38
rs532420269 Chr.8: 28776071-28776071 on Build GRCh38
rs532323381 Chr.8: 28775787-28775787 on Build GRCh38
rs769779753 Chr.8: 28775974-28775974 on Build GRCh38
rs531477331 Chr.8: 28776101-28776101 on Build GRCh38
rs751579461 Chr.8: 28775911-28775911 on Build GRCh38
rs762602475 Chr.8: 28775811-28775811 on Build GRCh38
rs372066824 Chr.8: 28775951-28775951 on Build GRCh38
rs755411492 Chr.8: 28775774-28775774 on Build GRCh38
rs941856671 Chr.8: 28775784-28775784 on Build GRCh38
rs750823393 Chr.8: 28775944-28775944 on Build GRCh38
rs982756388 Chr.8: 28775940-28775940 on Build GRCh38
rs1026941057 Chr.8: 28776015-28776015 on Build GRCh38
rs549719222 Chr.8: 28775773-28775773 on Build GRCh38
rs746047822 Chr.8: 28775846-28775846 on Build GRCh38
rs746773071 Chr.8: 28775800-28775800 on Build GRCh38
rs190888209 Chr.8: 28775990-28775990 on Build GRCh38
rs1056399587 Chr.8: 28776051-28776051 on Build GRCh38
rs1056923046 Chr.8: 28776106-28776106 on Build GRCh38
rs781748312 Chr.8: 28775708-28775708 on Build GRCh38
rs916948359 Chr.8: 28776107-28776107 on Build GRCh38
rs1020092032 Chr.8: 28776036-28776036 on Build GRCh38
rs552403467 Chr.8: 28776077-28776077 on Build GRCh38
rs757720202 Chr.8: 28775925-28775925 on Build GRCh38
rs764785865 Chr.8: 28775756-28775756 on Build GRCh38
rs199718736 Chr.8: 28775791-28775791 on Build GRCh38
rs890116563 Chr.8: 28775822-28775822 on Build GRCh38
rs879607359 Chr.8: 28776125-28776125 on Build GRCh38
rs146865172 Chr.8: 28775824-28775824 on Build GRCh38
rs780617509 Chr.8: 28775836-28775837 on Build GRCh38
rs541917974 Chr.8: 28776014-28776014 on Build GRCh38

View Full Product Details