Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28777737-28778169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS9
Entrez Gene ID
55756
Gene Name
integrator complex subunit 9
Gene Aliases
CPSF2L, INT9, RC74
UniGene
Hs.162397
Gene Chromosome Location
Chr.8: 28767658-28890226 on Build GRCh38
Transcripts
NM_001145159.2 NM_001172562.1 NM_018250.3 NR_026826.2 XM_006716356.2 XM_006716357.3 XM_011544573.1 XM_011544574.1 XM_011544575.2 XM_011544576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3834542 c.1346A>G p.D449G Chr.8: 28777878-28777878 on Build GRCh38
6312250 c.1338C>A p.C446* Chr.8: 28777886-28777886 on Build GRCh38
5874528 c.1349C>T p.T450I Chr.8: 28777875-28777875 on Build GRCh38
1210792 c.1309C>T p.Q437* Chr.8: 28777915-28777915 on Build GRCh38
1699924 c.1283C>T p.S428F Chr.8: 28777941-28777941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201166588 Chr.8: 28778003-28778003 on Build GRCh38
rs755142950 Chr.8: 28778116-28778116 on Build GRCh38
rs774992643 Chr.8: 28777943-28777943 on Build GRCh38
rs751890112 Chr.8: 28777873-28777873 on Build GRCh38
rs576258809 Chr.8: 28778001-28778001 on Build GRCh38
rs984579607 Chr.8: 28778049-28778050 on Build GRCh38
rs971775350 Chr.8: 28778039-28778042 on Build GRCh38
rs761011392 Chr.8: 28777823-28777823 on Build GRCh38
rs373909305 Chr.8: 28777829-28777829 on Build GRCh38
rs943274150 Chr.8: 28778078-28778078 on Build GRCh38
rs970411013 Chr.8: 28778142-28778142 on Build GRCh38
rs780420351 Chr.8: 28777872-28777872 on Build GRCh38
rs768332790 Chr.8: 28777962-28777962 on Build GRCh38
rs1036803099 Chr.8: 28777838-28777838 on Build GRCh38
rs151230593 Chr.8: 28778028-28778028 on Build GRCh38
rs771720047 Chr.8: 28777800-28777800 on Build GRCh38
rs903370078 Chr.8: 28777859-28777859 on Build GRCh38
rs750494854 Chr.8: 28777836-28777836 on Build GRCh38
rs777669891 Chr.8: 28777894-28777894 on Build GRCh38
rs757196607 Chr.8: 28777903-28777903 on Build GRCh38
rs771537343 Chr.8: 28777917-28777917 on Build GRCh38
rs745376105 Chr.8: 28777911-28777911 on Build GRCh38
rs563821485 Chr.8: 28778088-28778088 on Build GRCh38
rs981565056 Chr.8: 28778043-28778043 on Build GRCh38
rs919294067 Chr.8: 28778044-28778047 on Build GRCh38
rs779067666 Chr.8: 28778105-28778105 on Build GRCh38
rs374309788 Chr.8: 28777985-28777985 on Build GRCh38
rs775186973 Chr.8: 28777813-28777813 on Build GRCh38
rs749058173 Chr.8: 28777898-28777898 on Build GRCh38
rs116093564 Chr.8: 28777799-28777799 on Build GRCh38
rs751718846 Chr.8: 28778004-28778004 on Build GRCh38
rs762138428 Chr.8: 28777978-28777978 on Build GRCh38
rs73669443 Chr.8: 28778045-28778045 on Build GRCh38
rs746600632 Chr.8: 28777946-28777946 on Build GRCh38
rs764386717 Chr.8: 28777828-28777828 on Build GRCh38
rs773660176 Chr.8: 28777996-28777996 on Build GRCh38
rs1033270433 Chr.8: 28777965-28777965 on Build GRCh38
rs377388058 Chr.8: 28777868-28777868 on Build GRCh38
rs1007135083 Chr.8: 28778011-28778011 on Build GRCh38
rs1026884100 Chr.8: 28778106-28778106 on Build GRCh38
rs776935359 Chr.8: 28777964-28777964 on Build GRCh38
rs909174597 Chr.8: 28778063-28778063 on Build GRCh38
rs748405451 Chr.8: 28777838-28777840 on Build GRCh38
rs999619619 Chr.8: 28778051-28778051 on Build GRCh38
rs749169284 Chr.8: 28777785-28777785 on Build GRCh38
rs755213777 Chr.8: 28778069-28778069 on Build GRCh38
rs370956795 Chr.8: 28778087-28778087 on Build GRCh38
rs772330863 Chr.8: 28777939-28777941 on Build GRCh38
rs773861199 Chr.8: 28777796-28777796 on Build GRCh38
rs755200544 Chr.8: 28778005-28778005 on Build GRCh38
rs545172202 Chr.8: 28777780-28777780 on Build GRCh38
rs1039141367 Chr.8: 28778081-28778081 on Build GRCh38
rs11785319 Chr.8: 28777786-28777786 on Build GRCh38
rs777841731 Chr.8: 28777781-28777781 on Build GRCh38
rs755253516 Chr.8: 28777891-28777891 on Build GRCh38
rs368967611 Chr.8: 28777910-28777910 on Build GRCh38
rs200033074 Chr.8: 28777833-28777833 on Build GRCh38
rs765689623 Chr.8: 28777834-28777834 on Build GRCh38
rs535697202 Chr.8: 28778023-28778023 on Build GRCh38

View Full Product Details