Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101020618-101021146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT2B
Entrez Gene ID
79979
Gene Name
tRNA methyltransferase 2 homolog B
Gene Aliases
CXorf34, dJ341D10.3
UniGene
Hs.496501
Gene Chromosome Location
Chr.X: 100973358-101052118 on Build GRCh38
Transcripts
NM_001167970.1 NM_001167971.1 NM_001167972.1 NM_024917.5 XM_005262195.2 XM_005262196.2 XM_005262201.2 XM_006724705.2 XM_011531042.2 XM_011531044.2 XM_017029858.1 XM_017029859.1 XM_017029860.1 XM_017029861.1 XM_017029862.1 XM_017029863.1 XM_017029864.1 XM_017029865.1 XM_017029866.1 XM_017029867.1 XM_017029868.1 XM_017029869.1 XM_017029870.1 XM_017029871.1 XM_017029872.1 XM_017029873.1 XM_017029874.1 XM_017029875.1

SNP Detail

SNP ID Chromosome Location
rs1042420160 Chr.X: 101020957-101020957 on Build GRCh38
rs954426093 Chr.X: 101020710-101020710 on Build GRCh38
rs879000362 Chr.X: 101020713-101020713 on Build GRCh38
rs200657512 Chr.X: 101020642-101020645 on Build GRCh38
rs1031449939 Chr.X: 101020672-101020675 on Build GRCh38
rs773295316 Chr.X: 101021113-101021113 on Build GRCh38
rs1025181195 Chr.X: 101020741-101020741 on Build GRCh38
rs900435988 Chr.X: 101020701-101020701 on Build GRCh38
rs750405575 Chr.X: 101021054-101021054 on Build GRCh38
rs200061526 Chr.X: 101021072-101021072 on Build GRCh38
rs766164493 Chr.X: 101021109-101021109 on Build GRCh38
rs148532931 Chr.X: 101020989-101020989 on Build GRCh38
rs757360034 Chr.X: 101021114-101021114 on Build GRCh38
rs10527015 Chr.X: 101020650-101020650 on Build GRCh38
rs902601311 Chr.X: 101021028-101021028 on Build GRCh38
rs750324568 Chr.X: 101021082-101021082 on Build GRCh38
rs1000883093 Chr.X: 101020684-101020684 on Build GRCh38
rs983239121 Chr.X: 101020712-101020712 on Build GRCh38
rs990932862 Chr.X: 101020834-101020834 on Build GRCh38
rs1030198751 Chr.X: 101020708-101020708 on Build GRCh38
rs940341413 Chr.X: 101021006-101021006 on Build GRCh38
rs780105208 Chr.X: 101021067-101021067 on Build GRCh38
rs969576572 Chr.X: 101020762-101020762 on Build GRCh38
rs112699426 Chr.X: 101020650-101020650 on Build GRCh38
rs201998689 Chr.X: 101021093-101021093 on Build GRCh38
rs112210510 Chr.X: 101020653-101020653 on Build GRCh38
rs1032635586 Chr.X: 101020699-101020699 on Build GRCh38
rs984543278 Chr.X: 101020827-101020827 on Build GRCh38
rs1014589720 Chr.X: 101020724-101020724 on Build GRCh38
rs973094379 Chr.X: 101020763-101020763 on Build GRCh38
rs1053805426 Chr.X: 101021044-101021044 on Build GRCh38
rs144706265 Chr.X: 101020939-101020939 on Build GRCh38
rs938287127 Chr.X: 101020819-101020819 on Build GRCh38
rs202158383 Chr.X: 101020647-101020649 on Build GRCh38
rs980938397 Chr.X: 101020789-101020789 on Build GRCh38
rs200642952 Chr.X: 101021051-101021051 on Build GRCh38
rs963338758 Chr.X: 101020737-101020737 on Build GRCh38
rs996384887 Chr.X: 101020703-101020703 on Build GRCh38
rs936646358 Chr.X: 101021038-101021038 on Build GRCh38
rs918127581 Chr.X: 101020923-101020923 on Build GRCh38
rs184765846 Chr.X: 101020818-101020818 on Build GRCh38
rs181238058 Chr.X: 101021117-101021117 on Build GRCh38
rs748333501 Chr.X: 101021092-101021092 on Build GRCh38
rs756166696 Chr.X: 101021064-101021064 on Build GRCh38
rs188718384 Chr.X: 101020990-101020990 on Build GRCh38

View Full Product Details