Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33204235-33204737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B8
Entrez Gene ID
7923
Gene Name
hydroxysteroid 17-beta dehydrogenase 8
Gene Aliases
D6S2245E, FABG, FABGL, H2-KE6, HKE6, KE6, RING2, SDR30C1, dJ1033B10.9
UniGene
Hs.415058
Gene Chromosome Location
Chr.6: 33204637-33206831 on Build GRCh38
Transcripts
NM_014234.4 NM_001077516.1 NM_001288777.1 NM_006979.2

Gene Symbol
SLC39A7
Entrez Gene ID
7922
Gene Name
solute carrier family 39 member 7
Gene Aliases
D6S115E, D6S2244E, H2-KE4, HKE4, KE4, RING5, ZIP7
UniGene
Hs.631995
Gene Chromosome Location
Chr.6: 33200826-33204437 on Build GRCh38
Transcripts
NM_014234.4 NM_001077516.1 NM_001288777.1 NM_006979.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7130804 c.10C>G p.Q4E Chr.6: 33204678-33204678 on Build GRCh38
1621508 c.51A>G p.T17T Chr.6: 33204719-33204719 on Build GRCh38
6351132 c.34G>T p.A12S Chr.6: 33204702-33204702 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs995763147 Chr.6: 33204458-33204458 on Build GRCh38
rs182388247 Chr.6: 33204302-33204302 on Build GRCh38
rs765641312 Chr.6: 33204709-33204709 on Build GRCh38
rs958488817 Chr.6: 33204420-33204420 on Build GRCh38
rs1048782 Chr.6: 33204325-33204325 on Build GRCh38
rs890689189 Chr.6: 33204503-33204503 on Build GRCh38
rs759984516 Chr.6: 33204705-33204705 on Build GRCh38
rs547035463 Chr.6: 33204609-33204609 on Build GRCh38
rs558676405 Chr.6: 33204664-33204664 on Build GRCh38
rs1036429353 Chr.6: 33204528-33204528 on Build GRCh38
rs946168237 Chr.6: 33204309-33204309 on Build GRCh38
rs553333148 Chr.6: 33204434-33204434 on Build GRCh38
rs529663035 Chr.6: 33204320-33204320 on Build GRCh38
rs759026575 Chr.6: 33204689-33204694 on Build GRCh38
rs754302023 Chr.6: 33204667-33204667 on Build GRCh38
rs190065313 Chr.6: 33204473-33204473 on Build GRCh38
rs570962078 Chr.6: 33204472-33204472 on Build GRCh38
rs759296612 Chr.6: 33204648-33204648 on Build GRCh38
rs186100646 Chr.6: 33204407-33204407 on Build GRCh38
rs769773247 Chr.6: 33204654-33204654 on Build GRCh38
rs932120773 Chr.6: 33204492-33204492 on Build GRCh38
rs574075966 Chr.6: 33204697-33204697 on Build GRCh38
rs963200127 Chr.6: 33204494-33204494 on Build GRCh38
rs1029164459 Chr.6: 33204345-33204346 on Build GRCh38
rs376172570 Chr.6: 33204657-33204657 on Build GRCh38
rs1027488145 Chr.6: 33204408-33204408 on Build GRCh38
rs749196565 Chr.6: 33204686-33204686 on Build GRCh38
rs554888772 Chr.6: 33204556-33204556 on Build GRCh38
rs770521610 Chr.6: 33204680-33204696 on Build GRCh38
rs370208017 Chr.6: 33204499-33204500 on Build GRCh38
rs992830841 Chr.6: 33204298-33204298 on Build GRCh38
rs114699053 Chr.6: 33204529-33204529 on Build GRCh38
rs973625508 Chr.6: 33204261-33204261 on Build GRCh38
rs1008954258 Chr.6: 33204364-33204364 on Build GRCh38
rs1008591978 Chr.6: 33204509-33204509 on Build GRCh38
rs973302325 Chr.6: 33204254-33204254 on Build GRCh38
rs776370698 Chr.6: 33204646-33204646 on Build GRCh38
rs937322555 Chr.6: 33204281-33204281 on Build GRCh38
rs949331613 Chr.6: 33204301-33204301 on Build GRCh38
rs1034199255 Chr.6: 33204703-33204703 on Build GRCh38
rs142093440 Chr.6: 33204669-33204669 on Build GRCh38
rs772470105 Chr.6: 33204637-33204637 on Build GRCh38
rs768158059 Chr.6: 33204682-33204682 on Build GRCh38
rs369419148 Chr.6: 33204341-33204341 on Build GRCh38
rs1034228632 Chr.6: 33204524-33204524 on Build GRCh38
rs552781884 Chr.6: 33204465-33204465 on Build GRCh38
rs772071122 Chr.6: 33204690-33204690 on Build GRCh38
rs769508877 Chr.6: 33204680-33204680 on Build GRCh38
rs773380800 Chr.6: 33204691-33204691 on Build GRCh38
rs144584311 Chr.6: 33204273-33204273 on Build GRCh38
rs1030011633 Chr.6: 33204459-33204459 on Build GRCh38
rs372381285 Chr.6: 33204495-33204496 on Build GRCh38
rs935406852 Chr.6: 33204466-33204466 on Build GRCh38
rs767575152 Chr.6: 33204660-33204660 on Build GRCh38
rs749744272 Chr.6: 33204371-33204371 on Build GRCh38
rs958591184 Chr.6: 33204708-33204708 on Build GRCh38
rs376596225 Chr.6: 33204321-33204321 on Build GRCh38
rs926068012 Chr.6: 33204652-33204652 on Build GRCh38
rs140577933 Chr.6: 33204461-33204461 on Build GRCh38
rs890131901 Chr.6: 33204345-33204345 on Build GRCh38
rs370135457 Chr.6: 33204580-33204580 on Build GRCh38
rs552817372 Chr.6: 33204670-33204670 on Build GRCh38
rs557942898 Chr.6: 33204621-33204622 on Build GRCh38
rs201389008 Chr.6: 33204636-33204636 on Build GRCh38
rs371077739 Chr.6: 33204672-33204672 on Build GRCh38
rs147878855 Chr.6: 33204292-33204292 on Build GRCh38
rs145971755 Chr.6: 33204675-33204675 on Build GRCh38
rs912179628 Chr.6: 33204715-33204715 on Build GRCh38
rs766764231 Chr.6: 33204700-33204700 on Build GRCh38
rs917142867 Chr.6: 33204435-33204435 on Build GRCh38
rs902575405 Chr.6: 33204303-33204303 on Build GRCh38
rs373903352 Chr.6: 33204673-33204673 on Build GRCh38
rs546797834 Chr.6: 33204493-33204493 on Build GRCh38
rs994999284 Chr.6: 33204544-33204544 on Build GRCh38
rs150495100 Chr.6: 33204665-33204665 on Build GRCh38
rs991413150 Chr.6: 33204424-33204424 on Build GRCh38
rs776306585 Chr.6: 33204681-33204688 on Build GRCh38
rs916588591 Chr.6: 33204300-33204300 on Build GRCh38
rs746618614 Chr.6: 33204634-33204634 on Build GRCh38
rs962087406 Chr.6: 33204383-33204383 on Build GRCh38
rs761801597 Chr.6: 33204659-33204659 on Build GRCh38
rs138620921 Chr.6: 33204702-33204702 on Build GRCh38
rs946064697 Chr.6: 33204610-33204610 on Build GRCh38
rs372709810 Chr.6: 33204497-33204497 on Build GRCh38
rs765562999 Chr.6: 33204315-33204315 on Build GRCh38
rs961940972 Chr.6: 33204467-33204491 on Build GRCh38
rs992801004 Chr.6: 33204574-33204574 on Build GRCh38
rs780803866 Chr.6: 33204677-33204677 on Build GRCh38

View Full Product Details