Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 4111429-4112023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRM1
Entrez Gene ID
6240
Gene Name
ribonucleotide reductase catalytic subunit M1
Gene Aliases
R1, RIR1, RR1
UniGene
Hs.445705
Gene Chromosome Location
Chr.11: 4094685-4138925 on Build GRCh38
Transcripts
NM_001033.4 NM_001318064.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
466794 c.541G>A p.E181K Chr.11: 4111953-4111953 on Build GRCh38
6132337 c.501A>T p.P167P Chr.11: 4111913-4111913 on Build GRCh38
1289554 c.522A>T p.V174V Chr.11: 4111934-4111934 on Build GRCh38
6769859 c.549T>C p.I183I Chr.11: 4111961-4111961 on Build GRCh38
4032730 c.606G>A p.S202S Chr.11: 4112018-4112018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376581575 Chr.11: 4111815-4111815 on Build GRCh38
rs968439874 Chr.11: 4111820-4111820 on Build GRCh38
rs946856244 Chr.11: 4111737-4111737 on Build GRCh38
rs768752773 Chr.11: 4111961-4111961 on Build GRCh38
rs889449370 Chr.11: 4111520-4111527 on Build GRCh38
rs756725882 Chr.11: 4111580-4111582 on Build GRCh38
rs572316509 Chr.11: 4111775-4111775 on Build GRCh38
rs554441706 Chr.11: 4111809-4111809 on Build GRCh38
rs750119723 Chr.11: 4111562-4111562 on Build GRCh38
rs775832822 Chr.11: 4111665-4111665 on Build GRCh38
rs12808005 Chr.11: 4111948-4111948 on Build GRCh38
rs377479513 Chr.11: 4111632-4111632 on Build GRCh38
rs1052531119 Chr.11: 4111714-4111714 on Build GRCh38
rs969954465 Chr.11: 4111841-4111841 on Build GRCh38
rs551658215 Chr.11: 4111459-4111459 on Build GRCh38
rs991475623 Chr.11: 4111794-4111794 on Build GRCh38
rs137969593 Chr.11: 4111628-4111628 on Build GRCh38
rs750230648 Chr.11: 4111679-4111679 on Build GRCh38
rs372857715 Chr.11: 4111655-4111655 on Build GRCh38
rs753459541 Chr.11: 4111599-4111599 on Build GRCh38
rs889741042 Chr.11: 4111980-4111980 on Build GRCh38
rs778240323 Chr.11: 4111875-4111875 on Build GRCh38
rs774226307 Chr.11: 4111967-4111967 on Build GRCh38
rs771046548 Chr.11: 4111870-4111870 on Build GRCh38
rs578011277 Chr.11: 4111865-4111865 on Build GRCh38
rs377260439 Chr.11: 4111550-4111550 on Build GRCh38
rs772004423 Chr.11: 4111659-4111662 on Build GRCh38
rs562668206 Chr.11: 4111864-4111864 on Build GRCh38
rs774366453 Chr.11: 4111666-4111666 on Build GRCh38
rs774332452 Chr.11: 4111973-4111973 on Build GRCh38
rs113417540 Chr.11: 4111758-4111758 on Build GRCh38
rs780565423 Chr.11: 4111588-4111589 on Build GRCh38
rs770179085 Chr.11: 4111506-4111506 on Build GRCh38
rs771350104 Chr.11: 4111638-4111638 on Build GRCh38
rs1029750584 Chr.11: 4111764-4111764 on Build GRCh38
rs927996479 Chr.11: 4111856-4111856 on Build GRCh38
rs72555777 Chr.11: 4111805-4111805 on Build GRCh38
rs939432239 Chr.11: 4111675-4111675 on Build GRCh38
rs761680203 Chr.11: 4111554-4111554 on Build GRCh38
rs961142649 Chr.11: 4111640-4111640 on Build GRCh38
rs575532940 Chr.11: 4111507-4111508 on Build GRCh38
rs751431074 Chr.11: 4111531-4111531 on Build GRCh38
rs973894429 Chr.11: 4111816-4111816 on Build GRCh38
rs1044027378 Chr.11: 4111744-4111744 on Build GRCh38
rs768944195 Chr.11: 4111882-4111884 on Build GRCh38
rs780268388 Chr.11: 4111955-4111955 on Build GRCh38
rs746278014 Chr.11: 4111934-4111934 on Build GRCh38
rs200388706 Chr.11: 4111649-4111649 on Build GRCh38
rs763024799 Chr.11: 4111656-4111656 on Build GRCh38
rs1004403537 Chr.11: 4111791-4111791 on Build GRCh38
rs997073409 Chr.11: 4111760-4111760 on Build GRCh38
rs548971473 Chr.11: 4111582-4111582 on Build GRCh38
rs752140471 Chr.11: 4111877-4111877 on Build GRCh38
rs560861499 Chr.11: 4111502-4111502 on Build GRCh38
rs72555778 Chr.11: 4111840-4111840 on Build GRCh38
rs922430980 Chr.11: 4111896-4111896 on Build GRCh38
rs982519715 Chr.11: 4111541-4111541 on Build GRCh38
rs933880095 Chr.11: 4111902-4111902 on Build GRCh38
rs988661143 Chr.11: 4111916-4111916 on Build GRCh38
rs781374016 Chr.11: 4111644-4111644 on Build GRCh38
rs538849390 Chr.11: 4111729-4111729 on Build GRCh38
rs142136268 Chr.11: 4111982-4111982 on Build GRCh38
rs768662668 Chr.11: 4111660-4111660 on Build GRCh38
rs757746611 Chr.11: 4111887-4111887 on Build GRCh38
rs781708974 Chr.11: 4111932-4111932 on Build GRCh38
rs796673767 Chr.11: 4111749-4111749 on Build GRCh38
rs749460951 Chr.11: 4111956-4111956 on Build GRCh38
rs12789346 Chr.11: 4112001-4112001 on Build GRCh38
rs747076924 Chr.11: 4111842-4111842 on Build GRCh38
rs72555776 Chr.11: 4111736-4111736 on Build GRCh38
rs12807843 Chr.11: 4111913-4111913 on Build GRCh38
rs375890929 Chr.11: 4111849-4111849 on Build GRCh38
rs760529956 Chr.11: 4111686-4111686 on Build GRCh38
rs373213442 Chr.11: 4111563-4111563 on Build GRCh38
rs199510349 Chr.11: 4111668-4111668 on Build GRCh38
rs201413736 Chr.11: 4111949-4111949 on Build GRCh38
rs771956712 Chr.11: 4111999-4111999 on Build GRCh38
rs559690595 Chr.11: 4111537-4111537 on Build GRCh38
rs746212853 Chr.11: 4111474-4111474 on Build GRCh38
rs754535019 Chr.11: 4111603-4111603 on Build GRCh38
rs900104999 Chr.11: 4111755-4111755 on Build GRCh38
rs370037707 Chr.11: 4111551-4111551 on Build GRCh38

View Full Product Details