Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38767283-38767870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR5
Entrez Gene ID
79913
Gene Name
ARP5 actin-related protein 5 homolog
Gene Aliases
Arp5, INO80M
UniGene
Hs.371585
Gene Chromosome Location
Chr.20: 38748454-38772446 on Build GRCh38
Transcripts
NM_024855.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1181746 c.1552C>T p.R518W Chr.20: 38767582-38767582 on Build GRCh38
4468597 c.1551C>T p.F517F Chr.20: 38767581-38767581 on Build GRCh38
5744044 c.1493C>T p.T498M Chr.20: 38767523-38767523 on Build GRCh38
5857845 c.1501C>T p.P501S Chr.20: 38767531-38767531 on Build GRCh38
4569171 c.1562T>A p.F521Y Chr.20: 38767592-38767592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200364249 Chr.20: 38767501-38767501 on Build GRCh38
rs753017125 Chr.20: 38767560-38767560 on Build GRCh38
rs367992039 Chr.20: 38767515-38767515 on Build GRCh38
rs768091640 Chr.20: 38767602-38767603 on Build GRCh38
rs757635595 Chr.20: 38767581-38767581 on Build GRCh38
rs1049604731 Chr.20: 38767566-38767566 on Build GRCh38
rs867803649 Chr.20: 38767336-38767336 on Build GRCh38
rs766443901 Chr.20: 38767610-38767610 on Build GRCh38
rs754240396 Chr.20: 38767580-38767580 on Build GRCh38
rs779765563 Chr.20: 38767516-38767516 on Build GRCh38
rs17853827 Chr.20: 38767524-38767524 on Build GRCh38
rs894486301 Chr.20: 38767470-38767470 on Build GRCh38
rs922251720 Chr.20: 38767806-38767806 on Build GRCh38
rs767451036 Chr.20: 38767558-38767558 on Build GRCh38
rs372916844 Chr.20: 38767499-38767499 on Build GRCh38
rs760468689 Chr.20: 38767447-38767447 on Build GRCh38
rs2245231 Chr.20: 38767477-38767477 on Build GRCh38
rs878966993 Chr.20: 38767353-38767353 on Build GRCh38
rs1015281023 Chr.20: 38767333-38767333 on Build GRCh38
rs779942725 Chr.20: 38767496-38767496 on Build GRCh38
rs374188956 Chr.20: 38767614-38767614 on Build GRCh38
rs190638142 Chr.20: 38767368-38767368 on Build GRCh38
rs964958346 Chr.20: 38767766-38767766 on Build GRCh38
rs779564620 Chr.20: 38767582-38767582 on Build GRCh38
rs778351710 Chr.20: 38767575-38767575 on Build GRCh38
rs144462138 Chr.20: 38767474-38767474 on Build GRCh38
rs6071382 Chr.20: 38767460-38767460 on Build GRCh38
rs763786522 Chr.20: 38767459-38767459 on Build GRCh38
rs769678801 Chr.20: 38767535-38767535 on Build GRCh38
rs757723812 Chr.20: 38767640-38767640 on Build GRCh38
rs1039488763 Chr.20: 38767317-38767317 on Build GRCh38
rs774003741 Chr.20: 38767526-38767526 on Build GRCh38
rs1024256280 Chr.20: 38767686-38767686 on Build GRCh38
rs1031725450 Chr.20: 38767698-38767698 on Build GRCh38
rs774372395 Chr.20: 38767616-38767616 on Build GRCh38
rs556643366 Chr.20: 38767634-38767634 on Build GRCh38
rs879087874 Chr.20: 38767337-38767337 on Build GRCh38
rs754956369 Chr.20: 38767490-38767490 on Build GRCh38
rs111586858 Chr.20: 38767471-38767471 on Build GRCh38
rs2245237 Chr.20: 38767619-38767619 on Build GRCh38
rs933849116 Chr.20: 38767338-38767338 on Build GRCh38
rs761024281 Chr.20: 38767599-38767599 on Build GRCh38
rs756261891 Chr.20: 38767561-38767561 on Build GRCh38
rs1046929984 Chr.20: 38767397-38767397 on Build GRCh38
rs752826585 Chr.20: 38767495-38767495 on Build GRCh38
rs751432705 Chr.20: 38767550-38767550 on Build GRCh38
rs756527007 Chr.20: 38767500-38767500 on Build GRCh38
rs941179887 Chr.20: 38767625-38767625 on Build GRCh38
rs894356840 Chr.20: 38767642-38767642 on Build GRCh38
rs202044751 Chr.20: 38767531-38767531 on Build GRCh38
rs769424936 Chr.20: 38767603-38767603 on Build GRCh38
rs776085976 Chr.20: 38767464-38767464 on Build GRCh38
rs769748322 Chr.20: 38767466-38767466 on Build GRCh38
rs554240655 Chr.20: 38767609-38767609 on Build GRCh38
rs767677643 Chr.20: 38767621-38767621 on Build GRCh38
rs1005870051 Chr.20: 38767403-38767403 on Build GRCh38
rs201033138 Chr.20: 38767598-38767598 on Build GRCh38
rs749767793 Chr.20: 38767426-38767426 on Build GRCh38
rs975100412 Chr.20: 38767800-38767800 on Build GRCh38
rs907319305 Chr.20: 38767691-38767691 on Build GRCh38
rs1027769176 Chr.20: 38767392-38767392 on Build GRCh38
rs554132782 Chr.20: 38767538-38767538 on Build GRCh38
rs773803985 Chr.20: 38767417-38767417 on Build GRCh38
rs771761055 Chr.20: 38767436-38767436 on Build GRCh38
rs567037300 Chr.20: 38767594-38767594 on Build GRCh38
rs998881144 Chr.20: 38767697-38767697 on Build GRCh38
rs748830603 Chr.20: 38767598-38767601 on Build GRCh38
rs545647840 Chr.20: 38767414-38767414 on Build GRCh38
rs781481528 Chr.20: 38767492-38767492 on Build GRCh38
rs1016823734 Chr.20: 38767405-38767405 on Build GRCh38
rs991618542 Chr.20: 38767420-38767420 on Build GRCh38
rs752980824 Chr.20: 38767656-38767656 on Build GRCh38
rs957788323 Chr.20: 38767711-38767711 on Build GRCh38
rs765874064 Chr.20: 38767725-38767725 on Build GRCh38
rs34493530 Chr.20: 38767497-38767497 on Build GRCh38
rs990293615 Chr.20: 38767737-38767737 on Build GRCh38
rs775111101 Chr.20: 38767443-38767443 on Build GRCh38
rs375416188 Chr.20: 38767623-38767623 on Build GRCh38
rs1018310490 Chr.20: 38767762-38767762 on Build GRCh38
rs372519020 Chr.20: 38767555-38767555 on Build GRCh38
rs376703857 Chr.20: 38767696-38767696 on Build GRCh38
rs545586560 Chr.20: 38767805-38767805 on Build GRCh38
rs200120169 Chr.20: 38767633-38767633 on Build GRCh38
rs761668229 Chr.20: 38767468-38767468 on Build GRCh38
rs191027138 Chr.20: 38767843-38767843 on Build GRCh38
rs576579523 Chr.20: 38767801-38767801 on Build GRCh38
rs79240975 Chr.20: 38767337-38767337 on Build GRCh38
rs369269280 Chr.20: 38767641-38767641 on Build GRCh38
rs1026650712 Chr.20: 38767764-38767764 on Build GRCh38
rs933711849 Chr.20: 38767584-38767584 on Build GRCh38
rs758307673 Chr.20: 38767476-38767476 on Build GRCh38
rs576638282 Chr.20: 38767398-38767398 on Build GRCh38
rs772957767 Chr.20: 38767537-38767537 on Build GRCh38
rs187329040 Chr.20: 38767583-38767583 on Build GRCh38
rs533004644 Chr.20: 38767822-38767822 on Build GRCh38
rs746538809 Chr.20: 38767517-38767517 on Build GRCh38

View Full Product Details