Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 504604-505172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MADCAM1
Entrez Gene ID
8174
Gene Name
mucosal vascular addressin cell adhesion molecule 1
Gene Aliases
MACAM1
UniGene
Hs.102598
Gene Chromosome Location
Chr.19: 496490-505343 on Build GRCh38
Transcripts
NM_130760.2 NM_130762.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1742548 c.941C>T p.A314V Chr.19: 504757-504757 on Build GRCh38
1711547 c.1118G>A p.G373E Chr.19: 504934-504934 on Build GRCh38
227142 c.1108G>T p.G370W Chr.19: 504924-504924 on Build GRCh38
6229414 c.930G>A p.S310S Chr.19: 504746-504746 on Build GRCh38
6214951 c.1049A>C p.E350A Chr.19: 504865-504865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777482249 Chr.19: 504859-504860 on Build GRCh38
rs751707863 Chr.19: 504967-504967 on Build GRCh38
rs558308475 Chr.19: 505064-505064 on Build GRCh38
rs765389123 Chr.19: 504947-504947 on Build GRCh38
rs1050210570 Chr.19: 504636-504636 on Build GRCh38
rs373464736 Chr.19: 504847-504847 on Build GRCh38
rs147245939 Chr.19: 504869-504869 on Build GRCh38
rs756437236 Chr.19: 504813-504813 on Build GRCh38
rs374750697 Chr.19: 504962-504962 on Build GRCh38
rs535905706 Chr.19: 504719-504719 on Build GRCh38
rs935886675 Chr.19: 505125-505125 on Build GRCh38
rs775180241 Chr.19: 504794-504794 on Build GRCh38
rs755021998 Chr.19: 504758-504758 on Build GRCh38
rs777139660 Chr.19: 505016-505016 on Build GRCh38
rs759037630 Chr.19: 504788-504788 on Build GRCh38
rs184776902 Chr.19: 505110-505110 on Build GRCh38
rs370718340 Chr.19: 504712-504712 on Build GRCh38
rs1140823 Chr.19: 505071-505071 on Build GRCh38
rs754764277 Chr.19: 504787-504787 on Build GRCh38
rs774273915 Chr.19: 504705-504705 on Build GRCh38
rs776308081 Chr.19: 504849-504849 on Build GRCh38
rs944615867 Chr.19: 504659-504659 on Build GRCh38
rs143314648 Chr.19: 504914-504914 on Build GRCh38
rs773726132 Chr.19: 504929-504929 on Build GRCh38
rs920294257 Chr.19: 504867-504867 on Build GRCh38
rs953324217 Chr.19: 505102-505102 on Build GRCh38
rs532346585 Chr.19: 504939-504939 on Build GRCh38
rs986165807 Chr.19: 505115-505115 on Build GRCh38
rs375438687 Chr.19: 504648-504648 on Build GRCh38
rs566426814 Chr.19: 504735-504735 on Build GRCh38
rs529852754 Chr.19: 504974-504974 on Build GRCh38
rs776857920 Chr.19: 504943-504943 on Build GRCh38
rs755007127 Chr.19: 504823-504823 on Build GRCh38
rs145604202 Chr.19: 504757-504757 on Build GRCh38
rs1048430651 Chr.19: 505020-505020 on Build GRCh38
rs139350636 Chr.19: 504938-504938 on Build GRCh38
rs113593149 Chr.19: 504710-504710 on Build GRCh38
rs995743575 Chr.19: 505005-505006 on Build GRCh38
rs7246543 Chr.19: 504830-504830 on Build GRCh38
rs748049978 Chr.19: 504773-504773 on Build GRCh38
rs750794834 Chr.19: 504799-504799 on Build GRCh38
rs565184057 Chr.19: 504921-504921 on Build GRCh38
rs149092655 Chr.19: 504796-504796 on Build GRCh38
rs779800333 Chr.19: 504980-504980 on Build GRCh38
rs550472975 Chr.19: 504945-504945 on Build GRCh38
rs911742859 Chr.19: 504644-504644 on Build GRCh38
rs140417103 Chr.19: 504746-504746 on Build GRCh38
rs1015707551 Chr.19: 505046-505046 on Build GRCh38
rs771428543 Chr.19: 505152-505152 on Build GRCh38
rs374757993 Chr.19: 504727-504727 on Build GRCh38
rs778276933 Chr.19: 504997-504997 on Build GRCh38
rs935939711 Chr.19: 504944-504944 on Build GRCh38
rs550411289 Chr.19: 504694-504694 on Build GRCh38
rs762979960 Chr.19: 504961-504961 on Build GRCh38
rs768393049 Chr.19: 504994-504994 on Build GRCh38
rs1046243717 Chr.19: 504966-504966 on Build GRCh38
rs768298980 Chr.19: 504925-504925 on Build GRCh38
rs201733673 Chr.19: 504880-504880 on Build GRCh38
rs778945114 Chr.19: 504824-504824 on Build GRCh38
rs771280019 Chr.19: 505010-505010 on Build GRCh38
rs538493090 Chr.19: 505080-505080 on Build GRCh38
rs758064459 Chr.19: 504829-504830 on Build GRCh38
rs557375745 Chr.19: 504711-504711 on Build GRCh38
rs772150232 Chr.19: 504733-504733 on Build GRCh38
rs372736157 Chr.19: 504790-504790 on Build GRCh38
rs368846611 Chr.19: 504968-504968 on Build GRCh38
rs371067068 Chr.19: 505015-505015 on Build GRCh38
rs748977050 Chr.19: 504984-504984 on Build GRCh38
rs192760680 Chr.19: 505030-505030 on Build GRCh38
rs759745925 Chr.19: 504889-504889 on Build GRCh38
rs199656590 Chr.19: 504857-504857 on Build GRCh38
rs773603021 Chr.19: 504863-504863 on Build GRCh38
rs923239995 Chr.19: 504864-504864 on Build GRCh38
rs767963308 Chr.19: 504740-504740 on Build GRCh38
rs147543851 Chr.19: 504920-504920 on Build GRCh38
rs968394390 Chr.19: 505147-505147 on Build GRCh38
rs764638463 Chr.19: 504729-504729 on Build GRCh38
rs141825568 Chr.19: 504853-504853 on Build GRCh38
rs138718378 Chr.19: 504797-504797 on Build GRCh38
rs766563350 Chr.19: 504816-504816 on Build GRCh38
rs766007939 Chr.19: 504720-504720 on Build GRCh38
rs1004296185 Chr.19: 505037-505037 on Build GRCh38
rs752710973 Chr.19: 504909-504909 on Build GRCh38
rs770550961 Chr.19: 504776-504776 on Build GRCh38
rs777718695 Chr.19: 504911-504911 on Build GRCh38
rs1021238673 Chr.19: 505142-505142 on Build GRCh38
rs780204137 Chr.19: 504755-504755 on Build GRCh38
rs768630518 Chr.19: 504701-504701 on Build GRCh38
rs139304491 Chr.19: 504870-504870 on Build GRCh38
rs757260089 Chr.19: 504913-504913 on Build GRCh38
rs1041596815 Chr.19: 504933-504933 on Build GRCh38
rs748289153 Chr.19: 504825-504825 on Build GRCh38
rs371476209 Chr.19: 504924-504924 on Build GRCh38
rs150040042 Chr.19: 504948-504948 on Build GRCh38
rs199507986 Chr.19: 504745-504745 on Build GRCh38
rs1019113131 Chr.19: 505058-505058 on Build GRCh38
rs138020018 Chr.19: 504774-504774 on Build GRCh38
rs373642778 Chr.19: 504732-504732 on Build GRCh38
rs374689202 Chr.19: 504775-504775 on Build GRCh38
rs576214537 Chr.19: 505024-505024 on Build GRCh38
rs745478436 Chr.19: 504852-504852 on Build GRCh38
rs754025424 Chr.19: 504822-504822 on Build GRCh38
rs765512380 Chr.19: 504907-504907 on Build GRCh38
rs547863186 Chr.19: 504979-504979 on Build GRCh38
rs778769332 Chr.19: 504760-504760 on Build GRCh38
rs903412015 Chr.19: 504744-504744 on Build GRCh38
rs924499232 Chr.19: 505083-505083 on Build GRCh38
rs149388006 Chr.19: 504862-504862 on Build GRCh38

View Full Product Details