Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 156291526-156292116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNAB1
Entrez Gene ID
7881
Gene Name
potassium voltage-gated channel subfamily A member regulatory beta subunit 1
Gene Aliases
AKR6A3, KCNA1B, KV-BETA-1, Kvb1.3, hKvBeta3, hKvb3
UniGene
Hs.654519
Gene Chromosome Location
Chr.3: 156118211-156540603 on Build GRCh38
Transcripts
NM_001308217.1 NM_001308222.1 NM_003471.3 NM_172159.3 NM_172160.2 XM_017007171.1 XM_017007174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3005103 c.169C>T p.H57Y Chr.3: 156292076-156292076 on Build GRCh38
3005101 c.161C>G p.T54R Chr.3: 156292068-156292068 on Build GRCh38
5451707 c.102C>T p.N34N Chr.3: 156292009-156292009 on Build GRCh38
3005093 c.83C>A p.T28N Chr.3: 156291990-156291990 on Build GRCh38
6218948 c.141G>A p.A47A Chr.3: 156292048-156292048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918940708 Chr.3: 156291859-156291859 on Build GRCh38
rs369162310 Chr.3: 156291893-156291893 on Build GRCh38
rs921454119 Chr.3: 156291767-156291767 on Build GRCh38
rs1013443935 Chr.3: 156291843-156291843 on Build GRCh38
rs1019065679 Chr.3: 156291906-156291906 on Build GRCh38
rs768556987 Chr.3: 156292086-156292086 on Build GRCh38
rs888908547 Chr.3: 156291907-156291907 on Build GRCh38
rs79750297 Chr.3: 156291997-156291997 on Build GRCh38
rs1044224492 Chr.3: 156291851-156291851 on Build GRCh38
rs774757221 Chr.3: 156292080-156292080 on Build GRCh38
rs763909519 Chr.3: 156292005-156292005 on Build GRCh38
rs964851685 Chr.3: 156291764-156291764 on Build GRCh38
rs975604190 Chr.3: 156291765-156291765 on Build GRCh38
rs970112134 Chr.3: 156291816-156291816 on Build GRCh38
rs745583052 Chr.3: 156291892-156291892 on Build GRCh38
rs948862050 Chr.3: 156291574-156291574 on Build GRCh38
rs763268221 Chr.3: 156291994-156291994 on Build GRCh38
rs910173500 Chr.3: 156291580-156291580 on Build GRCh38
rs564452320 Chr.3: 156291616-156291616 on Build GRCh38
rs572439206 Chr.3: 156292018-156292018 on Build GRCh38
rs780403107 Chr.3: 156291939-156291939 on Build GRCh38
rs2290165 Chr.3: 156291825-156291825 on Build GRCh38
rs987136323 Chr.3: 156291745-156291745 on Build GRCh38
rs753866599 Chr.3: 156292006-156292006 on Build GRCh38
rs72558050 Chr.3: 156291796-156291796 on Build GRCh38
rs921518763 Chr.3: 156291564-156291564 on Build GRCh38
rs749470319 Chr.3: 156292070-156292070 on Build GRCh38
rs539860574 Chr.3: 156291810-156291810 on Build GRCh38
rs771307522 Chr.3: 156292076-156292076 on Build GRCh38
rs747459146 Chr.3: 156291950-156291950 on Build GRCh38
rs756986490 Chr.3: 156292075-156292076 on Build GRCh38
rs112100796 Chr.3: 156292072-156292072 on Build GRCh38
rs1012747447 Chr.3: 156291649-156291649 on Build GRCh38
rs1036058192 Chr.3: 156291638-156291638 on Build GRCh38
rs761220738 Chr.3: 156292008-156292008 on Build GRCh38
rs372404618 Chr.3: 156291972-156291972 on Build GRCh38
rs769974371 Chr.3: 156291991-156291991 on Build GRCh38
rs760368896 Chr.3: 156292082-156292082 on Build GRCh38
rs758060549 Chr.3: 156292028-156292028 on Build GRCh38
rs1031697649 Chr.3: 156291752-156291752 on Build GRCh38
rs769072570 Chr.3: 156291957-156291957 on Build GRCh38
rs868688359 Chr.3: 156291855-156291855 on Build GRCh38
rs1048361852 Chr.3: 156291578-156291578 on Build GRCh38
rs999168480 Chr.3: 156291744-156291744 on Build GRCh38
rs755419299 Chr.3: 156292032-156292032 on Build GRCh38
rs771932834 Chr.3: 156291996-156291996 on Build GRCh38
rs781742069 Chr.3: 156291973-156291973 on Build GRCh38
rs887136659 Chr.3: 156291591-156291591 on Build GRCh38
rs150626308 Chr.3: 156292009-156292009 on Build GRCh38
rs376778469 Chr.3: 156291651-156291651 on Build GRCh38
rs908792679 Chr.3: 156292036-156292036 on Build GRCh38
rs931550176 Chr.3: 156291779-156291779 on Build GRCh38
rs781537516 Chr.3: 156292033-156292033 on Build GRCh38
rs534966292 Chr.3: 156291880-156291880 on Build GRCh38
rs968659114 Chr.3: 156291742-156291742 on Build GRCh38
rs984286781 Chr.3: 156291819-156291819 on Build GRCh38
rs7620003 Chr.3: 156291720-156291720 on Build GRCh38
rs895900637 Chr.3: 156291831-156291831 on Build GRCh38
rs181223496 Chr.3: 156291674-156291674 on Build GRCh38
rs1004180953 Chr.3: 156291618-156291618 on Build GRCh38
rs138625130 Chr.3: 156292069-156292069 on Build GRCh38
rs552884415 Chr.3: 156292029-156292029 on Build GRCh38
rs759011239 Chr.3: 156291999-156291999 on Build GRCh38
rs773559627 Chr.3: 156291993-156291993 on Build GRCh38
rs151289713 Chr.3: 156291995-156291995 on Build GRCh38
rs752611476 Chr.3: 156291860-156291860 on Build GRCh38
rs16825964 Chr.3: 156291631-156291631 on Build GRCh38
rs895898449 Chr.3: 156291642-156291642 on Build GRCh38
rs940308850 Chr.3: 156291846-156291846 on Build GRCh38
rs944179785 Chr.3: 156291554-156291554 on Build GRCh38
rs551099358 Chr.3: 156291732-156291732 on Build GRCh38
rs1040282974 Chr.3: 156291563-156291563 on Build GRCh38
rs1031607313 Chr.3: 156291715-156291715 on Build GRCh38
rs931604346 Chr.3: 156291569-156291569 on Build GRCh38
rs748154435 Chr.3: 156291984-156291984 on Build GRCh38
rs368507650 Chr.3: 156291866-156291866 on Build GRCh38
rs1000142927 Chr.3: 156291664-156291664 on Build GRCh38
rs968646908 Chr.3: 156291656-156291656 on Build GRCh38
rs769692234 Chr.3: 156292068-156292068 on Build GRCh38
rs142304029 Chr.3: 156291814-156291814 on Build GRCh38
rs375183830 Chr.3: 156291865-156291865 on Build GRCh38
rs763608731 Chr.3: 156291871-156291871 on Build GRCh38
rs778411838 Chr.3: 156291891-156291891 on Build GRCh38
rs1048596765 Chr.3: 156291795-156291795 on Build GRCh38
rs748756556 Chr.3: 156292051-156292051 on Build GRCh38
rs762025688 Chr.3: 156292063-156292063 on Build GRCh38
rs776432870 Chr.3: 156292091-156292091 on Build GRCh38
rs911610810 Chr.3: 156291763-156291763 on Build GRCh38
rs756834632 Chr.3: 156291883-156291883 on Build GRCh38
rs1037663445 Chr.3: 156291852-156291852 on Build GRCh38

View Full Product Details