Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66591415-66592008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC87
Entrez Gene ID
55231
Gene Name
coiled-coil domain containing 87
Gene Aliases
-
UniGene
Hs.121072
Gene Chromosome Location
Chr.11: 66590169-66593083 on Build GRCh38
Transcripts
NM_018219.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4908225 c.1060G>A p.A354T Chr.11: 66591956-66591956 on Build GRCh38
3452134 c.1144C>T p.L382L Chr.11: 66591872-66591872 on Build GRCh38
4467660 c.1500C>T p.S500S Chr.11: 66591516-66591516 on Build GRCh38
3452132 c.1262C>T p.S421F Chr.11: 66591754-66591754 on Build GRCh38
6622526 c.1151T>C p.V384A Chr.11: 66591865-66591865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144092876 Chr.11: 66591648-66591648 on Build GRCh38
rs762173012 Chr.11: 66591766-66591767 on Build GRCh38
rs781347795 Chr.11: 66591603-66591603 on Build GRCh38
rs772116285 Chr.11: 66591501-66591501 on Build GRCh38
rs768785771 Chr.11: 66591696-66591696 on Build GRCh38
rs776515690 Chr.11: 66591704-66591704 on Build GRCh38
rs565595374 Chr.11: 66591505-66591505 on Build GRCh38
rs1056936915 Chr.11: 66591573-66591573 on Build GRCh38
rs758747499 Chr.11: 66591855-66591855 on Build GRCh38
rs750944700 Chr.11: 66591759-66591759 on Build GRCh38
rs770201652 Chr.11: 66591621-66591621 on Build GRCh38
rs778389835 Chr.11: 66591592-66591592 on Build GRCh38
rs758959276 Chr.11: 66591458-66591458 on Build GRCh38
rs143106957 Chr.11: 66591701-66591701 on Build GRCh38
rs142496256 Chr.11: 66591927-66591927 on Build GRCh38
rs913311758 Chr.11: 66591721-66591721 on Build GRCh38
rs754513838 Chr.11: 66591446-66591446 on Build GRCh38
rs776465296 Chr.11: 66591639-66591639 on Build GRCh38
rs200576384 Chr.11: 66591687-66591687 on Build GRCh38
rs752911174 Chr.11: 66591900-66591900 on Build GRCh38
rs759319157 Chr.11: 66591640-66591640 on Build GRCh38
rs750886174 Chr.11: 66591681-66591681 on Build GRCh38
rs748665254 Chr.11: 66591457-66591457 on Build GRCh38
rs138056551 Chr.11: 66591598-66591598 on Build GRCh38
rs755335832 Chr.11: 66591893-66591893 on Build GRCh38
rs371746457 Chr.11: 66591760-66591760 on Build GRCh38
rs749421239 Chr.11: 66591629-66591629 on Build GRCh38
rs780039959 Chr.11: 66591959-66591959 on Build GRCh38
rs377376167 Chr.11: 66591677-66591677 on Build GRCh38
rs780420434 Chr.11: 66591627-66591627 on Build GRCh38
rs761280214 Chr.11: 66591851-66591851 on Build GRCh38
rs780473418 Chr.11: 66591688-66591688 on Build GRCh38
rs778452891 Chr.11: 66591966-66591966 on Build GRCh38
rs771173697 Chr.11: 66591472-66591472 on Build GRCh38
rs763676422 Chr.11: 66591740-66591740 on Build GRCh38
rs775655299 Chr.11: 66591487-66591487 on Build GRCh38
rs766047380 Chr.11: 66591511-66591511 on Build GRCh38
rs139489472 Chr.11: 66591979-66591979 on Build GRCh38
rs111997006 Chr.11: 66591908-66591908 on Build GRCh38
rs757675570 Chr.11: 66591600-66591600 on Build GRCh38
rs779190045 Chr.11: 66591785-66591785 on Build GRCh38
rs977138145 Chr.11: 66591727-66591727 on Build GRCh38
rs887199177 Chr.11: 66591921-66591921 on Build GRCh38
rs773886806 Chr.11: 66591874-66591874 on Build GRCh38
rs373320692 Chr.11: 66591963-66591963 on Build GRCh38
rs759129000 Chr.11: 66591544-66591544 on Build GRCh38
rs762449518 Chr.11: 66591738-66591738 on Build GRCh38
rs778135888 Chr.11: 66591467-66591467 on Build GRCh38
rs534612416 Chr.11: 66591510-66591510 on Build GRCh38
rs1056976742 Chr.11: 66591816-66591816 on Build GRCh38
rs1051221128 Chr.11: 66591768-66591781 on Build GRCh38
rs905758342 Chr.11: 66591466-66591466 on Build GRCh38
rs975682611 Chr.11: 66591987-66591987 on Build GRCh38
rs898490727 Chr.11: 66591857-66591857 on Build GRCh38
rs770002210 Chr.11: 66591484-66591484 on Build GRCh38
rs748214053 Chr.11: 66591799-66591799 on Build GRCh38
rs374929199 Chr.11: 66591836-66591836 on Build GRCh38
rs773793192 Chr.11: 66591745-66591745 on Build GRCh38
rs532353345 Chr.11: 66591891-66591891 on Build GRCh38
rs148278836 Chr.11: 66591748-66591748 on Build GRCh38
rs146563586 Chr.11: 66591612-66591612 on Build GRCh38
rs141833535 Chr.11: 66591764-66591764 on Build GRCh38
rs927132168 Chr.11: 66591766-66591766 on Build GRCh38
rs771720516 Chr.11: 66591968-66591968 on Build GRCh38
rs768262949 Chr.11: 66591840-66591840 on Build GRCh38
rs775961114 Chr.11: 66591640-66591641 on Build GRCh38
rs879082571 Chr.11: 66591771-66591771 on Build GRCh38
rs367671433 Chr.11: 66591700-66591700 on Build GRCh38
rs753382789 Chr.11: 66591444-66591444 on Build GRCh38
rs777766804 Chr.11: 66591907-66591907 on Build GRCh38
rs370489699 Chr.11: 66591680-66591680 on Build GRCh38
rs753492468 Chr.11: 66591526-66591526 on Build GRCh38
rs375306181 Chr.11: 66591699-66591699 on Build GRCh38
rs375465158 Chr.11: 66591828-66591828 on Build GRCh38
rs766979684 Chr.11: 66591880-66591880 on Build GRCh38
rs374330821 Chr.11: 66591848-66591848 on Build GRCh38
rs746584823 Chr.11: 66591588-66591588 on Build GRCh38
rs768180205 Chr.11: 66591752-66591752 on Build GRCh38
rs749831512 Chr.11: 66591643-66591643 on Build GRCh38
rs767088192 Chr.11: 66591856-66591856 on Build GRCh38
rs376754440 Chr.11: 66591470-66591470 on Build GRCh38
rs749472686 Chr.11: 66591691-66591691 on Build GRCh38
rs766096691 Chr.11: 66591438-66591438 on Build GRCh38
rs199906600 Chr.11: 66591835-66591835 on Build GRCh38
rs1046013501 Chr.11: 66591445-66591445 on Build GRCh38
rs1008372278 Chr.11: 66591962-66591962 on Build GRCh38
rs774753466 Chr.11: 66591758-66591759 on Build GRCh38
rs755765185 Chr.11: 66591583-66591583 on Build GRCh38
rs765956457 Chr.11: 66591676-66591676 on Build GRCh38
rs141303580 Chr.11: 66591630-66591630 on Build GRCh38
rs137881283 Chr.11: 66591838-66591838 on Build GRCh38
rs765018973 Chr.11: 66591897-66591897 on Build GRCh38
rs747797538 Chr.11: 66591967-66591967 on Build GRCh38
rs768477307 Chr.11: 66591964-66591964 on Build GRCh38
rs1038303870 Chr.11: 66591819-66591819 on Build GRCh38
rs567481594 Chr.11: 66591614-66591614 on Build GRCh38
rs201117269 Chr.11: 66591777-66591777 on Build GRCh38
rs149088581 Chr.11: 66591611-66591611 on Build GRCh38
rs778325831 Chr.11: 66591450-66591450 on Build GRCh38
rs764907584 Chr.11: 66591657-66591657 on Build GRCh38
rs903016163 Chr.11: 66591547-66591547 on Build GRCh38
rs750141639 Chr.11: 66591574-66591574 on Build GRCh38
rs1041476035 Chr.11: 66591536-66591536 on Build GRCh38
rs947138341 Chr.11: 66591500-66591500 on Build GRCh38
rs769525293 Chr.11: 66591712-66591712 on Build GRCh38
rs769126762 Chr.11: 66591655-66591656 on Build GRCh38
rs747524308 Chr.11: 66591960-66591960 on Build GRCh38
rs778179008 Chr.11: 66591926-66591926 on Build GRCh38
rs761206869 Chr.11: 66591875-66591875 on Build GRCh38
rs372285398 Chr.11: 66591556-66591556 on Build GRCh38
rs775158538 Chr.11: 66591731-66591731 on Build GRCh38
rs770495671 Chr.11: 66591600-66591600 on Build GRCh38
rs556418445 Chr.11: 66591638-66591638 on Build GRCh38
rs150538178 Chr.11: 66591975-66591975 on Build GRCh38
rs776938671 Chr.11: 66591475-66591475 on Build GRCh38
rs80219769 Chr.11: 66591831-66591831 on Build GRCh38
rs745708026 Chr.11: 66591709-66591709 on Build GRCh38
rs746050902 Chr.11: 66591477-66591477 on Build GRCh38
rs777483736 Chr.11: 66591501-66591507 on Build GRCh38
rs369805771 Chr.11: 66591905-66591905 on Build GRCh38
rs762542551 Chr.11: 66591663-66591663 on Build GRCh38
rs760461232 Chr.11: 66591631-66591631 on Build GRCh38

View Full Product Details