Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49857915-49858172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTOV1
Entrez Gene ID
53635
Gene Name
prostate tumor overexpressed 1
Gene Aliases
ACID2, PTOV-1
UniGene
Hs.587979
Gene Chromosome Location
Chr.19: 49850726-49860744 on Build GRCh38
Transcripts
NM_001305105.1 NM_001305108.1 NM_017432.4 NR_130963.1 XM_005258998.4 XM_006723244.3 XM_011527033.1 XM_011527034.2 XM_011527035.1 XM_017026879.1 XM_017026880.1 NR_110730.1

Gene Symbol
PTOV1-AS2
Entrez Gene ID
101928378
Gene Name
PTOV1 antisense RNA 2
Gene Aliases
-
UniGene
Hs.710295
Gene Chromosome Location
Chr.19: 49856465-49859289 on Build GRCh38
Transcripts
NM_001305105.1 NM_001305108.1 NM_017432.4 NR_130963.1 XM_005258998.4 XM_006723244.3 XM_011527033.1 XM_011527034.2 XM_011527035.1 XM_017026879.1 XM_017026880.1 NR_110730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4654810 c.857A>T p.Y286F Chr.19: 49857956-49857956 on Build GRCh38
5856410 c.859G>A p.V287M Chr.19: 49857958-49857958 on Build GRCh38
5856411 c.763G>A p.V255M Chr.19: 49857958-49857958 on Build GRCh38
4285490 c.771G>A p.Q257Q Chr.19: 49857966-49857966 on Build GRCh38
5056845 c.812A>G p.Y271C Chr.19: 49858086-49858086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764310709 Chr.19: 49857988-49857988 on Build GRCh38
rs755010048 Chr.19: 49858122-49858122 on Build GRCh38
rs570399878 Chr.19: 49857993-49857993 on Build GRCh38
rs575957865 Chr.19: 49858064-49858064 on Build GRCh38
rs200657051 Chr.19: 49857986-49857986 on Build GRCh38
rs901674296 Chr.19: 49857952-49857952 on Build GRCh38
rs747481739 Chr.19: 49858144-49858144 on Build GRCh38
rs771524395 Chr.19: 49857964-49857964 on Build GRCh38
rs763418419 Chr.19: 49857961-49857961 on Build GRCh38
rs773657588 Chr.19: 49857958-49857958 on Build GRCh38
rs771181635 Chr.19: 49858033-49858033 on Build GRCh38
rs376581280 Chr.19: 49858120-49858120 on Build GRCh38
rs537423003 Chr.19: 49857994-49857994 on Build GRCh38
rs1011184824 Chr.19: 49857985-49857985 on Build GRCh38
rs555502907 Chr.19: 49858007-49858007 on Build GRCh38
rs746174573 Chr.19: 49858131-49858131 on Build GRCh38
rs146437714 Chr.19: 49858148-49858148 on Build GRCh38
rs200629879 Chr.19: 49857975-49857975 on Build GRCh38
rs772817837 Chr.19: 49858117-49858117 on Build GRCh38
rs779661768 Chr.19: 49857944-49857944 on Build GRCh38
rs775130692 Chr.19: 49858153-49858153 on Build GRCh38
rs745766571 Chr.19: 49858106-49858106 on Build GRCh38
rs751074444 Chr.19: 49858060-49858060 on Build GRCh38
rs371426666 Chr.19: 49857951-49857951 on Build GRCh38
rs375634367 Chr.19: 49858130-49858130 on Build GRCh38
rs12461139 Chr.19: 49858069-49858069 on Build GRCh38
rs770526938 Chr.19: 49858066-49858066 on Build GRCh38
rs749548797 Chr.19: 49857990-49857991 on Build GRCh38
rs772389280 Chr.19: 49858082-49858082 on Build GRCh38
rs771967042 Chr.19: 49858119-49858119 on Build GRCh38
rs755661713 Chr.19: 49858128-49858128 on Build GRCh38
rs758399521 Chr.19: 49857997-49857997 on Build GRCh38
rs368113668 Chr.19: 49858012-49858012 on Build GRCh38
rs769216423 Chr.19: 49858147-49858147 on Build GRCh38
rs780046219 Chr.19: 49858059-49858059 on Build GRCh38
rs768891544 Chr.19: 49857992-49857992 on Build GRCh38
rs755864677 Chr.19: 49858068-49858068 on Build GRCh38
rs779418615 Chr.19: 49857999-49857999 on Build GRCh38
rs746532230 Chr.19: 49858002-49858002 on Build GRCh38
rs543377969 Chr.19: 49858087-49858087 on Build GRCh38
rs574048043 Chr.19: 49858018-49858018 on Build GRCh38
rs760139273 Chr.19: 49857974-49857974 on Build GRCh38
rs202196523 Chr.19: 49858037-49858037 on Build GRCh38
rs146945099 Chr.19: 49858063-49858063 on Build GRCh38
rs776730341 Chr.19: 49858102-49858102 on Build GRCh38
rs200354830 Chr.19: 49858028-49858028 on Build GRCh38
rs764908574 Chr.19: 49858048-49858048 on Build GRCh38
rs746254567 Chr.19: 49858075-49858075 on Build GRCh38
rs765512552 Chr.19: 49858111-49858111 on Build GRCh38
rs200072872 Chr.19: 49858038-49858038 on Build GRCh38
rs769868353 Chr.19: 49858149-49858149 on Build GRCh38
rs566283053 Chr.19: 49857957-49857957 on Build GRCh38
rs112931931 Chr.19: 49858055-49858055 on Build GRCh38
rs754561665 Chr.19: 49858062-49858062 on Build GRCh38
rs753558526 Chr.19: 49857992-49857992 on Build GRCh38
rs750293477 Chr.19: 49858052-49858052 on Build GRCh38
rs183611086 Chr.19: 49858035-49858035 on Build GRCh38
rs767133321 Chr.19: 49858126-49858126 on Build GRCh38
rs370977468 Chr.19: 49857991-49857991 on Build GRCh38
rs2290286 Chr.19: 49858096-49858096 on Build GRCh38
rs969352671 Chr.19: 49858030-49858030 on Build GRCh38
rs780444391 Chr.19: 49858140-49858140 on Build GRCh38
rs768699383 Chr.19: 49858099-49858099 on Build GRCh38
rs540090112 Chr.19: 49857969-49857969 on Build GRCh38
rs776003296 Chr.19: 49858093-49858093 on Build GRCh38
rs759954073 Chr.19: 49858034-49858034 on Build GRCh38
rs540934989 Chr.19: 49858045-49858045 on Build GRCh38
rs766337690 Chr.19: 49858057-49858057 on Build GRCh38

View Full Product Details