Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 36066184-36066783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARAP2
Entrez Gene ID
116984
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2
Gene Aliases
CENTD1, PARX
UniGene
Hs.479451
Gene Chromosome Location
Chr.4: 36005186-36244832 on Build GRCh38
Transcripts
NM_015230.3 XM_017007698.1 XM_017007699.1 XM_017007700.1 XM_017007701.1 XM_017007702.1

SNP Detail

SNP ID Chromosome Location
rs779212830 Chr.4: 36066458-36066458 on Build GRCh38
rs899812132 Chr.4: 36066374-36066374 on Build GRCh38
rs1045320940 Chr.4: 36066727-36066727 on Build GRCh38
rs998200859 Chr.4: 36066380-36066380 on Build GRCh38
rs953318118 Chr.4: 36066288-36066288 on Build GRCh38
rs548037148 Chr.4: 36066456-36066456 on Build GRCh38
rs1016136951 Chr.4: 36066580-36066580 on Build GRCh38
rs899955264 Chr.4: 36066592-36066592 on Build GRCh38
rs1045351211 Chr.4: 36066492-36066492 on Build GRCh38
rs1001461617 Chr.4: 36066505-36066505 on Build GRCh38
rs537114063 Chr.4: 36066466-36066466 on Build GRCh38
rs888590265 Chr.4: 36066571-36066571 on Build GRCh38
rs1022716547 Chr.4: 36066716-36066716 on Build GRCh38
rs903983751 Chr.4: 36066500-36066500 on Build GRCh38
rs985976629 Chr.4: 36066556-36066556 on Build GRCh38
rs570446526 Chr.4: 36066656-36066656 on Build GRCh38
rs971202609 Chr.4: 36066719-36066719 on Build GRCh38
rs755299675 Chr.4: 36066571-36066574 on Build GRCh38
rs918996313 Chr.4: 36066661-36066661 on Build GRCh38
rs991210188 Chr.4: 36066461-36066461 on Build GRCh38
rs956706554 Chr.4: 36066226-36066226 on Build GRCh38
rs948320751 Chr.4: 36066487-36066487 on Build GRCh38
rs976499930 Chr.4: 36066243-36066243 on Build GRCh38
rs1030327484 Chr.4: 36066545-36066545 on Build GRCh38
rs772261558 Chr.4: 36066684-36066684 on Build GRCh38
rs997506206 Chr.4: 36066544-36066544 on Build GRCh38
rs777195909 Chr.4: 36066243-36066246 on Build GRCh38
rs981498636 Chr.4: 36066726-36066726 on Build GRCh38
rs1018331170 Chr.4: 36066348-36066348 on Build GRCh38
rs768698115 Chr.4: 36066387-36066387 on Build GRCh38
rs749687567 Chr.4: 36066297-36066297 on Build GRCh38
rs879455876 Chr.4: 36066570-36066571 on Build GRCh38
rs972336624 Chr.4: 36066639-36066639 on Build GRCh38
rs948372767 Chr.4: 36066710-36066710 on Build GRCh38
rs930131328 Chr.4: 36066242-36066242 on Build GRCh38
rs903083739 Chr.4: 36066663-36066663 on Build GRCh38
rs558699932 Chr.4: 36066549-36066549 on Build GRCh38
rs941866049 Chr.4: 36066314-36066314 on Build GRCh38
rs963218970 Chr.4: 36066596-36066596 on Build GRCh38
rs1029692713 Chr.4: 36066645-36066646 on Build GRCh38
rs776680109 Chr.4: 36066688-36066688 on Build GRCh38
rs915547455 Chr.4: 36066708-36066708 on Build GRCh38
rs554444210 Chr.4: 36066701-36066701 on Build GRCh38
rs923175802 Chr.4: 36066251-36066251 on Build GRCh38
rs981168455 Chr.4: 36066513-36066513 on Build GRCh38
rs971801121 Chr.4: 36066333-36066333 on Build GRCh38
rs1031508834 Chr.4: 36066223-36066223 on Build GRCh38
rs977647547 Chr.4: 36066451-36066451 on Build GRCh38
rs1005615257 Chr.4: 36066268-36066268 on Build GRCh38
rs998586289 Chr.4: 36066624-36066624 on Build GRCh38
rs953287508 Chr.4: 36066546-36066546 on Build GRCh38
rs182271449 Chr.4: 36066459-36066459 on Build GRCh38
rs1029991309 Chr.4: 36066389-36066389 on Build GRCh38
rs1031054157 Chr.4: 36066508-36066508 on Build GRCh38
rs534366950 Chr.4: 36066248-36066251 on Build GRCh38
rs1000925630 Chr.4: 36066206-36066206 on Build GRCh38
rs565536711 Chr.4: 36066305-36066305 on Build GRCh38
rs985922502 Chr.4: 36066292-36066292 on Build GRCh38
rs896738747 Chr.4: 36066482-36066482 on Build GRCh38
rs967354717 Chr.4: 36066390-36066394 on Build GRCh38
rs892572484 Chr.4: 36066518-36066518 on Build GRCh38
rs909083454 Chr.4: 36066293-36066293 on Build GRCh38
rs2271810 Chr.4: 36066664-36066664 on Build GRCh38
rs1039724830 Chr.4: 36066571-36066571 on Build GRCh38

View Full Product Details