Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184117633-184118033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP6
Entrez Gene ID
133121
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 6
Gene Aliases
NPP6
UniGene
Hs.297814
Gene Chromosome Location
Chr.4: 184088706-184217961 on Build GRCh38
Transcripts
NM_153343.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5743054 c.598G>A p.G200R Chr.4: 184117836-184117836 on Build GRCh38
1053685 c.560T>C p.I187T Chr.4: 184117874-184117874 on Build GRCh38
5767045 c.579C>T p.D193D Chr.4: 184117855-184117855 on Build GRCh38
3602426 c.568G>A p.E190K Chr.4: 184117866-184117866 on Build GRCh38
3602427 c.540C>T p.G180G Chr.4: 184117894-184117894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200817506 Chr.4: 184117885-184117885 on Build GRCh38
rs575580266 Chr.4: 184117829-184117829 on Build GRCh38
rs749916932 Chr.4: 184117936-184117936 on Build GRCh38
rs781129375 Chr.4: 184117711-184117711 on Build GRCh38
rs763550965 Chr.4: 184117862-184117862 on Build GRCh38
rs201529329 Chr.4: 184117906-184117906 on Build GRCh38
rs759136476 Chr.4: 184117898-184117898 on Build GRCh38
rs143413078 Chr.4: 184117892-184117892 on Build GRCh38
rs754469609 Chr.4: 184117865-184117865 on Build GRCh38
rs138463843 Chr.4: 184117868-184117868 on Build GRCh38
rs866481439 Chr.4: 184117830-184117830 on Build GRCh38
rs201041142 Chr.4: 184117908-184117908 on Build GRCh38
rs74865283 Chr.4: 184117822-184117822 on Build GRCh38
rs762262213 Chr.4: 184117854-184117854 on Build GRCh38
rs1035941780 Chr.4: 184117998-184117998 on Build GRCh38
rs928380374 Chr.4: 184117851-184117851 on Build GRCh38
rs766044558 Chr.4: 184117863-184117863 on Build GRCh38
rs200918603 Chr.4: 184117772-184117772 on Build GRCh38
rs543843235 Chr.4: 184117823-184117823 on Build GRCh38
rs145954744 Chr.4: 184117663-184117663 on Build GRCh38
rs375249112 Chr.4: 184117741-184117741 on Build GRCh38
rs754663839 Chr.4: 184117801-184117801 on Build GRCh38
rs1037865957 Chr.4: 184117660-184117660 on Build GRCh38
rs191005965 Chr.4: 184117723-184117723 on Build GRCh38
rs979919577 Chr.4: 184117996-184117996 on Build GRCh38
rs753876812 Chr.4: 184117875-184117875 on Build GRCh38
rs748287540 Chr.4: 184117888-184117888 on Build GRCh38
rs1035169324 Chr.4: 184117954-184117954 on Build GRCh38
rs778846976 Chr.4: 184117932-184117932 on Build GRCh38
rs369757915 Chr.4: 184117887-184117887 on Build GRCh38
rs140638467 Chr.4: 184117855-184117855 on Build GRCh38
rs756877237 Chr.4: 184117710-184117711 on Build GRCh38
rs781128011 Chr.4: 184117940-184117940 on Build GRCh38
rs151030639 Chr.4: 184117790-184117790 on Build GRCh38
rs753583719 Chr.4: 184117920-184117920 on Build GRCh38
rs565924948 Chr.4: 184117992-184117992 on Build GRCh38
rs765957723 Chr.4: 184117789-184117789 on Build GRCh38
rs900709147 Chr.4: 184117687-184117687 on Build GRCh38
rs370715887 Chr.4: 184118009-184118009 on Build GRCh38
rs114691085 Chr.4: 184117837-184117837 on Build GRCh38
rs371882346 Chr.4: 184117838-184117838 on Build GRCh38
rs139870308 Chr.4: 184117693-184117693 on Build GRCh38
rs769245882 Chr.4: 184117719-184117719 on Build GRCh38
rs777396356 Chr.4: 184117871-184117871 on Build GRCh38
rs113173347 Chr.4: 184117965-184117965 on Build GRCh38
rs747978016 Chr.4: 184117816-184117816 on Build GRCh38
rs1021513147 Chr.4: 184117978-184117978 on Build GRCh38
rs780933102 Chr.4: 184117804-184117804 on Build GRCh38
rs757009538 Chr.4: 184117927-184117927 on Build GRCh38
rs563640719 Chr.4: 184117947-184117947 on Build GRCh38
rs774957563 Chr.4: 184117896-184117896 on Build GRCh38
rs757203954 Chr.4: 184117880-184117880 on Build GRCh38
rs115181649 Chr.4: 184117738-184117738 on Build GRCh38
rs772954559 Chr.4: 184117721-184117721 on Build GRCh38
rs75116728 Chr.4: 184117969-184117969 on Build GRCh38
rs147979605 Chr.4: 184117893-184117893 on Build GRCh38
rs747125622 Chr.4: 184117884-184117884 on Build GRCh38
rs1030967482 Chr.4: 184117771-184117771 on Build GRCh38
rs375176663 Chr.4: 184117882-184117882 on Build GRCh38
rs746548467 Chr.4: 184117739-184117739 on Build GRCh38
rs548790121 Chr.4: 184117991-184117991 on Build GRCh38
rs371057717 Chr.4: 184117945-184117945 on Build GRCh38
rs772088679 Chr.4: 184117734-184117734 on Build GRCh38
rs764806393 Chr.4: 184117820-184117820 on Build GRCh38
rs755823029 Chr.4: 184117817-184117817 on Build GRCh38
rs752430549 Chr.4: 184117907-184117907 on Build GRCh38
rs748462092 Chr.4: 184117942-184117942 on Build GRCh38
rs748093395 Chr.4: 184117713-184117713 on Build GRCh38
rs890785461 Chr.4: 184117665-184117665 on Build GRCh38
rs763747839 Chr.4: 184117917-184117917 on Build GRCh38
rs116426631 Chr.4: 184117861-184117861 on Build GRCh38
rs559930706 Chr.4: 184117864-184117864 on Build GRCh38
rs149213000 Chr.4: 184117869-184117869 on Build GRCh38
rs149795320 Chr.4: 184117836-184117836 on Build GRCh38
rs749755989 Chr.4: 184117673-184117673 on Build GRCh38
rs757976046 Chr.4: 184117776-184117776 on Build GRCh38
rs769782111 Chr.4: 184117717-184117717 on Build GRCh38
rs750339734 Chr.4: 184117938-184117938 on Build GRCh38
rs974176198 Chr.4: 184117993-184117993 on Build GRCh38
rs369581585 Chr.4: 184117768-184117768 on Build GRCh38
rs113089442 Chr.4: 184117796-184117796 on Build GRCh38
rs762020121 Chr.4: 184117743-184117743 on Build GRCh38
rs111690890 Chr.4: 184117946-184117946 on Build GRCh38
rs776679814 Chr.4: 184117840-184117840 on Build GRCh38
rs765805733 Chr.4: 184117753-184117753 on Build GRCh38
rs767308783 Chr.4: 184117867-184117867 on Build GRCh38

View Full Product Details