Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 24698185-24698783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DFNA5
Entrez Gene ID
1687
Gene Name
DFNA5, deafness associated tumor suppressor
Gene Aliases
ICERE-1
UniGene
Hs.520708
Gene Chromosome Location
Chr.7: 24698355-24758020 on Build GRCh38
Transcripts
NM_001127453.1 NM_001127454.1 NM_004403.2 XM_017011802.1

SNP Detail

SNP ID Chromosome Location
rs538502935 Chr.7: 24698597-24698597 on Build GRCh38
rs953508727 Chr.7: 24698603-24698603 on Build GRCh38
rs144061292 Chr.7: 24698578-24698578 on Build GRCh38
rs910686693 Chr.7: 24698350-24698350 on Build GRCh38
rs113887389 Chr.7: 24698231-24698231 on Build GRCh38
rs914806198 Chr.7: 24698236-24698236 on Build GRCh38
rs577672103 Chr.7: 24698727-24698727 on Build GRCh38
rs1021234765 Chr.7: 24698535-24698535 on Build GRCh38
rs545583173 Chr.7: 24698240-24698240 on Build GRCh38
rs554412975 Chr.7: 24698700-24698700 on Build GRCh38
rs566482254 Chr.7: 24698543-24698544 on Build GRCh38
rs990122927 Chr.7: 24698614-24698614 on Build GRCh38
rs1004089153 Chr.7: 24698519-24698519 on Build GRCh38
rs6957782 Chr.7: 24698499-24698499 on Build GRCh38
rs562252726 Chr.7: 24698375-24698379 on Build GRCh38
rs928788764 Chr.7: 24698394-24698394 on Build GRCh38
rs967581374 Chr.7: 24698640-24698640 on Build GRCh38
rs1021646273 Chr.7: 24698636-24698636 on Build GRCh38
rs370650011 Chr.7: 24698468-24698468 on Build GRCh38
rs77276759 Chr.7: 24698521-24698521 on Build GRCh38
rs938900490 Chr.7: 24698397-24698397 on Build GRCh38
rs569722624 Chr.7: 24698579-24698579 on Build GRCh38
rs886062221 Chr.7: 24698419-24698419 on Build GRCh38
rs569478245 Chr.7: 24698620-24698620 on Build GRCh38
rs868737385 Chr.7: 24698407-24698407 on Build GRCh38
rs779682652 Chr.7: 24698219-24698219 on Build GRCh38
rs556430889 Chr.7: 24698745-24698745 on Build GRCh38
rs915622012 Chr.7: 24698707-24698707 on Build GRCh38
rs753423977 Chr.7: 24698254-24698254 on Build GRCh38
rs772900361 Chr.7: 24698683-24698683 on Build GRCh38
rs919456250 Chr.7: 24698629-24698629 on Build GRCh38
rs894253402 Chr.7: 24698436-24698436 on Build GRCh38
rs12979 Chr.7: 24698545-24698545 on Build GRCh38
rs879645458 Chr.7: 24698688-24698688 on Build GRCh38
rs748962180 Chr.7: 24698485-24698486 on Build GRCh38
rs547049769 Chr.7: 24698424-24698424 on Build GRCh38
rs192289965 Chr.7: 24698548-24698548 on Build GRCh38
rs928935167 Chr.7: 24698669-24698669 on Build GRCh38
rs771701952 Chr.7: 24698659-24698659 on Build GRCh38
rs953696195 Chr.7: 24698571-24698571 on Build GRCh38
rs201456060 Chr.7: 24698540-24698541 on Build GRCh38
rs898066909 Chr.7: 24698419-24698419 on Build GRCh38
rs1002177709 Chr.7: 24698723-24698723 on Build GRCh38
rs1041871713 Chr.7: 24698306-24698306 on Build GRCh38
rs942195114 Chr.7: 24698405-24698405 on Build GRCh38
rs534617327 Chr.7: 24698693-24698693 on Build GRCh38
rs542923347 Chr.7: 24698558-24698558 on Build GRCh38
rs17149888 Chr.7: 24698753-24698753 on Build GRCh38
rs772614606 Chr.7: 24698415-24698415 on Build GRCh38
rs762544686 Chr.7: 24698575-24698575 on Build GRCh38
rs572856899 Chr.7: 24698209-24698210 on Build GRCh38
rs538537133 Chr.7: 24698730-24698730 on Build GRCh38
rs886062222 Chr.7: 24698443-24698443 on Build GRCh38
rs893879215 Chr.7: 24698752-24698752 on Build GRCh38
rs1010707070 Chr.7: 24698528-24698528 on Build GRCh38
rs946296309 Chr.7: 24698241-24698241 on Build GRCh38
rs573722628 Chr.7: 24698705-24698705 on Build GRCh38
rs1033727122 Chr.7: 24698503-24698503 on Build GRCh38
rs565290367 Chr.7: 24698427-24698427 on Build GRCh38
rs966650715 Chr.7: 24698538-24698539 on Build GRCh38
rs1047092084 Chr.7: 24698683-24698684 on Build GRCh38
rs941121185 Chr.7: 24698613-24698613 on Build GRCh38
rs931077495 Chr.7: 24698726-24698726 on Build GRCh38
rs117584512 Chr.7: 24698489-24698489 on Build GRCh38
rs556056755 Chr.7: 24698520-24698520 on Build GRCh38
rs144928530 Chr.7: 24698680-24698680 on Build GRCh38
rs118110860 Chr.7: 24698602-24698602 on Build GRCh38
rs1046388505 Chr.7: 24698475-24698475 on Build GRCh38

View Full Product Details