Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235166804-235167224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID4B
Entrez Gene ID
51742
Gene Name
AT-rich interaction domain 4B
Gene Aliases
BCAA, BRCAA1, RBBP1L1, RBP1L1, SAP180
UniGene
Hs.575782
Gene Chromosome Location
Chr.1: 235166895-235328240 on Build GRCh38
Transcripts
NM_001206794.1 NM_016374.5 NM_031371.3 NR_135074.1 XM_006711781.2 XM_011544212.2 XM_017001468.1 XM_017001469.1 XM_017001470.1 XM_017001472.1

SNP Detail

SNP ID Chromosome Location
rs772448517 Chr.1: 235166981-235166981 on Build GRCh38
rs570781902 Chr.1: 235167087-235167087 on Build GRCh38
rs760996959 Chr.1: 235167124-235167124 on Build GRCh38
rs918201138 Chr.1: 235166986-235166989 on Build GRCh38
rs1006410779 Chr.1: 235166876-235166876 on Build GRCh38
rs961790921 Chr.1: 235166916-235166916 on Build GRCh38
rs925228730 Chr.1: 235167085-235167085 on Build GRCh38
rs970962080 Chr.1: 235167000-235167000 on Build GRCh38
rs913733518 Chr.1: 235167152-235167152 on Build GRCh38
rs773693560 Chr.1: 235167074-235167074 on Build GRCh38
rs987963266 Chr.1: 235167151-235167151 on Build GRCh38
rs936426814 Chr.1: 235167030-235167030 on Build GRCh38
rs995526392 Chr.1: 235166899-235166899 on Build GRCh38
rs112339865 Chr.1: 235167023-235167023 on Build GRCh38
rs1038861311 Chr.1: 235167205-235167205 on Build GRCh38
rs1046273167 Chr.1: 235167197-235167197 on Build GRCh38
rs928610586 Chr.1: 235167200-235167200 on Build GRCh38
rs11195 Chr.1: 235167115-235167115 on Build GRCh38
rs534002467 Chr.1: 235166997-235166997 on Build GRCh38
rs913576747 Chr.1: 235167054-235167054 on Build GRCh38
rs983055953 Chr.1: 235166980-235166980 on Build GRCh38
rs1015523663 Chr.1: 235166915-235166915 on Build GRCh38
rs1053918582 Chr.1: 235167052-235167052 on Build GRCh38
rs964119773 Chr.1: 235166856-235166856 on Build GRCh38
rs997681475 Chr.1: 235166966-235166966 on Build GRCh38
rs946114334 Chr.1: 235167173-235167173 on Build GRCh38
rs887931994 Chr.1: 235166854-235166854 on Build GRCh38
rs1029952585 Chr.1: 235167012-235167012 on Build GRCh38
rs375206731 Chr.1: 235166845-235166845 on Build GRCh38
rs1026998457 Chr.1: 235166910-235166910 on Build GRCh38

View Full Product Details