Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62210369-62210930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
L1TD1
Entrez Gene ID
54596
Gene Name
LINE1 type transposase domain containing 1
Gene Aliases
ECAT11
UniGene
Hs.685462
Gene Chromosome Location
Chr.1: 62194802-62212328 on Build GRCh38
Transcripts
NM_001164835.1 NM_019079.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6564981 c.1956G>C p.L652L Chr.1: 62210730-62210730 on Build GRCh38
5533603 c.1609G>A p.E537K Chr.1: 62210383-62210383 on Build GRCh38
911322 c.2142G>T p.E714D Chr.1: 62210916-62210916 on Build GRCh38
3491581 c.1656C>A p.T552T Chr.1: 62210430-62210430 on Build GRCh38
911318 c.2016A>C p.E672D Chr.1: 62210790-62210790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775919925 Chr.1: 62210513-62210513 on Build GRCh38
rs763813062 Chr.1: 62210752-62210752 on Build GRCh38
rs991272739 Chr.1: 62210853-62210857 on Build GRCh38
rs377478013 Chr.1: 62210674-62210674 on Build GRCh38
rs986976636 Chr.1: 62210767-62210767 on Build GRCh38
rs772771600 Chr.1: 62210684-62210684 on Build GRCh38
rs150779448 Chr.1: 62210533-62210533 on Build GRCh38
rs749914250 Chr.1: 62210798-62210798 on Build GRCh38
rs746559652 Chr.1: 62210515-62210515 on Build GRCh38
rs552938019 Chr.1: 62210700-62210700 on Build GRCh38
rs753974080 Chr.1: 62210541-62210543 on Build GRCh38
rs147452990 Chr.1: 62210421-62210421 on Build GRCh38
rs768923412 Chr.1: 62210667-62210667 on Build GRCh38
rs770867900 Chr.1: 62210661-62210662 on Build GRCh38
rs752133168 Chr.1: 62210650-62210651 on Build GRCh38
rs773410172 Chr.1: 62210699-62210699 on Build GRCh38
rs559373020 Chr.1: 62210493-62210493 on Build GRCh38
rs751434685 Chr.1: 62210762-62210762 on Build GRCh38
rs751646211 Chr.1: 62210452-62210452 on Build GRCh38
rs758340016 Chr.1: 62210487-62210487 on Build GRCh38
rs745536159 Chr.1: 62210499-62210499 on Build GRCh38
rs529756494 Chr.1: 62210541-62210541 on Build GRCh38
rs779718860 Chr.1: 62210830-62210830 on Build GRCh38
rs568498567 Chr.1: 62210881-62210881 on Build GRCh38
rs375076178 Chr.1: 62210898-62210898 on Build GRCh38
rs143856414 Chr.1: 62210503-62210503 on Build GRCh38
rs949250789 Chr.1: 62210655-62210655 on Build GRCh38
rs763522788 Chr.1: 62210450-62210450 on Build GRCh38
rs747956601 Chr.1: 62210481-62210481 on Build GRCh38
rs777615995 Chr.1: 62210490-62210490 on Build GRCh38
rs896285167 Chr.1: 62210633-62210633 on Build GRCh38
rs757463322 Chr.1: 62210455-62210455 on Build GRCh38
rs2886644 Chr.1: 62210612-62210612 on Build GRCh38
rs779622892 Chr.1: 62210640-62210640 on Build GRCh38
rs747900222 Chr.1: 62210553-62210554 on Build GRCh38
rs900192306 Chr.1: 62210730-62210730 on Build GRCh38
rs202029696 Chr.1: 62210576-62210579 on Build GRCh38
rs766564924 Chr.1: 62210563-62210563 on Build GRCh38
rs955577840 Chr.1: 62210542-62210542 on Build GRCh38
rs541806917 Chr.1: 62210553-62210553 on Build GRCh38
rs996298120 Chr.1: 62210736-62210736 on Build GRCh38
rs777729013 Chr.1: 62210598-62210598 on Build GRCh38
rs190573555 Chr.1: 62210390-62210390 on Build GRCh38
rs147208373 Chr.1: 62210516-62210516 on Build GRCh38
rs779235664 Chr.1: 62210561-62210562 on Build GRCh38
rs778103879 Chr.1: 62210432-62210433 on Build GRCh38
rs546879656 Chr.1: 62210854-62210854 on Build GRCh38
rs781166127 Chr.1: 62210777-62210777 on Build GRCh38
rs774941220 Chr.1: 62210669-62210669 on Build GRCh38
rs771414993 Chr.1: 62210428-62210428 on Build GRCh38
rs766588112 Chr.1: 62210709-62210709 on Build GRCh38
rs769184330 Chr.1: 62210521-62210521 on Build GRCh38
rs764578131 Chr.1: 62210451-62210451 on Build GRCh38
rs919912915 Chr.1: 62210811-62210811 on Build GRCh38
rs754851639 Chr.1: 62210417-62210417 on Build GRCh38
rs528655805 Chr.1: 62210764-62210764 on Build GRCh38
rs1032003600 Chr.1: 62210446-62210446 on Build GRCh38
rs751524660 Chr.1: 62210606-62210606 on Build GRCh38
rs549166182 Chr.1: 62210613-62210615 on Build GRCh38
rs755620644 Chr.1: 62210806-62210806 on Build GRCh38
rs750695998 Chr.1: 62210463-62210463 on Build GRCh38
rs745450964 Chr.1: 62210615-62210615 on Build GRCh38
rs370890823 Chr.1: 62210614-62210614 on Build GRCh38
rs774219143 Chr.1: 62210680-62210680 on Build GRCh38
rs770543773 Chr.1: 62210445-62210445 on Build GRCh38
rs1015805590 Chr.1: 62210790-62210790 on Build GRCh38
rs11207933 Chr.1: 62210419-62210419 on Build GRCh38
rs781622703 Chr.1: 62210398-62210398 on Build GRCh38
rs768998315 Chr.1: 62210882-62210882 on Build GRCh38
rs1030102894 Chr.1: 62210740-62210740 on Build GRCh38
rs758163738 Chr.1: 62210583-62210583 on Build GRCh38
rs754759991 Chr.1: 62210467-62210467 on Build GRCh38
rs939968607 Chr.1: 62210581-62210581 on Build GRCh38
rs1031951649 Chr.1: 62210725-62210725 on Build GRCh38
rs747744420 Chr.1: 62210613-62210614 on Build GRCh38
rs769158189 Chr.1: 62210638-62210639 on Build GRCh38
rs752512393 Chr.1: 62210568-62210568 on Build GRCh38
rs749635272 Chr.1: 62210649-62210649 on Build GRCh38
rs866849348 Chr.1: 62210749-62210749 on Build GRCh38
rs780064975 Chr.1: 62210390-62210391 on Build GRCh38
rs772772111 Chr.1: 62210434-62210434 on Build GRCh38
rs140755361 Chr.1: 62210657-62210657 on Build GRCh38
rs569988115 Chr.1: 62210593-62210593 on Build GRCh38
rs571071314 Chr.1: 62210719-62210719 on Build GRCh38
rs746554359 Chr.1: 62210441-62210441 on Build GRCh38
rs767509504 Chr.1: 62210836-62210836 on Build GRCh38
rs577004201 Chr.1: 62210458-62210458 on Build GRCh38
rs377660865 Chr.1: 62210528-62210528 on Build GRCh38
rs775264428 Chr.1: 62210849-62210850 on Build GRCh38
rs761934726 Chr.1: 62210540-62210540 on Build GRCh38
rs938039434 Chr.1: 62210852-62210852 on Build GRCh38
rs749070660 Chr.1: 62210402-62210402 on Build GRCh38
rs762520115 Chr.1: 62210745-62210745 on Build GRCh38
rs955691359 Chr.1: 62210751-62210751 on Build GRCh38
rs759573169 Chr.1: 62210833-62210836 on Build GRCh38
rs762502674 Chr.1: 62210658-62210662 on Build GRCh38
rs775805713 Chr.1: 62210449-62210449 on Build GRCh38
rs757549990 Chr.1: 62210393-62210393 on Build GRCh38
rs770463644 Chr.1: 62210495-62210495 on Build GRCh38
rs370854912 Chr.1: 62210892-62210892 on Build GRCh38
rs540884928 Chr.1: 62210472-62210472 on Build GRCh38
rs746379678 Chr.1: 62210571-62210573 on Build GRCh38
rs762763244 Chr.1: 62210741-62210741 on Build GRCh38
rs776220924 Chr.1: 62210498-62210498 on Build GRCh38
rs759858325 Chr.1: 62210448-62210448 on Build GRCh38
rs769476610 Chr.1: 62210630-62210630 on Build GRCh38
rs760973536 Chr.1: 62210558-62210558 on Build GRCh38
rs776310502 Chr.1: 62210447-62210447 on Build GRCh38

View Full Product Details