Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 284359-284913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM5A
Entrez Gene ID
5927
Gene Name
lysine demethylase 5A
Gene Aliases
RBBP-2, RBBP2, RBP2
UniGene
Hs.76272
Gene Chromosome Location
Chr.12: 280057-389455 on Build GRCh38
Transcripts
NM_001042603.2

SNP Detail

SNP ID Chromosome Location
rs781042752 Chr.12: 284395-284395 on Build GRCh38
rs118142329 Chr.12: 284605-284605 on Build GRCh38
rs773420021 Chr.12: 284629-284631 on Build GRCh38
rs7973426 Chr.12: 284731-284731 on Build GRCh38
rs997267655 Chr.12: 284800-284800 on Build GRCh38
rs116745728 Chr.12: 284558-284558 on Build GRCh38
rs988981216 Chr.12: 284762-284762 on Build GRCh38
rs745786366 Chr.12: 284574-284574 on Build GRCh38
rs766650765 Chr.12: 284842-284842 on Build GRCh38
rs979123395 Chr.12: 284551-284551 on Build GRCh38
rs541812095 Chr.12: 284610-284610 on Build GRCh38
rs79783495 Chr.12: 284639-284639 on Build GRCh38
rs1038989268 Chr.12: 284597-284597 on Build GRCh38
rs192907672 Chr.12: 284793-284793 on Build GRCh38
rs138695488 Chr.12: 284603-284603 on Build GRCh38
rs983461371 Chr.12: 284436-284436 on Build GRCh38
rs983717893 Chr.12: 284837-284837 on Build GRCh38
rs916717189 Chr.12: 284501-284501 on Build GRCh38
rs982710588 Chr.12: 284517-284517 on Build GRCh38
rs557335094 Chr.12: 284460-284460 on Build GRCh38
rs541136053 Chr.12: 284600-284600 on Build GRCh38
rs991039162 Chr.12: 284500-284500 on Build GRCh38
rs967689630 Chr.12: 284491-284491 on Build GRCh38
rs76565406 Chr.12: 284620-284620 on Build GRCh38
rs1017242745 Chr.12: 284739-284739 on Build GRCh38
rs762024580 Chr.12: 284598-284598 on Build GRCh38
rs891963166 Chr.12: 284625-284625 on Build GRCh38
rs964434478 Chr.12: 284573-284573 on Build GRCh38
rs575608478 Chr.12: 284482-284482 on Build GRCh38
rs74665848 Chr.12: 284638-284638 on Build GRCh38
rs574193851 Chr.12: 284586-284586 on Build GRCh38
rs964382394 Chr.12: 284791-284791 on Build GRCh38
rs950560340 Chr.12: 284818-284818 on Build GRCh38
rs546413552 Chr.12: 284789-284789 on Build GRCh38
rs79500657 Chr.12: 284649-284649 on Build GRCh38
rs914219244 Chr.12: 284776-284776 on Build GRCh38
rs76286555 Chr.12: 284704-284704 on Build GRCh38
rs934394828 Chr.12: 284758-284758 on Build GRCh38
rs1010846532 Chr.12: 284628-284628 on Build GRCh38
rs142708753 Chr.12: 284675-284675 on Build GRCh38
rs949572140 Chr.12: 284504-284504 on Build GRCh38
rs963556526 Chr.12: 284472-284472 on Build GRCh38
rs904748934 Chr.12: 284729-284729 on Build GRCh38
rs145879157 Chr.12: 284589-284589 on Build GRCh38
rs199915159 Chr.12: 284638-284639 on Build GRCh38
rs938726916 Chr.12: 284611-284611 on Build GRCh38
rs1023823941 Chr.12: 284725-284725 on Build GRCh38
rs963359601 Chr.12: 284693-284693 on Build GRCh38
rs76812733 Chr.12: 284417-284417 on Build GRCh38
rs531061792 Chr.12: 284646-284646 on Build GRCh38
rs963239331 Chr.12: 284861-284861 on Build GRCh38
rs535493072 Chr.12: 284875-284875 on Build GRCh38
rs148398656 Chr.12: 284863-284863 on Build GRCh38
rs934444834 Chr.12: 284561-284561 on Build GRCh38
rs1016714885 Chr.12: 284443-284443 on Build GRCh38
rs1005377853 Chr.12: 284615-284615 on Build GRCh38
rs1030534405 Chr.12: 284813-284813 on Build GRCh38

View Full Product Details