Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103573959-103574344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOPT1
Entrez Gene ID
84334
Gene Name
apoptogenic 1, mitochondrial
Gene Aliases
APOP, APOP1, C14orf153
UniGene
Hs.598441
Gene Chromosome Location
Chr.14: 103562957-103592187 on Build GRCh38
Transcripts
NM_001302652.1 NM_001302653.1 NM_001302654.1 NM_032374.4 NR_126431.1 NR_126432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6602481 c.374T>G p.F125C Chr.14: 103574120-103574120 on Build GRCh38
5468710 c.328G>A p.E110K Chr.14: 103574113-103574113 on Build GRCh38
1678285 c.335T>G p.F112C Chr.14: 103574120-103574120 on Build GRCh38
3689954 c.351A>G p.L117L Chr.14: 103574136-103574136 on Build GRCh38
1477385 c.363C>T p.G121G Chr.14: 103574148-103574148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs73361345 Chr.14: 103574295-103574295 on Build GRCh38
rs754793274 Chr.14: 103574218-103574218 on Build GRCh38
rs773747904 Chr.14: 103574104-103574104 on Build GRCh38
rs1046288137 Chr.14: 103574267-103574268 on Build GRCh38
rs772669466 Chr.14: 103574264-103574264 on Build GRCh38
rs753612286 Chr.14: 103574212-103574212 on Build GRCh38
rs377493532 Chr.14: 103574014-103574014 on Build GRCh38
rs781278777 Chr.14: 103574226-103574226 on Build GRCh38
rs951020033 Chr.14: 103574041-103574041 on Build GRCh38
rs150059895 Chr.14: 103574246-103574246 on Build GRCh38
rs1049769938 Chr.14: 103574090-103574090 on Build GRCh38
rs562504344 Chr.14: 103574191-103574192 on Build GRCh38
rs759039475 Chr.14: 103574106-103574106 on Build GRCh38
rs754918522 Chr.14: 103574108-103574108 on Build GRCh38
rs778923080 Chr.14: 103574066-103574067 on Build GRCh38
rs540383297 Chr.14: 103574079-103574079 on Build GRCh38
rs977868831 Chr.14: 103574247-103574247 on Build GRCh38
rs1027674154 Chr.14: 103574200-103574200 on Build GRCh38
rs749643552 Chr.14: 103574176-103574176 on Build GRCh38
rs540525175 Chr.14: 103574244-103574244 on Build GRCh38
rs766094324 Chr.14: 103574102-103574102 on Build GRCh38
rs539140983 Chr.14: 103574074-103574074 on Build GRCh38
rs187033388 Chr.14: 103574322-103574322 on Build GRCh38
rs763402953 Chr.14: 103574057-103574057 on Build GRCh38
rs772896010 Chr.14: 103574269-103574269 on Build GRCh38
rs770550856 Chr.14: 103574103-103574104 on Build GRCh38
rs1025331312 Chr.14: 103573992-103573992 on Build GRCh38
rs775742260 Chr.14: 103574141-103574141 on Build GRCh38
rs1054141068 Chr.14: 103574046-103574046 on Build GRCh38
rs546395018 Chr.14: 103574283-103574283 on Build GRCh38
rs11337243 Chr.14: 103574103-103574103 on Build GRCh38
rs201389506 Chr.14: 103574167-103574167 on Build GRCh38
rs974768824 Chr.14: 103574155-103574155 on Build GRCh38
rs752643761 Chr.14: 103574224-103574224 on Build GRCh38
rs79347665 Chr.14: 103574303-103574303 on Build GRCh38
rs769084534 Chr.14: 103574179-103574179 on Build GRCh38
rs139436329 Chr.14: 103574222-103574222 on Build GRCh38
rs760229338 Chr.14: 103574272-103574272 on Build GRCh38
rs781188986 Chr.14: 103574122-103574122 on Build GRCh38
rs772982123 Chr.14: 103574079-103574081 on Build GRCh38
rs1016822245 Chr.14: 103574062-103574062 on Build GRCh38
rs71417867 Chr.14: 103574136-103574136 on Build GRCh38
rs375549685 Chr.14: 103574211-103574211 on Build GRCh38
rs747124583 Chr.14: 103574116-103574116 on Build GRCh38
rs773782984 Chr.14: 103574073-103574073 on Build GRCh38
rs577396949 Chr.14: 103574249-103574249 on Build GRCh38
rs769966495 Chr.14: 103574137-103574137 on Build GRCh38
rs776178624 Chr.14: 103574191-103574191 on Build GRCh38
rs772783943 Chr.14: 103574204-103574204 on Build GRCh38
rs755337484 Chr.14: 103574080-103574081 on Build GRCh38
rs764611703 Chr.14: 103574192-103574192 on Build GRCh38
rs749682558 Chr.14: 103574245-103574245 on Build GRCh38
rs557614166 Chr.14: 103574316-103574316 on Build GRCh38
rs1030821072 Chr.14: 103574270-103574270 on Build GRCh38
rs894788503 Chr.14: 103574203-103574203 on Build GRCh38
rs370053620 Chr.14: 103574280-103574280 on Build GRCh38
rs757315655 Chr.14: 103574118-103574118 on Build GRCh38
rs888121190 Chr.14: 103574318-103574318 on Build GRCh38
rs35140000 Chr.14: 103574055-103574055 on Build GRCh38
rs757421250 Chr.14: 103574273-103574273 on Build GRCh38
rs775854624 Chr.14: 103574056-103574056 on Build GRCh38
rs778874587 Chr.14: 103574113-103574113 on Build GRCh38
rs200886784 Chr.14: 103574181-103574181 on Build GRCh38
rs748672371 Chr.14: 103574259-103574259 on Build GRCh38
rs761471179 Chr.14: 103574189-103574189 on Build GRCh38
rs756341441 Chr.14: 103574240-103574240 on Build GRCh38
rs750090713 Chr.14: 103574076-103574076 on Build GRCh38
rs377184826 Chr.14: 103574210-103574210 on Build GRCh38
rs754225304 Chr.14: 103574249-103574250 on Build GRCh38
rs4906337 Chr.14: 103574077-103574077 on Build GRCh38
rs745901992 Chr.14: 103574236-103574236 on Build GRCh38
rs891216333 Chr.14: 103574103-103574103 on Build GRCh38
rs745890692 Chr.14: 103574076-103574077 on Build GRCh38
rs772221375 Chr.14: 103574079-103574080 on Build GRCh38
rs770552390 Chr.14: 103574273-103574273 on Build GRCh38
rs764614443 Chr.14: 103574064-103574064 on Build GRCh38
rs770239408 Chr.14: 103574078-103574079 on Build GRCh38
rs587777787 Chr.14: 103574116-103574118 on Build GRCh38
rs753617371 Chr.14: 103574107-103574107 on Build GRCh38
rs767151805 Chr.14: 103574191-103574191 on Build GRCh38
rs758262953 Chr.14: 103574225-103574225 on Build GRCh38
rs772559167 Chr.14: 103574300-103574300 on Build GRCh38

View Full Product Details