Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27270424-27270987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASL11A
Entrez Gene ID
387496
Gene Name
RAS like family 11 member A
Gene Aliases
-
UniGene
Hs.192131
Gene Chromosome Location
Chr.13: 27270327-27273690 on Build GRCh38
Transcripts
NM_206827.1

SNP Detail

SNP ID Chromosome Location
rs186768561 Chr.13: 27270869-27270869 on Build GRCh38
rs561969353 Chr.13: 27270799-27270799 on Build GRCh38
rs935918094 Chr.13: 27270941-27270941 on Build GRCh38
rs182479086 Chr.13: 27270814-27270814 on Build GRCh38
rs754487168 Chr.13: 27270802-27270802 on Build GRCh38
rs529478414 Chr.13: 27270770-27270770 on Build GRCh38
rs1025828225 Chr.13: 27270815-27270815 on Build GRCh38
rs955426846 Chr.13: 27270833-27270833 on Build GRCh38
rs1041554793 Chr.13: 27270757-27270757 on Build GRCh38
rs758614171 Chr.13: 27270719-27270719 on Build GRCh38
rs548830357 Chr.13: 27270566-27270566 on Build GRCh38
rs1018284001 Chr.13: 27270839-27270839 on Build GRCh38
rs945835632 Chr.13: 27270738-27270738 on Build GRCh38
rs373350199 Chr.13: 27270596-27270596 on Build GRCh38
rs78525380 Chr.13: 27270546-27270546 on Build GRCh38
rs942605272 Chr.13: 27270466-27270466 on Build GRCh38
rs539134049 Chr.13: 27270710-27270710 on Build GRCh38
rs552676458 Chr.13: 27270860-27270860 on Build GRCh38
rs1000477741 Chr.13: 27270617-27270617 on Build GRCh38
rs1023594761 Chr.13: 27270654-27270654 on Build GRCh38
rs945813004 Chr.13: 27270526-27270526 on Build GRCh38
rs751390719 Chr.13: 27270929-27270929 on Build GRCh38
rs892981921 Chr.13: 27270525-27270525 on Build GRCh38
rs544558691 Chr.13: 27270819-27270819 on Build GRCh38
rs779243499 Chr.13: 27270681-27270681 on Build GRCh38
rs1054714329 Chr.13: 27270811-27270811 on Build GRCh38
rs931704441 Chr.13: 27270468-27270468 on Build GRCh38
rs528645824 Chr.13: 27270874-27270874 on Build GRCh38
rs7331765 Chr.13: 27270556-27270556 on Build GRCh38
rs914403265 Chr.13: 27270732-27270732 on Build GRCh38
rs977629560 Chr.13: 27270878-27270878 on Build GRCh38
rs1008338086 Chr.13: 27270836-27270836 on Build GRCh38
rs759588455 Chr.13: 27270921-27270921 on Build GRCh38
rs556538130 Chr.13: 27270692-27270692 on Build GRCh38
rs963245968 Chr.13: 27270671-27270671 on Build GRCh38
rs970668099 Chr.13: 27270659-27270662 on Build GRCh38
rs367718372 Chr.13: 27270930-27270930 on Build GRCh38
rs552669491 Chr.13: 27270717-27270717 on Build GRCh38
rs866994045 Chr.13: 27270948-27270948 on Build GRCh38
rs564689059 Chr.13: 27270680-27270680 on Build GRCh38
rs781382504 Chr.13: 27270920-27270920 on Build GRCh38
rs1050104177 Chr.13: 27270472-27270472 on Build GRCh38
rs1041381614 Chr.13: 27270793-27270793 on Build GRCh38
rs765619350 Chr.13: 27270560-27270560 on Build GRCh38
rs780604923 Chr.13: 27270851-27270851 on Build GRCh38
rs1032828391 Chr.13: 27270628-27270628 on Build GRCh38
rs944610497 Chr.13: 27270488-27270488 on Build GRCh38
rs914434774 Chr.13: 27270868-27270868 on Build GRCh38
rs938416975 Chr.13: 27270884-27270884 on Build GRCh38
rs765375868 Chr.13: 27270922-27270922 on Build GRCh38
rs779919095 Chr.13: 27270934-27270934 on Build GRCh38
rs570110390 Chr.13: 27270447-27270447 on Build GRCh38
rs994455525 Chr.13: 27270946-27270946 on Build GRCh38
rs1002454162 Chr.13: 27270584-27270584 on Build GRCh38
rs890104783 Chr.13: 27270482-27270482 on Build GRCh38
rs767983147 Chr.13: 27270954-27270954 on Build GRCh38
rs563657762 Chr.13: 27270847-27270847 on Build GRCh38
rs541055848 Chr.13: 27270461-27270463 on Build GRCh38
rs368072480 Chr.13: 27270467-27270467 on Build GRCh38
rs894169808 Chr.13: 27270792-27270792 on Build GRCh38
rs1017084922 Chr.13: 27270646-27270646 on Build GRCh38
rs750514059 Chr.13: 27270713-27270713 on Build GRCh38
rs911253307 Chr.13: 27270716-27270716 on Build GRCh38
rs962300611 Chr.13: 27270696-27270696 on Build GRCh38
rs568670325 Chr.13: 27270640-27270640 on Build GRCh38
rs893921817 Chr.13: 27270635-27270635 on Build GRCh38
rs572955723 Chr.13: 27270726-27270726 on Build GRCh38
rs924271919 Chr.13: 27270938-27270938 on Build GRCh38
rs895373414 Chr.13: 27270614-27270614 on Build GRCh38
rs977519987 Chr.13: 27270935-27270935 on Build GRCh38
rs576473485 Chr.13: 27270703-27270703 on Build GRCh38
rs964424884 Chr.13: 27270842-27270842 on Build GRCh38
rs780804797 Chr.13: 27270932-27270932 on Build GRCh38
rs371620480 Chr.13: 27270933-27270933 on Build GRCh38
rs1033995534 Chr.13: 27270589-27270589 on Build GRCh38
rs928500384 Chr.13: 27270882-27270882 on Build GRCh38
rs753906835 Chr.13: 27270911-27270911 on Build GRCh38
rs1012499945 Chr.13: 27270798-27270798 on Build GRCh38
rs748794891 Chr.13: 27270953-27270953 on Build GRCh38
rs972837566 Chr.13: 27270679-27270679 on Build GRCh38
rs373623695 Chr.13: 27270896-27270896 on Build GRCh38
rs1055376212 Chr.13: 27270791-27270791 on Build GRCh38
rs376471623 Chr.13: 27270952-27270952 on Build GRCh38
rs532580298 Chr.13: 27270857-27270857 on Build GRCh38
rs938447978 Chr.13: 27270782-27270782 on Build GRCh38
rs541666865 Chr.13: 27270776-27270776 on Build GRCh38
rs915572166 Chr.13: 27270915-27270915 on Build GRCh38
rs552738023 Chr.13: 27270523-27270523 on Build GRCh38
rs532642725 Chr.13: 27270452-27270452 on Build GRCh38
rs1054280782 Chr.13: 27270490-27270490 on Build GRCh38

View Full Product Details