Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 100643052-100643608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRG7
Entrez Gene ID
84873
Gene Name
adhesion G protein-coupled receptor G7
Gene Aliases
GPR128
UniGene
Hs.334511
Gene Chromosome Location
Chr.3: 100609589-100696174 on Build GRCh38
Transcripts
NM_001308362.1 NM_032787.2 XM_011513245.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5863088 c.735delG p.G246fs*10 Chr.3: 100643302-100643302 on Build GRCh38
7289055 c.738T>C p.G246G Chr.3: 100643305-100643305 on Build GRCh38
6095490 c.714G>C p.M238I Chr.3: 100643281-100643281 on Build GRCh38
1566369 c.725C>T p.S242F Chr.3: 100643292-100643292 on Build GRCh38
4687978 c.800C>T p.A267V Chr.3: 100643367-100643367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907522734 Chr.3: 100643082-100643082 on Build GRCh38
rs760069834 Chr.3: 100643178-100643178 on Build GRCh38
rs61212523 Chr.3: 100643317-100643317 on Build GRCh38
rs890084007 Chr.3: 100643126-100643126 on Build GRCh38
rs574712840 Chr.3: 100643418-100643418 on Build GRCh38
rs576424574 Chr.3: 100643212-100643212 on Build GRCh38
rs766478554 Chr.3: 100643333-100643333 on Build GRCh38
rs184610361 Chr.3: 100643148-100643148 on Build GRCh38
rs143786841 Chr.3: 100643264-100643264 on Build GRCh38
rs528132805 Chr.3: 100643492-100643492 on Build GRCh38
rs779730760 Chr.3: 100643387-100643387 on Build GRCh38
rs1012327046 Chr.3: 100643563-100643563 on Build GRCh38
rs779278985 Chr.3: 100643400-100643400 on Build GRCh38
rs1041671966 Chr.3: 100643487-100643487 on Build GRCh38
rs371369020 Chr.3: 100643557-100643557 on Build GRCh38
rs191874939 Chr.3: 100643438-100643438 on Build GRCh38
rs893654692 Chr.3: 100643213-100643213 on Build GRCh38
rs768505512 Chr.3: 100643508-100643509 on Build GRCh38
rs370228436 Chr.3: 100643298-100643298 on Build GRCh38
rs774786770 Chr.3: 100643318-100643318 on Build GRCh38
rs944280496 Chr.3: 100643138-100643138 on Build GRCh38
rs751437270 Chr.3: 100643353-100643353 on Build GRCh38
rs969236359 Chr.3: 100643375-100643375 on Build GRCh38
rs375365345 Chr.3: 100643519-100643519 on Build GRCh38
rs755835902 Chr.3: 100643498-100643498 on Build GRCh38
rs754068092 Chr.3: 100643296-100643296 on Build GRCh38
rs750572142 Chr.3: 100643248-100643248 on Build GRCh38
rs540674410 Chr.3: 100643227-100643227 on Build GRCh38
rs369615471 Chr.3: 100643516-100643516 on Build GRCh38
rs145200978 Chr.3: 100643572-100643572 on Build GRCh38
rs369645034 Chr.3: 100643276-100643276 on Build GRCh38
rs1032401918 Chr.3: 100643199-100643199 on Build GRCh38
rs753257531 Chr.3: 100643198-100643198 on Build GRCh38
rs755784886 Chr.3: 100643384-100643384 on Build GRCh38
rs370081148 Chr.3: 100643388-100643388 on Build GRCh38
rs944327164 Chr.3: 100643448-100643448 on Build GRCh38
rs747305307 Chr.3: 100643566-100643566 on Build GRCh38
rs376767128 Chr.3: 100643271-100643271 on Build GRCh38
rs760784655 Chr.3: 100643331-100643331 on Build GRCh38
rs375778486 Chr.3: 100643334-100643334 on Build GRCh38
rs766128499 Chr.3: 100643504-100643504 on Build GRCh38
rs774536133 Chr.3: 100643225-100643225 on Build GRCh38
rs750066112 Chr.3: 100643374-100643374 on Build GRCh38
rs769276093 Chr.3: 100643394-100643394 on Build GRCh38
rs756988832 Chr.3: 100643367-100643367 on Build GRCh38
rs768990409 Chr.3: 100643314-100643314 on Build GRCh38
rs931452110 Chr.3: 100643105-100643105 on Build GRCh38
rs761861284 Chr.3: 100643584-100643584 on Build GRCh38
rs769031133 Chr.3: 100643224-100643224 on Build GRCh38
rs987902569 Chr.3: 100643118-100643118 on Build GRCh38
rs778125412 Chr.3: 100643549-100643549 on Build GRCh38
rs960402650 Chr.3: 100643112-100643112 on Build GRCh38
rs371504737 Chr.3: 100643581-100643581 on Build GRCh38
rs776695277 Chr.3: 100643431-100643431 on Build GRCh38
rs41272971 Chr.3: 100643270-100643270 on Build GRCh38
rs902527467 Chr.3: 100643488-100643488 on Build GRCh38
rs749830310 Chr.3: 100643311-100643311 on Build GRCh38
rs940477706 Chr.3: 100643092-100643092 on Build GRCh38
rs913650050 Chr.3: 100643124-100643124 on Build GRCh38
rs1023495878 Chr.3: 100643564-100643564 on Build GRCh38
rs368699106 Chr.3: 100643522-100643522 on Build GRCh38
rs1049973565 Chr.3: 100643114-100643114 on Build GRCh38
rs757459939 Chr.3: 100643300-100643300 on Build GRCh38
rs765207432 Chr.3: 100643266-100643266 on Build GRCh38
rs372870298 Chr.3: 100643330-100643330 on Build GRCh38
rs1053877344 Chr.3: 100643503-100643503 on Build GRCh38
rs373414391 Chr.3: 100643326-100643326 on Build GRCh38
rs774130113 Chr.3: 100643445-100643445 on Build GRCh38
rs546307092 Chr.3: 100643573-100643573 on Build GRCh38
rs780986641 Chr.3: 100643278-100643278 on Build GRCh38
rs747256166 Chr.3: 100643427-100643427 on Build GRCh38
rs147822261 Chr.3: 100643184-100643184 on Build GRCh38
rs183730793 Chr.3: 100643440-100643440 on Build GRCh38
rs758854996 Chr.3: 100643524-100643524 on Build GRCh38
rs773526403 Chr.3: 100643419-100643419 on Build GRCh38
rs762111259 Chr.3: 100643226-100643226 on Build GRCh38
rs1034970208 Chr.3: 100643489-100643489 on Build GRCh38
rs758488167 Chr.3: 100643434-100643434 on Build GRCh38
rs771085330 Chr.3: 100643430-100643430 on Build GRCh38
rs777647174 Chr.3: 100643275-100643275 on Build GRCh38
rs1043635629 Chr.3: 100643177-100643177 on Build GRCh38
rs770456590 Chr.3: 100643095-100643095 on Build GRCh38
rs780743304 Chr.3: 100643308-100643308 on Build GRCh38
rs75566957 Chr.3: 100643098-100643098 on Build GRCh38
rs762959765 Chr.3: 100643337-100643337 on Build GRCh38
rs372913854 Chr.3: 100643411-100643411 on Build GRCh38
rs745556172 Chr.3: 100643147-100643147 on Build GRCh38
rs189937466 Chr.3: 100643191-100643191 on Build GRCh38
rs761704801 Chr.3: 100643453-100643453 on Build GRCh38
rs763870316 Chr.3: 100643339-100643339 on Build GRCh38
rs769727441 Chr.3: 100643575-100643575 on Build GRCh38
rs781035694 Chr.3: 100643368-100643368 on Build GRCh38

View Full Product Details