Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173487947-173488502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDX6
Entrez Gene ID
9588
Gene Name
peroxiredoxin 6
Gene Aliases
1-Cys, AOP2, HEL-S-128m, NSGPx, PRX, aiPLA2, p29
UniGene
Hs.120
Gene Chromosome Location
Chr.1: 173477347-173488807 on Build GRCh38
Transcripts
NM_004905.2

SNP Detail

SNP ID Chromosome Location
rs34931994 Chr.1: 173488223-173488224 on Build GRCh38
rs918179063 Chr.1: 173488440-173488440 on Build GRCh38
rs995693548 Chr.1: 173488285-173488285 on Build GRCh38
rs545090954 Chr.1: 173488208-173488208 on Build GRCh38
rs3211528 Chr.1: 173488439-173488439 on Build GRCh38
rs905847532 Chr.1: 173488292-173488292 on Build GRCh38
rs950709287 Chr.1: 173487980-173487980 on Build GRCh38
rs1005178202 Chr.1: 173488306-173488306 on Build GRCh38
rs558462837 Chr.1: 173488013-173488013 on Build GRCh38
rs181078869 Chr.1: 173488475-173488475 on Build GRCh38
rs988601827 Chr.1: 173488157-173488157 on Build GRCh38
rs953159998 Chr.1: 173488399-173488399 on Build GRCh38
rs35364344 Chr.1: 173488237-173488237 on Build GRCh38
rs1004539367 Chr.1: 173488118-173488118 on Build GRCh38
rs534502965 Chr.1: 173488442-173488442 on Build GRCh38
rs144526723 Chr.1: 173488323-173488323 on Build GRCh38
rs1045189323 Chr.1: 173487988-173487988 on Build GRCh38
rs970104262 Chr.1: 173488150-173488150 on Build GRCh38
rs905661905 Chr.1: 173488034-173488034 on Build GRCh38
rs577921693 Chr.1: 173488349-173488350 on Build GRCh38
rs149636735 Chr.1: 173488409-173488410 on Build GRCh38
rs36005931 Chr.1: 173488462-173488462 on Build GRCh38
rs1020105676 Chr.1: 173488350-173488350 on Build GRCh38
rs571239002 Chr.1: 173488343-173488343 on Build GRCh38
rs1042710441 Chr.1: 173487971-173487971 on Build GRCh38
rs141152960 Chr.1: 173488228-173488228 on Build GRCh38
rs75435712 Chr.1: 173488156-173488156 on Build GRCh38
rs921576119 Chr.1: 173488216-173488216 on Build GRCh38
rs35570547 Chr.1: 173488178-173488178 on Build GRCh38
rs185836646 Chr.1: 173488194-173488194 on Build GRCh38
rs1048952720 Chr.1: 173488248-173488248 on Build GRCh38
rs1037243660 Chr.1: 173488104-173488104 on Build GRCh38
rs534463922 Chr.1: 173488103-173488103 on Build GRCh38
rs995836978 Chr.1: 173488357-173488357 on Build GRCh38
rs897345468 Chr.1: 173488260-173488260 on Build GRCh38
rs778432448 Chr.1: 173488151-173488151 on Build GRCh38
rs921627944 Chr.1: 173487973-173487973 on Build GRCh38
rs987116213 Chr.1: 173488401-173488401 on Build GRCh38
rs1008385169 Chr.1: 173488074-173488074 on Build GRCh38
rs369545647 Chr.1: 173488152-173488152 on Build GRCh38
rs1048666229 Chr.1: 173488290-173488290 on Build GRCh38
rs138744415 Chr.1: 173488259-173488259 on Build GRCh38
rs1027374787 Chr.1: 173488137-173488137 on Build GRCh38
rs573473830 Chr.1: 173488052-173488052 on Build GRCh38
rs921906947 Chr.1: 173488381-173488381 on Build GRCh38
rs567268129 Chr.1: 173488459-173488459 on Build GRCh38
rs761638283 Chr.1: 173488440-173488440 on Build GRCh38
rs931694993 Chr.1: 173488226-173488226 on Build GRCh38
rs7314 Chr.1: 173488460-173488460 on Build GRCh38
rs112926420 Chr.1: 173488238-173488238 on Build GRCh38
rs1010338067 Chr.1: 173488349-173488349 on Build GRCh38
rs758149204 Chr.1: 173488160-173488160 on Build GRCh38
rs541397227 Chr.1: 173488214-173488215 on Build GRCh38
rs931409903 Chr.1: 173488477-173488481 on Build GRCh38
rs995725603 Chr.1: 173488130-173488130 on Build GRCh38
rs931700460 Chr.1: 173487978-173487978 on Build GRCh38
rs979602211 Chr.1: 173488201-173488201 on Build GRCh38
rs1051544679 Chr.1: 173488027-173488027 on Build GRCh38
rs141751235 Chr.1: 173488309-173488309 on Build GRCh38
rs189429240 Chr.1: 173488432-173488432 on Build GRCh38
rs546729955 Chr.1: 173488033-173488033 on Build GRCh38
rs74819145 Chr.1: 173488366-173488366 on Build GRCh38
rs538233696 Chr.1: 173488011-173488011 on Build GRCh38
rs967799365 Chr.1: 173488370-173488370 on Build GRCh38
rs1056737552 Chr.1: 173488215-173488215 on Build GRCh38
rs869206676 Chr.1: 173488410-173488411 on Build GRCh38
rs1804051 Chr.1: 173488243-173488243 on Build GRCh38
rs979073073 Chr.1: 173488378-173488378 on Build GRCh38
rs886340928 Chr.1: 173488278-173488278 on Build GRCh38
rs113281070 Chr.1: 173488420-173488420 on Build GRCh38
rs756042615 Chr.1: 173488143-173488144 on Build GRCh38
rs960074108 Chr.1: 173488342-173488342 on Build GRCh38
rs768649446 Chr.1: 173488202-173488202 on Build GRCh38
rs555997943 Chr.1: 173488105-173488105 on Build GRCh38

View Full Product Details