Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 20784933-20785488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEBL
Entrez Gene ID
10529
Gene Name
nebulette
Gene Aliases
LASP2, LNEBL
UniGene
Hs.5025
Gene Chromosome Location
Chr.10: 20779973-21174187 on Build GRCh38
Transcripts
NM_001173484.1 NM_006393.2 NM_213569.2 XM_005252342.4 XM_005252343.4 XM_005252344.4 XM_011519291.2 XM_017015467.1 XM_017015468.1

SNP Detail

SNP ID Chromosome Location
rs942009970 Chr.10: 20785014-20785014 on Build GRCh38
rs530622273 Chr.10: 20785298-20785298 on Build GRCh38
rs942080218 Chr.10: 20785265-20785265 on Build GRCh38
rs1020734361 Chr.10: 20785183-20785184 on Build GRCh38
rs990794821 Chr.10: 20784978-20784978 on Build GRCh38
rs750449390 Chr.10: 20785384-20785384 on Build GRCh38
rs924170899 Chr.10: 20785327-20785327 on Build GRCh38
rs924167156 Chr.10: 20785008-20785008 on Build GRCh38
rs771158677 Chr.10: 20785159-20785159 on Build GRCh38
rs779620965 Chr.10: 20785170-20785170 on Build GRCh38
rs946917040 Chr.10: 20784987-20784987 on Build GRCh38
rs897787283 Chr.10: 20785296-20785296 on Build GRCh38
rs534768800 Chr.10: 20785430-20785430 on Build GRCh38
rs542688075 Chr.10: 20785330-20785330 on Build GRCh38
rs1051769858 Chr.10: 20785363-20785363 on Build GRCh38
rs1007663035 Chr.10: 20785413-20785413 on Build GRCh38
rs1023601113 Chr.10: 20785211-20785211 on Build GRCh38
rs563267856 Chr.10: 20785249-20785249 on Build GRCh38
rs16921058 Chr.10: 20785091-20785091 on Build GRCh38
rs911654955 Chr.10: 20785414-20785414 on Build GRCh38
rs144336187 Chr.10: 20785177-20785177 on Build GRCh38
rs935486749 Chr.10: 20785037-20785037 on Build GRCh38
rs968033239 Chr.10: 20785239-20785239 on Build GRCh38
rs994749583 Chr.10: 20785115-20785115 on Build GRCh38
rs990827448 Chr.10: 20785196-20785196 on Build GRCh38
rs553137188 Chr.10: 20785442-20785442 on Build GRCh38
rs979710675 Chr.10: 20785254-20785254 on Build GRCh38
rs1037820356 Chr.10: 20785023-20785023 on Build GRCh38
rs537026268 Chr.10: 20785187-20785187 on Build GRCh38
rs2296612 Chr.10: 20785415-20785415 on Build GRCh38
rs935533390 Chr.10: 20785367-20785367 on Build GRCh38
rs900431656 Chr.10: 20785103-20785103 on Build GRCh38
rs909779181 Chr.10: 20785257-20785257 on Build GRCh38
rs890641209 Chr.10: 20785386-20785386 on Build GRCh38
rs12784926 Chr.10: 20785161-20785161 on Build GRCh38
rs760421425 Chr.10: 20785003-20785003 on Build GRCh38
rs147151207 Chr.10: 20784998-20784998 on Build GRCh38
rs752116744 Chr.10: 20785174-20785174 on Build GRCh38
rs866426155 Chr.10: 20785042-20785042 on Build GRCh38
rs770456228 Chr.10: 20785033-20785033 on Build GRCh38
rs756400273 Chr.10: 20785395-20785395 on Build GRCh38
rs972956751 Chr.10: 20784976-20784977 on Build GRCh38
rs916846967 Chr.10: 20784983-20784983 on Build GRCh38
rs1038104354 Chr.10: 20785279-20785279 on Build GRCh38
rs1021040022 Chr.10: 20785464-20785464 on Build GRCh38
rs548445384 Chr.10: 20785094-20785094 on Build GRCh38
rs890180711 Chr.10: 20785059-20785059 on Build GRCh38
rs772579104 Chr.10: 20784971-20784971 on Build GRCh38
rs141574784 Chr.10: 20784994-20784994 on Build GRCh38
rs929401549 Chr.10: 20785322-20785322 on Build GRCh38
rs552437129 Chr.10: 20785193-20785193 on Build GRCh38
rs140281057 Chr.10: 20785039-20785039 on Build GRCh38

View Full Product Details