Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29555025-29555624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIPSNAP1
Entrez Gene ID
8508
Gene Name
nipsnap homolog 1 (C. elegans)
Gene Aliases
-
UniGene
Hs.173878
Gene Chromosome Location
Chr.22: 29554808-29581337 on Build GRCh38
Transcripts
NM_001202502.1 NM_003634.3

SNP Detail

SNP ID Chromosome Location
rs459556 Chr.22: 29555093-29555093 on Build GRCh38
rs1038707956 Chr.22: 29555464-29555464 on Build GRCh38
rs974894111 Chr.22: 29555534-29555534 on Build GRCh38
rs6006189 Chr.22: 29555075-29555075 on Build GRCh38
rs41278849 Chr.22: 29555177-29555177 on Build GRCh38
rs565768950 Chr.22: 29555333-29555333 on Build GRCh38
rs966305429 Chr.22: 29555419-29555419 on Build GRCh38
rs555762565 Chr.22: 29555598-29555599 on Build GRCh38
rs1016160437 Chr.22: 29555241-29555241 on Build GRCh38
rs1022964103 Chr.22: 29555292-29555292 on Build GRCh38
rs779055796 Chr.22: 29555535-29555535 on Build GRCh38
rs145661949 Chr.22: 29555147-29555148 on Build GRCh38
rs1048816431 Chr.22: 29555195-29555195 on Build GRCh38
rs1045615694 Chr.22: 29555245-29555245 on Build GRCh38
rs965120985 Chr.22: 29555144-29555144 on Build GRCh38
rs952265233 Chr.22: 29555537-29555537 on Build GRCh38
rs538950525 Chr.22: 29555401-29555401 on Build GRCh38
rs561052524 Chr.22: 29555291-29555291 on Build GRCh38
rs867887669 Chr.22: 29555140-29555140 on Build GRCh38
rs552914852 Chr.22: 29555205-29555205 on Build GRCh38
rs13055175 Chr.22: 29555271-29555271 on Build GRCh38
rs531656515 Chr.22: 29555306-29555306 on Build GRCh38
rs189398707 Chr.22: 29555221-29555221 on Build GRCh38
rs1004820946 Chr.22: 29555231-29555231 on Build GRCh38
rs751504064 Chr.22: 29555346-29555346 on Build GRCh38
rs970582652 Chr.22: 29555293-29555293 on Build GRCh38
rs560570584 Chr.22: 29555112-29555112 on Build GRCh38
rs769603370 Chr.22: 29555458-29555458 on Build GRCh38
rs907168776 Chr.22: 29555301-29555301 on Build GRCh38
rs907651669 Chr.22: 29555552-29555552 on Build GRCh38
rs976416197 Chr.22: 29555152-29555152 on Build GRCh38
rs71704684 Chr.22: 29555221-29555223 on Build GRCh38
rs930416146 Chr.22: 29555170-29555170 on Build GRCh38
rs1045397881 Chr.22: 29555579-29555579 on Build GRCh38
rs1055583729 Chr.22: 29555307-29555307 on Build GRCh38
rs1011386861 Chr.22: 29555326-29555326 on Build GRCh38
rs944847565 Chr.22: 29555141-29555141 on Build GRCh38
rs548810610 Chr.22: 29555217-29555217 on Build GRCh38
rs984907761 Chr.22: 29555551-29555551 on Build GRCh38
rs745614262 Chr.22: 29555402-29555402 on Build GRCh38
rs974944871 Chr.22: 29555143-29555143 on Build GRCh38
rs748365576 Chr.22: 29555583-29555583 on Build GRCh38
rs571084178 Chr.22: 29555386-29555386 on Build GRCh38
rs995467276 Chr.22: 29555247-29555247 on Build GRCh38
rs9613953 Chr.22: 29555278-29555278 on Build GRCh38
rs796769017 Chr.22: 29555215-29555217 on Build GRCh38
rs370620039 Chr.22: 29555294-29555294 on Build GRCh38
rs1022690788 Chr.22: 29555077-29555077 on Build GRCh38
rs377427788 Chr.22: 29555128-29555128 on Build GRCh38
rs940359309 Chr.22: 29555571-29555571 on Build GRCh38
rs547357686 Chr.22: 29555576-29555576 on Build GRCh38
rs892984262 Chr.22: 29555316-29555316 on Build GRCh38
rs757964086 Chr.22: 29555319-29555319 on Build GRCh38
rs13054859 Chr.22: 29555196-29555196 on Build GRCh38
rs959746638 Chr.22: 29555052-29555052 on Build GRCh38
rs978298514 Chr.22: 29555130-29555130 on Build GRCh38
rs565707540 Chr.22: 29555587-29555587 on Build GRCh38
rs900286033 Chr.22: 29555202-29555202 on Build GRCh38

View Full Product Details