Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 56385642-56386150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMFR
Entrez Gene ID
267
Gene Name
autocrine motility factor receptor
Gene Aliases
GP78, RNF45
UniGene
Hs.295137
Gene Chromosome Location
Chr.16: 56361452-56425538 on Build GRCh38
Transcripts
NM_001144.5 NM_001323511.1 NM_001323512.1 XM_005255890.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4061333 c.1380+1G>A p.? Chr.16: 56385918-56385918 on Build GRCh38
5030936 c.1373A>G p.N458S Chr.16: 56385926-56385926 on Build GRCh38
6599188 c.1301C>T p.P434L Chr.16: 56385998-56385998 on Build GRCh38
6599187 c.1283G>A p.R428Q Chr.16: 56386016-56386016 on Build GRCh38
971660 c.1378A>C p.M460L Chr.16: 56385921-56385921 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs182529055 Chr.16: 56385847-56385847 on Build GRCh38
rs530527145 Chr.16: 56385754-56385754 on Build GRCh38
rs370219059 Chr.16: 56385921-56385921 on Build GRCh38
rs142197069 Chr.16: 56385988-56385988 on Build GRCh38
rs1050746366 Chr.16: 56385807-56385807 on Build GRCh38
rs187528539 Chr.16: 56385889-56385889 on Build GRCh38
rs779957946 Chr.16: 56385886-56385886 on Build GRCh38
rs766901764 Chr.16: 56385955-56385955 on Build GRCh38
rs1012281776 Chr.16: 56386024-56386024 on Build GRCh38
rs941969245 Chr.16: 56385858-56385858 on Build GRCh38
rs780797126 Chr.16: 56385869-56385869 on Build GRCh38
rs372760626 Chr.16: 56386026-56386026 on Build GRCh38
rs760101716 Chr.16: 56386060-56386060 on Build GRCh38
rs756403470 Chr.16: 56385873-56385875 on Build GRCh38
rs909143312 Chr.16: 56385842-56385842 on Build GRCh38
rs766164120 Chr.16: 56386098-56386098 on Build GRCh38
rs749300282 Chr.16: 56386035-56386035 on Build GRCh38
rs772504579 Chr.16: 56385852-56385852 on Build GRCh38
rs773233341 Chr.16: 56386045-56386045 on Build GRCh38
rs562463818 Chr.16: 56385818-56385818 on Build GRCh38
rs762843736 Chr.16: 56386009-56386009 on Build GRCh38
rs202204058 Chr.16: 56385909-56385909 on Build GRCh38
rs761117811 Chr.16: 56385954-56385954 on Build GRCh38
rs555467705 Chr.16: 56385715-56385715 on Build GRCh38
rs376135211 Chr.16: 56385985-56385985 on Build GRCh38
rs200380370 Chr.16: 56385965-56385965 on Build GRCh38
rs777108965 Chr.16: 56386057-56386057 on Build GRCh38
rs1053726446 Chr.16: 56386111-56386111 on Build GRCh38
rs544545869 Chr.16: 56385681-56385681 on Build GRCh38
rs749104432 Chr.16: 56385888-56385888 on Build GRCh38
rs751376369 Chr.16: 56386116-56386116 on Build GRCh38
rs957744536 Chr.16: 56385693-56385693 on Build GRCh38
rs745778867 Chr.16: 56385878-56385878 on Build GRCh38
rs2432540 Chr.16: 56385716-56385716 on Build GRCh38
rs975864622 Chr.16: 56385755-56385755 on Build GRCh38
rs953132661 Chr.16: 56386099-56386099 on Build GRCh38
rs922898604 Chr.16: 56385760-56385760 on Build GRCh38
rs754134863 Chr.16: 56385961-56385961 on Build GRCh38
rs756983668 Chr.16: 56385989-56385989 on Build GRCh38
rs778804937 Chr.16: 56386040-56386040 on Build GRCh38
rs1034794500 Chr.16: 56385982-56385982 on Build GRCh38
rs191248857 Chr.16: 56385689-56385689 on Build GRCh38
rs549904887 Chr.16: 56386124-56386124 on Build GRCh38
rs967117781 Chr.16: 56385717-56385717 on Build GRCh38
rs561005231 Chr.16: 56386074-56386074 on Build GRCh38
rs761930447 Chr.16: 56385805-56385805 on Build GRCh38
rs139530029 Chr.16: 56385950-56385950 on Build GRCh38
rs533227435 Chr.16: 56386021-56386021 on Build GRCh38
rs377202918 Chr.16: 56386043-56386043 on Build GRCh38
rs1001558731 Chr.16: 56385981-56385981 on Build GRCh38
rs569280705 Chr.16: 56386047-56386047 on Build GRCh38
rs768334761 Chr.16: 56386038-56386038 on Build GRCh38
rs544182112 Chr.16: 56385765-56385765 on Build GRCh38
rs765497157 Chr.16: 56386072-56386072 on Build GRCh38
rs140819075 Chr.16: 56386075-56386075 on Build GRCh38
rs769875610 Chr.16: 56385880-56385880 on Build GRCh38
rs146293237 Chr.16: 56385949-56385949 on Build GRCh38
rs748291710 Chr.16: 56386042-56386042 on Build GRCh38
rs201017867 Chr.16: 56386058-56386058 on Build GRCh38
rs756051344 Chr.16: 56386018-56386018 on Build GRCh38
rs369549246 Chr.16: 56385997-56385997 on Build GRCh38
rs762158280 Chr.16: 56385926-56385926 on Build GRCh38

View Full Product Details