Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 127524717-127525087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRRC1
Entrez Gene ID
133619
Gene Name
proline rich coiled-coil 1
Gene Aliases
-
UniGene
Hs.483259
Gene Chromosome Location
Chr.5: 127517609-127555089 on Build GRCh38
Transcripts
NM_001286808.1 NM_130809.4 XM_006714529.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4718129 c.358C>A p.L120I Chr.5: 127524785-127524785 on Build GRCh38
4718128 c.358C>A p.L120I Chr.5: 127524785-127524785 on Build GRCh38
6449920 c.307G>A p.G103S Chr.5: 127524734-127524734 on Build GRCh38
6284098 c.308G>A p.G103D Chr.5: 127524735-127524735 on Build GRCh38
1287415 c.466C>T p.P156S Chr.5: 127524893-127524893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765228149 Chr.5: 127524893-127524893 on Build GRCh38
rs764658417 Chr.5: 127524948-127524948 on Build GRCh38
rs61641410 Chr.5: 127524998-127524998 on Build GRCh38
rs979555429 Chr.5: 127524959-127524959 on Build GRCh38
rs112986457 Chr.5: 127524817-127524817 on Build GRCh38
rs148088843 Chr.5: 127524801-127524801 on Build GRCh38
rs776990478 Chr.5: 127524795-127524795 on Build GRCh38
rs367829860 Chr.5: 127524805-127524805 on Build GRCh38
rs767356076 Chr.5: 127524812-127524812 on Build GRCh38
rs191579681 Chr.5: 127524844-127524844 on Build GRCh38
rs767875561 Chr.5: 127524919-127524919 on Build GRCh38
rs750274879 Chr.5: 127524820-127524820 on Build GRCh38
rs747368545 Chr.5: 127524784-127524784 on Build GRCh38
rs753776557 Chr.5: 127524931-127524931 on Build GRCh38
rs943308431 Chr.5: 127525045-127525045 on Build GRCh38
rs766334285 Chr.5: 127524929-127524929 on Build GRCh38
rs187978100 Chr.5: 127525002-127525002 on Build GRCh38
rs375478806 Chr.5: 127524848-127524848 on Build GRCh38
rs768799240 Chr.5: 127524877-127524877 on Build GRCh38
rs143578156 Chr.5: 127524800-127524800 on Build GRCh38
rs149336967 Chr.5: 127524797-127524797 on Build GRCh38
rs746242728 Chr.5: 127524971-127524971 on Build GRCh38
rs754990463 Chr.5: 127524862-127524862 on Build GRCh38
rs779577581 Chr.5: 127524756-127524756 on Build GRCh38
rs762087830 Chr.5: 127524897-127524897 on Build GRCh38
rs143589506 Chr.5: 127524870-127524870 on Build GRCh38
rs756009873 Chr.5: 127524748-127524748 on Build GRCh38
rs750330055 Chr.5: 127524745-127524745 on Build GRCh38
rs949314167 Chr.5: 127524798-127524798 on Build GRCh38
rs754511369 Chr.5: 127524839-127524839 on Build GRCh38
rs182513400 Chr.5: 127525000-127525000 on Build GRCh38
rs773117730 Chr.5: 127524922-127524922 on Build GRCh38
rs911711377 Chr.5: 127525028-127525028 on Build GRCh38
rs752230930 Chr.5: 127524949-127524949 on Build GRCh38
rs770574755 Chr.5: 127525021-127525021 on Build GRCh38
rs774579380 Chr.5: 127524888-127524888 on Build GRCh38
rs376777554 Chr.5: 127524957-127524957 on Build GRCh38
rs753794686 Chr.5: 127524786-127524787 on Build GRCh38
rs147277611 Chr.5: 127524858-127524858 on Build GRCh38
rs770307535 Chr.5: 127524867-127524867 on Build GRCh38
rs766263227 Chr.5: 127524739-127524741 on Build GRCh38
rs374637146 Chr.5: 127524767-127524767 on Build GRCh38
rs756031565 Chr.5: 127524823-127524823 on Build GRCh38
rs749377323 Chr.5: 127524875-127524875 on Build GRCh38
rs1021184452 Chr.5: 127524898-127524898 on Build GRCh38
rs546751321 Chr.5: 127525017-127525018 on Build GRCh38
rs373618612 Chr.5: 127524757-127524757 on Build GRCh38
rs143797909 Chr.5: 127524804-127524804 on Build GRCh38
rs938078212 Chr.5: 127524773-127524773 on Build GRCh38
rs574379865 Chr.5: 127524987-127524987 on Build GRCh38
rs747738559 Chr.5: 127524846-127524846 on Build GRCh38
rs148035544 Chr.5: 127524927-127524927 on Build GRCh38
rs113631271 Chr.5: 127524786-127524786 on Build GRCh38
rs966957795 Chr.5: 127524924-127524924 on Build GRCh38
rs143091955 Chr.5: 127524882-127524882 on Build GRCh38
rs753358432 Chr.5: 127524836-127524836 on Build GRCh38
rs766276073 Chr.5: 127524830-127524830 on Build GRCh38
rs768332093 Chr.5: 127524758-127524758 on Build GRCh38
rs949389515 Chr.5: 127525048-127525048 on Build GRCh38
rs781492319 Chr.5: 127524854-127524854 on Build GRCh38
rs777445776 Chr.5: 127524965-127524965 on Build GRCh38
rs754953477 Chr.5: 127524945-127524945 on Build GRCh38
rs570625717 Chr.5: 127524743-127524743 on Build GRCh38
rs1008879105 Chr.5: 127524896-127524896 on Build GRCh38

View Full Product Details