Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43567709-43568302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF239
Entrez Gene ID
8187
Gene Name
zinc finger protein 239
Gene Aliases
HOK-2, MOK2
UniGene
Hs.25040
Gene Chromosome Location
Chr.10: 43554516-43574619 on Build GRCh38
Transcripts
NM_001099282.1 NM_001099283.1 NM_001099284.1 NM_001324347.1 NM_001324348.1 NM_001324349.1 NM_001324350.1 NM_001324351.1 NM_001324352.1 NM_001324353.1 NM_005674.2 XM_006718003.3 XM_011540232.2 XM_011540234.2 XM_011540235.2 XM_011540236.2 XM_011540237.2 XM_017016740.1

SNP Detail

SNP ID Chromosome Location
rs533433851 Chr.10: 43567931-43567931 on Build GRCh38
rs985551003 Chr.10: 43568245-43568245 on Build GRCh38
rs1001574292 Chr.10: 43567754-43567754 on Build GRCh38
rs959970818 Chr.10: 43567820-43567820 on Build GRCh38
rs935809929 Chr.10: 43568181-43568181 on Build GRCh38
rs553116869 Chr.10: 43568135-43568135 on Build GRCh38
rs1042432583 Chr.10: 43568160-43568160 on Build GRCh38
rs567572726 Chr.10: 43568109-43568109 on Build GRCh38
rs149268118 Chr.10: 43567768-43567768 on Build GRCh38
rs543793607 Chr.10: 43567825-43567825 on Build GRCh38
rs917915044 Chr.10: 43568140-43568140 on Build GRCh38
rs925387545 Chr.10: 43568174-43568174 on Build GRCh38
rs1054305430 Chr.10: 43568192-43568192 on Build GRCh38
rs996775305 Chr.10: 43568223-43568223 on Build GRCh38
rs569526907 Chr.10: 43567832-43567832 on Build GRCh38
rs760876611 Chr.10: 43567983-43567983 on Build GRCh38
rs895456714 Chr.10: 43568080-43568080 on Build GRCh38
rs1053336710 Chr.10: 43567968-43567968 on Build GRCh38
rs954296665 Chr.10: 43568177-43568177 on Build GRCh38
rs944302727 Chr.10: 43567950-43567950 on Build GRCh38
rs35528392 Chr.10: 43567949-43567950 on Build GRCh38
rs963155950 Chr.10: 43567864-43567864 on Build GRCh38
rs905648371 Chr.10: 43568016-43568016 on Build GRCh38
rs565906583 Chr.10: 43567885-43567885 on Build GRCh38
rs183108134 Chr.10: 43567982-43567982 on Build GRCh38
rs144472611 Chr.10: 43568079-43568079 on Build GRCh38
rs919969461 Chr.10: 43567873-43567873 on Build GRCh38
rs187646674 Chr.10: 43567860-43567860 on Build GRCh38
rs12258131 Chr.10: 43567973-43567973 on Build GRCh38
rs148034515 Chr.10: 43568203-43568203 on Build GRCh38
rs78330278 Chr.10: 43567830-43567830 on Build GRCh38
rs1014753943 Chr.10: 43568131-43568131 on Build GRCh38
rs985906464 Chr.10: 43567926-43567926 on Build GRCh38
rs772399050 Chr.10: 43567795-43567795 on Build GRCh38
rs899763241 Chr.10: 43567871-43567871 on Build GRCh38
rs974185972 Chr.10: 43567868-43567868 on Build GRCh38
rs759608341 Chr.10: 43568265-43568265 on Build GRCh38
rs182071572 Chr.10: 43567779-43567779 on Build GRCh38
rs1028825960 Chr.10: 43568152-43568152 on Build GRCh38
rs111627988 Chr.10: 43567872-43567872 on Build GRCh38
rs903009470 Chr.10: 43567747-43567747 on Build GRCh38
rs1035359862 Chr.10: 43567772-43567772 on Build GRCh38
rs1004773969 Chr.10: 43568124-43568124 on Build GRCh38
rs899710148 Chr.10: 43568220-43568220 on Build GRCh38
rs551877286 Chr.10: 43567937-43567937 on Build GRCh38
rs535006105 Chr.10: 43568121-43568121 on Build GRCh38
rs191302714 Chr.10: 43567938-43567938 on Build GRCh38
rs776867659 Chr.10: 43568151-43568151 on Build GRCh38
rs537903829 Chr.10: 43567890-43567890 on Build GRCh38
rs766533675 Chr.10: 43568041-43568041 on Build GRCh38

View Full Product Details