Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 20768645-20769197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS4Y2
Entrez Gene ID
140032
Gene Name
ribosomal protein S4, Y-linked 2
Gene Aliases
RPS4Y2P
UniGene
Hs.367761
Gene Chromosome Location
Chr.Y: 20756068-20781032 on Build GRCh38
Transcripts
NM_001039567.2

SNP Detail

SNP ID Chromosome Location
rs758585155 Chr.Y: 20768994-20768995 on Build GRCh38
rs78109950 Chr.Y: 20769171-20769171 on Build GRCh38
rs376309524 Chr.Y: 20768834-20768834 on Build GRCh38
rs753453211 Chr.Y: 20768873-20768873 on Build GRCh38
rs758305048 Chr.Y: 20768915-20768915 on Build GRCh38
rs751302551 Chr.Y: 20768934-20768934 on Build GRCh38
rs780242103 Chr.Y: 20768970-20768970 on Build GRCh38
rs774839475 Chr.Y: 20768803-20768803 on Build GRCh38
rs773579090 Chr.Y: 20768808-20768808 on Build GRCh38
rs760246582 Chr.Y: 20768827-20768827 on Build GRCh38
rs369641476 Chr.Y: 20768797-20768797 on Build GRCh38
rs73626981 Chr.Y: 20768993-20768993 on Build GRCh38
rs372057453 Chr.Y: 20768807-20768807 on Build GRCh38
rs779447849 Chr.Y: 20768997-20768997 on Build GRCh38
rs369926175 Chr.Y: 20768872-20768872 on Build GRCh38
rs777458513 Chr.Y: 20768916-20768916 on Build GRCh38
rs756304517 Chr.Y: 20768939-20768939 on Build GRCh38
rs765996121 Chr.Y: 20768841-20768841 on Build GRCh38
rs796068040 Chr.Y: 20768903-20768903 on Build GRCh38
rs780910003 Chr.Y: 20768994-20768994 on Build GRCh38
rs769210253 Chr.Y: 20768802-20768802 on Build GRCh38
rs752962850 Chr.Y: 20768995-20768995 on Build GRCh38
rs759109719 Chr.Y: 20768877-20768877 on Build GRCh38
rs752658709 Chr.Y: 20768901-20768901 on Build GRCh38
rs748628726 Chr.Y: 20769005-20769005 on Build GRCh38
rs544793767 Chr.Y: 20768886-20768886 on Build GRCh38
rs751459297 Chr.Y: 20768723-20768723 on Build GRCh38
rs765305070 Chr.Y: 20768878-20768878 on Build GRCh38
rs778145562 Chr.Y: 20769022-20769023 on Build GRCh38
rs749440420 Chr.Y: 20768995-20768995 on Build GRCh38
rs762209594 Chr.Y: 20768805-20768805 on Build GRCh38

View Full Product Details