Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 108863291-108863803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTL7A
Entrez Gene ID
10881
Gene Name
actin like 7A
Gene Aliases
-
UniGene
Hs.123530
Gene Chromosome Location
Chr.9: 108862228-108863755 on Build GRCh38
Transcripts
NM_006687.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3902858 c.1035G>A p.K345K Chr.9: 108863357-108863357 on Build GRCh38
3902859 c.1171G>T p.V391L Chr.9: 108863493-108863493 on Build GRCh38
3902860 c.1255C>T p.R419C Chr.9: 108863577-108863577 on Build GRCh38
5875305 c.1270G>A p.E424K Chr.9: 108863592-108863592 on Build GRCh38
4163197 c.1126A>T p.K376* Chr.9: 108863448-108863448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957486229 Chr.9: 108863578-108863578 on Build GRCh38
rs781311712 Chr.9: 108863571-108863571 on Build GRCh38
rs574508377 Chr.9: 108863685-108863685 on Build GRCh38
rs751153348 Chr.9: 108863519-108863519 on Build GRCh38
rs771282721 Chr.9: 108863589-108863589 on Build GRCh38
rs781247340 Chr.9: 108863419-108863419 on Build GRCh38
rs768716390 Chr.9: 108863503-108863503 on Build GRCh38
rs912502096 Chr.9: 108863648-108863648 on Build GRCh38
rs771744553 Chr.9: 108863663-108863663 on Build GRCh38
rs767000576 Chr.9: 108863524-108863524 on Build GRCh38
rs761069418 Chr.9: 108863373-108863373 on Build GRCh38
rs763615233 Chr.9: 108863376-108863376 on Build GRCh38
rs763714192 Chr.9: 108863609-108863609 on Build GRCh38
rs557241796 Chr.9: 108863684-108863684 on Build GRCh38
rs757700013 Chr.9: 108863379-108863379 on Build GRCh38
rs113170447 Chr.9: 108863462-108863462 on Build GRCh38
rs752334307 Chr.9: 108863405-108863406 on Build GRCh38
rs776258222 Chr.9: 108863339-108863339 on Build GRCh38
rs930788420 Chr.9: 108863478-108863478 on Build GRCh38
rs766665155 Chr.9: 108863518-108863518 on Build GRCh38
rs771565075 Chr.9: 108863480-108863480 on Build GRCh38
rs775666120 Chr.9: 108863368-108863368 on Build GRCh38
rs372604597 Chr.9: 108863459-108863459 on Build GRCh38
rs1054744 Chr.9: 108863636-108863636 on Build GRCh38
rs748238839 Chr.9: 108863683-108863683 on Build GRCh38
rs775137726 Chr.9: 108863602-108863602 on Build GRCh38
rs775405375 Chr.9: 108863439-108863439 on Build GRCh38
rs773168581 Chr.9: 108863427-108863427 on Build GRCh38
rs748292929 Chr.9: 108863574-108863574 on Build GRCh38
rs559816581 Chr.9: 108863750-108863750 on Build GRCh38
rs770996962 Chr.9: 108863743-108863743 on Build GRCh38
rs774743819 Chr.9: 108863650-108863650 on Build GRCh38
rs886621305 Chr.9: 108863699-108863700 on Build GRCh38
rs752301041 Chr.9: 108863525-108863525 on Build GRCh38
rs757172980 Chr.9: 108863632-108863632 on Build GRCh38
rs762552847 Chr.9: 108863366-108863366 on Build GRCh38
rs377728306 Chr.9: 108863655-108863655 on Build GRCh38
rs1003169039 Chr.9: 108863758-108863758 on Build GRCh38
rs548785692 Chr.9: 108863517-108863517 on Build GRCh38
rs746034667 Chr.9: 108863428-108863428 on Build GRCh38
rs17851211 Chr.9: 108863567-108863567 on Build GRCh38
rs749742786 Chr.9: 108863645-108863645 on Build GRCh38
rs1042546069 Chr.9: 108863747-108863747 on Build GRCh38
rs200133779 Chr.9: 108863662-108863662 on Build GRCh38
rs774597225 Chr.9: 108863595-108863595 on Build GRCh38
rs758729154 Chr.9: 108863440-108863440 on Build GRCh38
rs759140056 Chr.9: 108863522-108863522 on Build GRCh38
rs765600389 Chr.9: 108863387-108863387 on Build GRCh38
rs769033580 Chr.9: 108863456-108863456 on Build GRCh38
rs76316936 Chr.9: 108863482-108863482 on Build GRCh38
rs114738906 Chr.9: 108863417-108863417 on Build GRCh38
rs916101607 Chr.9: 108863624-108863624 on Build GRCh38
rs371393958 Chr.9: 108863405-108863405 on Build GRCh38
rs142813052 Chr.9: 108863341-108863341 on Build GRCh38
rs756119180 Chr.9: 108863640-108863640 on Build GRCh38
rs770269864 Chr.9: 108863577-108863577 on Build GRCh38
rs373429002 Chr.9: 108863597-108863597 on Build GRCh38
rs750011882 Chr.9: 108863390-108863390 on Build GRCh38
rs371023600 Chr.9: 108863635-108863635 on Build GRCh38
rs773561113 Chr.9: 108863516-108863516 on Build GRCh38
rs760728125 Chr.9: 108863393-108863394 on Build GRCh38
rs7872077 Chr.9: 108863340-108863340 on Build GRCh38
rs775229445 Chr.9: 108863672-108863672 on Build GRCh38
rs755231009 Chr.9: 108863712-108863712 on Build GRCh38
rs770324864 Chr.9: 108863514-108863514 on Build GRCh38
rs1035904912 Chr.9: 108863550-108863550 on Build GRCh38
rs142768089 Chr.9: 108863493-108863493 on Build GRCh38
rs201781647 Chr.9: 108863673-108863673 on Build GRCh38
rs747214131 Chr.9: 108863558-108863558 on Build GRCh38
rs777927399 Chr.9: 108863526-108863526 on Build GRCh38
rs138636206 Chr.9: 108863425-108863425 on Build GRCh38
rs754152142 Chr.9: 108863377-108863377 on Build GRCh38
rs757063598 Chr.9: 108863535-108863535 on Build GRCh38
rs768562040 Chr.9: 108863608-108863608 on Build GRCh38
rs748383059 Chr.9: 108863639-108863639 on Build GRCh38
rs1034679027 Chr.9: 108863761-108863761 on Build GRCh38
rs902147504 Chr.9: 108863749-108863749 on Build GRCh38
rs768784361 Chr.9: 108863564-108863564 on Build GRCh38
rs762561313 Chr.9: 108863464-108863464 on Build GRCh38
rs750810632 Chr.9: 108863463-108863463 on Build GRCh38
rs147410710 Chr.9: 108863483-108863483 on Build GRCh38
rs781449366 Chr.9: 108863638-108863638 on Build GRCh38
rs989169864 Chr.9: 108863332-108863332 on Build GRCh38
rs746569380 Chr.9: 108863408-108863408 on Build GRCh38
rs762260566 Chr.9: 108863513-108863513 on Build GRCh38
rs776605955 Chr.9: 108863512-108863512 on Build GRCh38
rs780160013 Chr.9: 108863504-108863505 on Build GRCh38
rs763658734 Chr.9: 108863677-108863677 on Build GRCh38
rs1040944718 Chr.9: 108863647-108863647 on Build GRCh38
rs200711916 Chr.9: 108863469-108863469 on Build GRCh38
rs369257453 Chr.9: 108863498-108863498 on Build GRCh38
rs56031956 Chr.9: 108863349-108863349 on Build GRCh38
rs184507726 Chr.9: 108863475-108863475 on Build GRCh38
rs534593253 Chr.9: 108863598-108863598 on Build GRCh38
rs759952745 Chr.9: 108863386-108863387 on Build GRCh38
rs778577594 Chr.9: 108863554-108863554 on Build GRCh38
rs542842157 Chr.9: 108863701-108863701 on Build GRCh38
rs760323674 Chr.9: 108863603-108863603 on Build GRCh38
rs745634839 Chr.9: 108863334-108863334 on Build GRCh38
rs151067634 Chr.9: 108863529-108863529 on Build GRCh38
rs777858496 Chr.9: 108863642-108863642 on Build GRCh38
rs751018299 Chr.9: 108863474-108863474 on Build GRCh38
rs567391469 Chr.9: 108863582-108863582 on Build GRCh38
rs912226594 Chr.9: 108863454-108863454 on Build GRCh38
rs779515458 Chr.9: 108863406-108863406 on Build GRCh38
rs749139340 Chr.9: 108863492-108863492 on Build GRCh38
rs1001934172 Chr.9: 108863732-108863732 on Build GRCh38
rs772475262 Chr.9: 108863438-108863438 on Build GRCh38
rs4978370 Chr.9: 108863633-108863633 on Build GRCh38
rs749799893 Chr.9: 108863588-108863588 on Build GRCh38
rs746881341 Chr.9: 108863316-108863316 on Build GRCh38
rs916003118 Chr.9: 108863396-108863396 on Build GRCh38
rs753762911 Chr.9: 108863625-108863625 on Build GRCh38
rs1048387362 Chr.9: 108863416-108863416 on Build GRCh38
rs368815599 Chr.9: 108863367-108863367 on Build GRCh38

View Full Product Details