Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132877921-132878429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFF4
Entrez Gene ID
27125
Gene Name
AF4/FMR2 family member 4
Gene Aliases
AF5Q31, CHOPS, MCEF
UniGene
Hs.519313
Gene Chromosome Location
Chr.5: 132875379-132963662 on Build GRCh38
Transcripts
NM_014423.3 XM_005271963.4 XM_005271964.4 XM_006714587.3

SNP Detail

SNP ID Chromosome Location
rs979006078 Chr.5: 132878396-132878396 on Build GRCh38
rs576870553 Chr.5: 132878062-132878062 on Build GRCh38
rs888705237 Chr.5: 132878230-132878230 on Build GRCh38
rs984576321 Chr.5: 132878394-132878394 on Build GRCh38
rs965705875 Chr.5: 132878334-132878334 on Build GRCh38
rs911598491 Chr.5: 132878032-132878032 on Build GRCh38
rs192880053 Chr.5: 132877985-132877985 on Build GRCh38
rs969893703 Chr.5: 132878379-132878379 on Build GRCh38
rs776896255 Chr.5: 132878206-132878206 on Build GRCh38
rs562491593 Chr.5: 132878274-132878274 on Build GRCh38
rs957599154 Chr.5: 132878257-132878257 on Build GRCh38
rs531224152 Chr.5: 132878278-132878278 on Build GRCh38
rs142128818 Chr.5: 132877963-132877963 on Build GRCh38
rs912929097 Chr.5: 132877945-132877945 on Build GRCh38
rs574654858 Chr.5: 132878002-132878002 on Build GRCh38
rs1001682653 Chr.5: 132878239-132878239 on Build GRCh38
rs747164115 Chr.5: 132877962-132877962 on Build GRCh38
rs530193196 Chr.5: 132878347-132878347 on Build GRCh38
rs953155744 Chr.5: 132878373-132878373 on Build GRCh38
rs897194381 Chr.5: 132878152-132878152 on Build GRCh38
rs188630185 Chr.5: 132878044-132878044 on Build GRCh38
rs1037433304 Chr.5: 132878108-132878108 on Build GRCh38
rs559386205 Chr.5: 132878204-132878204 on Build GRCh38
rs561250237 Chr.5: 132878309-132878309 on Build GRCh38
rs925880098 Chr.5: 132877978-132877978 on Build GRCh38
rs184603649 Chr.5: 132878031-132878031 on Build GRCh38
rs151175527 Chr.5: 132878299-132878299 on Build GRCh38
rs1033204841 Chr.5: 132878253-132878253 on Build GRCh38
rs944419243 Chr.5: 132878033-132878033 on Build GRCh38
rs769970194 Chr.5: 132878338-132878338 on Build GRCh38
rs796366240 Chr.5: 132878036-132878036 on Build GRCh38
rs905962742 Chr.5: 132878232-132878232 on Build GRCh38
rs1014280753 Chr.5: 132878283-132878283 on Build GRCh38
rs1045784246 Chr.5: 132878205-132878205 on Build GRCh38
rs42540 Chr.5: 132878263-132878263 on Build GRCh38
rs899971080 Chr.5: 132878072-132878072 on Build GRCh38
rs545610637 Chr.5: 132878093-132878093 on Build GRCh38
rs1048899409 Chr.5: 132878011-132878011 on Build GRCh38
rs966183865 Chr.5: 132877956-132877956 on Build GRCh38
rs909115671 Chr.5: 132878020-132878020 on Build GRCh38
rs538124600 Chr.5: 132877943-132877943 on Build GRCh38
rs771152519 Chr.5: 132878201-132878201 on Build GRCh38

View Full Product Details