Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 21816908-21817457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH12
Entrez Gene ID
1010
Gene Name
cadherin 12
Gene Aliases
CDHB
UniGene
Hs.113684
Gene Chromosome Location
Chr.5: 21750191-22853622 on Build GRCh38
Transcripts
NM_001317227.1 NM_001317228.1 NM_004061.4 XM_011513927.2 XM_017008920.1 XM_017008921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3975483 c.893T>G p.F298C Chr.5: 21817054-21817054 on Build GRCh38
3615172 c.982G>A p.G328R Chr.5: 21816965-21816965 on Build GRCh38
3975482 c.955A>C p.I319L Chr.5: 21816992-21816992 on Build GRCh38
1637643 c.940G>C p.G314R Chr.5: 21817007-21817007 on Build GRCh38
6170780 c.866G>A p.G289E Chr.5: 21817081-21817081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779713059 Chr.5: 21816942-21816942 on Build GRCh38
rs112294842 Chr.5: 21817364-21817364 on Build GRCh38
rs772425182 Chr.5: 21817137-21817137 on Build GRCh38
rs543329996 Chr.5: 21817117-21817117 on Build GRCh38
rs757175486 Chr.5: 21817013-21817013 on Build GRCh38
rs932473365 Chr.5: 21817285-21817285 on Build GRCh38
rs765658408 Chr.5: 21816995-21816998 on Build GRCh38
rs1025550735 Chr.5: 21817411-21817411 on Build GRCh38
rs951018833 Chr.5: 21817423-21817423 on Build GRCh38
rs182375489 Chr.5: 21817344-21817344 on Build GRCh38
rs779723483 Chr.5: 21817042-21817042 on Build GRCh38
rs778677354 Chr.5: 21816935-21816935 on Build GRCh38
rs571403721 Chr.5: 21817348-21817348 on Build GRCh38
rs992378540 Chr.5: 21817410-21817410 on Build GRCh38
rs369167882 Chr.5: 21816970-21816970 on Build GRCh38
rs746658599 Chr.5: 21816944-21816944 on Build GRCh38
rs191053108 Chr.5: 21817148-21817148 on Build GRCh38
rs772899019 Chr.5: 21817383-21817383 on Build GRCh38
rs1000459555 Chr.5: 21817372-21817372 on Build GRCh38
rs767189901 Chr.5: 21817132-21817132 on Build GRCh38
rs941985962 Chr.5: 21817239-21817239 on Build GRCh38
rs17328673 Chr.5: 21817097-21817097 on Build GRCh38
rs967801376 Chr.5: 21817006-21817006 on Build GRCh38
rs900551934 Chr.5: 21817255-21817255 on Build GRCh38
rs983040903 Chr.5: 21817106-21817106 on Build GRCh38
rs1033416127 Chr.5: 21817020-21817020 on Build GRCh38
rs757969224 Chr.5: 21816941-21816941 on Build GRCh38
rs1051366344 Chr.5: 21817289-21817289 on Build GRCh38
rs760949752 Chr.5: 21817147-21817147 on Build GRCh38
rs910105365 Chr.5: 21817119-21817119 on Build GRCh38
rs562250740 Chr.5: 21817292-21817292 on Build GRCh38
rs891080923 Chr.5: 21817294-21817294 on Build GRCh38
rs369293815 Chr.5: 21817104-21817104 on Build GRCh38
rs540978271 Chr.5: 21816985-21816985 on Build GRCh38
rs959076673 Chr.5: 21817405-21817405 on Build GRCh38
rs768152116 Chr.5: 21816947-21816947 on Build GRCh38
rs941527022 Chr.5: 21817120-21817120 on Build GRCh38
rs1033367087 Chr.5: 21817069-21817069 on Build GRCh38
rs1009887987 Chr.5: 21817321-21817321 on Build GRCh38
rs909185500 Chr.5: 21817236-21817236 on Build GRCh38
rs150544028 Chr.5: 21816978-21816978 on Build GRCh38
rs918178865 Chr.5: 21817115-21817115 on Build GRCh38
rs754743363 Chr.5: 21817086-21817086 on Build GRCh38
rs114414442 Chr.5: 21817138-21817138 on Build GRCh38
rs774834819 Chr.5: 21816972-21816972 on Build GRCh38
rs746602261 Chr.5: 21817077-21817077 on Build GRCh38
rs778611536 Chr.5: 21817140-21817140 on Build GRCh38
rs764095766 Chr.5: 21816992-21816992 on Build GRCh38
rs377660503 Chr.5: 21816989-21816989 on Build GRCh38
rs761924989 Chr.5: 21817174-21817174 on Build GRCh38
rs370457596 Chr.5: 21817226-21817226 on Build GRCh38
rs750312273 Chr.5: 21817183-21817183 on Build GRCh38
rs1022127732 Chr.5: 21816988-21816988 on Build GRCh38
rs765213108 Chr.5: 21817017-21817017 on Build GRCh38
rs771574374 Chr.5: 21817299-21817299 on Build GRCh38
rs376029399 Chr.5: 21817318-21817318 on Build GRCh38
rs570780292 Chr.5: 21817416-21817416 on Build GRCh38
rs776674900 Chr.5: 21817168-21817168 on Build GRCh38
rs533650180 Chr.5: 21817426-21817426 on Build GRCh38
rs776283058 Chr.5: 21816959-21816959 on Build GRCh38
rs1039148688 Chr.5: 21817253-21817253 on Build GRCh38
rs765319640 Chr.5: 21817176-21817176 on Build GRCh38
rs1051138354 Chr.5: 21817196-21817196 on Build GRCh38
rs749806867 Chr.5: 21816967-21816967 on Build GRCh38
rs372716639 Chr.5: 21816977-21816977 on Build GRCh38
rs375741531 Chr.5: 21816939-21816939 on Build GRCh38
rs774959670 Chr.5: 21817105-21817105 on Build GRCh38
rs1021034866 Chr.5: 21816990-21816990 on Build GRCh38
rs750157329 Chr.5: 21817031-21817031 on Build GRCh38
rs747709119 Chr.5: 21817100-21817100 on Build GRCh38
rs753988533 Chr.5: 21817002-21817002 on Build GRCh38
rs189208219 Chr.5: 21817381-21817381 on Build GRCh38
rs758159930 Chr.5: 21817041-21817041 on Build GRCh38

View Full Product Details