Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 93709504-93710008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM92A1
Entrez Gene ID
137392
Gene Name
family with sequence similarity 92 member A1
Gene Aliases
-
UniGene
Hs.125038
Gene Chromosome Location
Chr.8: 93700252-93732041 on Build GRCh38
Transcripts
NM_001283034.1 NM_145269.4 NR_104267.1 NR_104268.1 XM_005250787.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2790956 c.439-7T>A p.? Chr.8: 93709764-93709764 on Build GRCh38
3651806 c.499G>A p.E167K Chr.8: 93709831-93709831 on Build GRCh38
3651805 c.462T>A p.A154A Chr.8: 93709794-93709794 on Build GRCh38
1102455 c.487C>T p.R163C Chr.8: 93709819-93709819 on Build GRCh38
4791021 c.474T>A p.A158A Chr.8: 93709806-93709806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027958688 Chr.8: 93709941-93709941 on Build GRCh38
rs774504781 Chr.8: 93709820-93709820 on Build GRCh38
rs778514678 Chr.8: 93709757-93709757 on Build GRCh38
rs1037118505 Chr.8: 93709643-93709643 on Build GRCh38
rs370173587 Chr.8: 93709744-93709744 on Build GRCh38
rs989751147 Chr.8: 93709885-93709885 on Build GRCh38
rs1042540603 Chr.8: 93709701-93709701 on Build GRCh38
rs566268607 Chr.8: 93709877-93709877 on Build GRCh38
rs368652620 Chr.8: 93709810-93709810 on Build GRCh38
rs779906556 Chr.8: 93709894-93709894 on Build GRCh38
rs753791006 Chr.8: 93709748-93709748 on Build GRCh38
rs773760672 Chr.8: 93709672-93709672 on Build GRCh38
rs113840650 Chr.8: 93709866-93709866 on Build GRCh38
rs777888229 Chr.8: 93709733-93709734 on Build GRCh38
rs766226124 Chr.8: 93709640-93709640 on Build GRCh38
rs543712323 Chr.8: 93709845-93709845 on Build GRCh38
rs1024671355 Chr.8: 93709795-93709795 on Build GRCh38
rs757237814 Chr.8: 93709875-93709875 on Build GRCh38
rs759778738 Chr.8: 93709703-93709703 on Build GRCh38
rs771861329 Chr.8: 93709764-93709764 on Build GRCh38
rs758061253 Chr.8: 93709638-93709638 on Build GRCh38
rs141977697 Chr.8: 93709746-93709746 on Build GRCh38
rs143396120 Chr.8: 93709610-93709610 on Build GRCh38
rs1031411162 Chr.8: 93709864-93709864 on Build GRCh38
rs932923894 Chr.8: 93709536-93709537 on Build GRCh38
rs766454874 Chr.8: 93709688-93709688 on Build GRCh38
rs912717862 Chr.8: 93709552-93709552 on Build GRCh38
rs746193366 Chr.8: 93709925-93709926 on Build GRCh38
rs931266226 Chr.8: 93709663-93709663 on Build GRCh38
rs940209910 Chr.8: 93709563-93709563 on Build GRCh38
rs574583897 Chr.8: 93709755-93709755 on Build GRCh38
rs1034227564 Chr.8: 93709977-93709977 on Build GRCh38
rs532537380 Chr.8: 93709896-93709896 on Build GRCh38
rs750383872 Chr.8: 93709632-93709632 on Build GRCh38
rs757510053 Chr.8: 93709917-93709917 on Build GRCh38
rs759546035 Chr.8: 93709830-93709830 on Build GRCh38
rs757973150 Chr.8: 93709889-93709889 on Build GRCh38
rs758471500 Chr.8: 93709686-93709687 on Build GRCh38
rs979949210 Chr.8: 93709976-93709976 on Build GRCh38
rs541216491 Chr.8: 93709743-93709743 on Build GRCh38
rs771296198 Chr.8: 93709811-93709811 on Build GRCh38
rs572073067 Chr.8: 93709690-93709690 on Build GRCh38
rs953742606 Chr.8: 93709967-93709969 on Build GRCh38
rs147949730 Chr.8: 93709682-93709682 on Build GRCh38
rs1017378989 Chr.8: 93709825-93709825 on Build GRCh38
rs559796739 Chr.8: 93709937-93709937 on Build GRCh38
rs760510809 Chr.8: 93709862-93709862 on Build GRCh38
rs980001079 Chr.8: 93709592-93709592 on Build GRCh38
rs971211757 Chr.8: 93709868-93709868 on Build GRCh38
rs751760080 Chr.8: 93709880-93709881 on Build GRCh38
rs987054154 Chr.8: 93709974-93709974 on Build GRCh38
rs935672073 Chr.8: 93709636-93709636 on Build GRCh38
rs994679806 Chr.8: 93709857-93709857 on Build GRCh38
rs779782028 Chr.8: 93709765-93709765 on Build GRCh38
rs753534077 Chr.8: 93709870-93709870 on Build GRCh38
rs748586181 Chr.8: 93709772-93709772 on Build GRCh38
rs781300646 Chr.8: 93709916-93709917 on Build GRCh38
rs752494545 Chr.8: 93709725-93709725 on Build GRCh38
rs926926863 Chr.8: 93709614-93709614 on Build GRCh38
rs547948762 Chr.8: 93709745-93709745 on Build GRCh38
rs745313526 Chr.8: 93709759-93709759 on Build GRCh38
rs770264980 Chr.8: 93709778-93709778 on Build GRCh38
rs752903152 Chr.8: 93709854-93709854 on Build GRCh38
rs765839780 Chr.8: 93709667-93709667 on Build GRCh38
rs749356557 Chr.8: 93709887-93709887 on Build GRCh38
rs898617519 Chr.8: 93709662-93709662 on Build GRCh38
rs987346028 Chr.8: 93709551-93709552 on Build GRCh38
rs538261132 Chr.8: 93709659-93709659 on Build GRCh38
rs768386653 Chr.8: 93709921-93709921 on Build GRCh38
rs778655068 Chr.8: 93709876-93709876 on Build GRCh38
rs552633735 Chr.8: 93709930-93709930 on Build GRCh38
rs767670231 Chr.8: 93709710-93709710 on Build GRCh38

View Full Product Details