Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 8055051-8055548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA3
Entrez Gene ID
2625
Gene Name
GATA binding protein 3
Gene Aliases
HDR, HDRS
UniGene
Hs.524134
Gene Chromosome Location
Chr.10: 8045421-8075201 on Build GRCh38
Transcripts
NM_001002295.1 NM_002051.2 XM_005252442.2 XM_005252443.4

SNP Detail

SNP ID Chromosome Location
rs922603588 Chr.10: 8055473-8055473 on Build GRCh38
rs35971003 Chr.10: 8055392-8055392 on Build GRCh38
rs982156414 Chr.10: 8055164-8055164 on Build GRCh38
rs932631467 Chr.10: 8055474-8055474 on Build GRCh38
rs1005684248 Chr.10: 8055507-8055507 on Build GRCh38
rs888623765 Chr.10: 8055504-8055504 on Build GRCh38
rs917635399 Chr.10: 8055227-8055227 on Build GRCh38
rs1020679544 Chr.10: 8055144-8055144 on Build GRCh38
rs892465866 Chr.10: 8055143-8055143 on Build GRCh38
rs1041429196 Chr.10: 8055266-8055266 on Build GRCh38
rs930443733 Chr.10: 8055212-8055212 on Build GRCh38
rs991151861 Chr.10: 8055367-8055367 on Build GRCh38
rs971700489 Chr.10: 8055400-8055400 on Build GRCh38
rs917560892 Chr.10: 8055404-8055404 on Build GRCh38
rs940330001 Chr.10: 8055219-8055219 on Build GRCh38
rs940081068 Chr.10: 8055449-8055449 on Build GRCh38
rs543137882 Chr.10: 8055178-8055178 on Build GRCh38
rs887117098 Chr.10: 8055079-8055079 on Build GRCh38
rs1041316077 Chr.10: 8055098-8055098 on Build GRCh38
rs886047298 Chr.10: 8055369-8055369 on Build GRCh38
rs894033689 Chr.10: 8055478-8055478 on Build GRCh38
rs1010670508 Chr.10: 8055130-8055130 on Build GRCh38
rs901466431 Chr.10: 8055268-8055268 on Build GRCh38
rs996978916 Chr.10: 8055484-8055484 on Build GRCh38
rs1009485452 Chr.10: 8055336-8055336 on Build GRCh38
rs920420141 Chr.10: 8055201-8055201 on Build GRCh38
rs936476604 Chr.10: 8055429-8055429 on Build GRCh38
rs1042562995 Chr.10: 8055495-8055495 on Build GRCh38
rs1009643744 Chr.10: 8055154-8055154 on Build GRCh38
rs1015818820 Chr.10: 8055389-8055389 on Build GRCh38
rs771883196 Chr.10: 8055291-8055291 on Build GRCh38
rs34628951 Chr.10: 8055305-8055305 on Build GRCh38
rs1049977228 Chr.10: 8055276-8055276 on Build GRCh38
rs957809117 Chr.10: 8055120-8055120 on Build GRCh38
rs946897270 Chr.10: 8055479-8055479 on Build GRCh38
rs1035094141 Chr.10: 8055365-8055365 on Build GRCh38
rs11567885 Chr.10: 8055519-8055519 on Build GRCh38
rs562705113 Chr.10: 8055200-8055200 on Build GRCh38
rs138512915 Chr.10: 8055297-8055297 on Build GRCh38
rs745703902 Chr.10: 8055187-8055187 on Build GRCh38
rs959351374 Chr.10: 8055366-8055366 on Build GRCh38
rs892363056 Chr.10: 8055434-8055434 on Build GRCh38
rs945389432 Chr.10: 8055452-8055452 on Build GRCh38
rs886047297 Chr.10: 8055276-8055276 on Build GRCh38
rs1050262337 Chr.10: 8055503-8055503 on Build GRCh38
rs1027489498 Chr.10: 8055390-8055390 on Build GRCh38
rs902252286 Chr.10: 8055338-8055338 on Build GRCh38
rs936632170 Chr.10: 8055269-8055269 on Build GRCh38
rs984404346 Chr.10: 8055384-8055384 on Build GRCh38
rs1011056435 Chr.10: 8055300-8055300 on Build GRCh38
rs10905277 Chr.10: 8055405-8055405 on Build GRCh38
rs1028908661 Chr.10: 8055111-8055111 on Build GRCh38
rs901391483 Chr.10: 8055103-8055103 on Build GRCh38
rs551025607 Chr.10: 8055431-8055431 on Build GRCh38
rs905081288 Chr.10: 8055122-8055122 on Build GRCh38
rs11567884 Chr.10: 8055097-8055097 on Build GRCh38
rs976814818 Chr.10: 8055465-8055465 on Build GRCh38
rs886047299 Chr.10: 8055508-8055508 on Build GRCh38
rs927932954 Chr.10: 8055177-8055177 on Build GRCh38
rs1000725339 Chr.10: 8055128-8055128 on Build GRCh38
rs926513282 Chr.10: 8055416-8055416 on Build GRCh38
rs965934407 Chr.10: 8055184-8055184 on Build GRCh38
rs997501872 Chr.10: 8055274-8055274 on Build GRCh38
rs908659563 Chr.10: 8055408-8055408 on Build GRCh38
rs1048465824 Chr.10: 8055215-8055215 on Build GRCh38
rs1003235315 Chr.10: 8055355-8055355 on Build GRCh38
rs368662863 Chr.10: 8055371-8055371 on Build GRCh38
rs1036109987 Chr.10: 8055090-8055090 on Build GRCh38
rs971815020 Chr.10: 8055221-8055221 on Build GRCh38
rs940036481 Chr.10: 8055239-8055239 on Build GRCh38
rs997052361 Chr.10: 8055108-8055108 on Build GRCh38
rs902304650 Chr.10: 8055508-8055508 on Build GRCh38
rs952853061 Chr.10: 8055381-8055381 on Build GRCh38
rs953019356 Chr.10: 8055209-8055209 on Build GRCh38
rs966384873 Chr.10: 8055157-8055157 on Build GRCh38
rs1044934934 Chr.10: 8055255-8055255 on Build GRCh38
rs1050026072 Chr.10: 8055476-8055476 on Build GRCh38
rs908607142 Chr.10: 8055234-8055234 on Build GRCh38
rs370853904 Chr.10: 8055280-8055280 on Build GRCh38

View Full Product Details