Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 83639192-83639750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAPLN1
Entrez Gene ID
1404
Gene Name
hyaluronan and proteoglycan link protein 1
Gene Aliases
CRT1, CRTL1
UniGene
Hs.2799
Gene Chromosome Location
Chr.5: 83638198-83721210 on Build GRCh38
Transcripts
NM_001884.3 XM_011543168.2 XM_017009051.1 XM_017009052.1 XM_017009053.1 XM_017009054.1

SNP Detail

SNP ID Chromosome Location
rs559249337 Chr.5: 83639640-83639640 on Build GRCh38
rs950071756 Chr.5: 83639458-83639458 on Build GRCh38
rs181814510 Chr.5: 83639627-83639627 on Build GRCh38
rs192011788 Chr.5: 83639508-83639508 on Build GRCh38
rs961536753 Chr.5: 83639649-83639649 on Build GRCh38
rs1028389452 Chr.5: 83639396-83639396 on Build GRCh38
rs780553610 Chr.5: 83639456-83639456 on Build GRCh38
rs965056759 Chr.5: 83639376-83639376 on Build GRCh38
rs552764671 Chr.5: 83639725-83639725 on Build GRCh38
rs1049433958 Chr.5: 83639295-83639295 on Build GRCh38
rs535512222 Chr.5: 83639391-83639391 on Build GRCh38
rs530525221 Chr.5: 83639714-83639714 on Build GRCh38
rs187955793 Chr.5: 83639267-83639267 on Build GRCh38
rs377694525 Chr.5: 83639488-83639488 on Build GRCh38
rs1044884773 Chr.5: 83639282-83639283 on Build GRCh38
rs760014372 Chr.5: 83639623-83639623 on Build GRCh38
rs574358756 Chr.5: 83639664-83639664 on Build GRCh38
rs954290961 Chr.5: 83639611-83639611 on Build GRCh38
rs186214955 Chr.5: 83639459-83639459 on Build GRCh38
rs879107708 Chr.5: 83639284-83639284 on Build GRCh38
rs568466038 Chr.5: 83639292-83639292 on Build GRCh38
rs998499441 Chr.5: 83639539-83639539 on Build GRCh38
rs1032202512 Chr.5: 83639293-83639293 on Build GRCh38
rs971648684 Chr.5: 83639681-83639681 on Build GRCh38
rs1002588745 Chr.5: 83639450-83639450 on Build GRCh38
rs769301162 Chr.5: 83639541-83639541 on Build GRCh38
rs914078485 Chr.5: 83639253-83639253 on Build GRCh38
rs1024158172 Chr.5: 83639423-83639423 on Build GRCh38
rs540850485 Chr.5: 83639609-83639611 on Build GRCh38
rs949874113 Chr.5: 83639710-83639710 on Build GRCh38
rs972849873 Chr.5: 83639667-83639667 on Build GRCh38
rs770577709 Chr.5: 83639434-83639434 on Build GRCh38
rs984270739 Chr.5: 83639418-83639418 on Build GRCh38
rs182020995 Chr.5: 83639436-83639436 on Build GRCh38
rs1037232889 Chr.5: 83639349-83639349 on Build GRCh38
rs1037242027 Chr.5: 83639266-83639266 on Build GRCh38
rs953841093 Chr.5: 83639579-83639579 on Build GRCh38
rs138862475 Chr.5: 83639670-83639670 on Build GRCh38
rs777882542 Chr.5: 83639306-83639306 on Build GRCh38
rs186842254 Chr.5: 83639703-83639703 on Build GRCh38
rs73133803 Chr.5: 83639489-83639489 on Build GRCh38
rs905844296 Chr.5: 83639483-83639483 on Build GRCh38
rs1005303132 Chr.5: 83639612-83639612 on Build GRCh38
rs972967529 Chr.5: 83639445-83639445 on Build GRCh38
rs896986212 Chr.5: 83639358-83639358 on Build GRCh38
rs7731588 Chr.5: 83639277-83639277 on Build GRCh38
rs901114081 Chr.5: 83639529-83639529 on Build GRCh38
rs976940245 Chr.5: 83639603-83639603 on Build GRCh38
rs966862964 Chr.5: 83639584-83639584 on Build GRCh38
rs1028024319 Chr.5: 83639549-83639549 on Build GRCh38
rs576691472 Chr.5: 83639525-83639525 on Build GRCh38
rs922885649 Chr.5: 83639276-83639276 on Build GRCh38
rs1036781113 Chr.5: 83639690-83639690 on Build GRCh38
rs537937193 Chr.5: 83639518-83639518 on Build GRCh38
rs1014031197 Chr.5: 83639486-83639486 on Build GRCh38
rs571112729 Chr.5: 83639257-83639257 on Build GRCh38
rs941058353 Chr.5: 83639686-83639686 on Build GRCh38
rs905921457 Chr.5: 83639283-83639283 on Build GRCh38
rs753216300 Chr.5: 83639709-83639709 on Build GRCh38
rs1054910670 Chr.5: 83639487-83639487 on Build GRCh38
rs953850446 Chr.5: 83639398-83639398 on Build GRCh38
rs990033151 Chr.5: 83639227-83639227 on Build GRCh38
rs1045555955 Chr.5: 83639717-83639717 on Build GRCh38
rs893643327 Chr.5: 83639307-83639307 on Build GRCh38
rs910089938 Chr.5: 83639422-83639422 on Build GRCh38
rs1020902116 Chr.5: 83639517-83639517 on Build GRCh38
rs922835413 Chr.5: 83639604-83639604 on Build GRCh38
rs941137193 Chr.5: 83639308-83639308 on Build GRCh38

View Full Product Details