Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 64858845-64859358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC37A3
Entrez Gene ID
374819
Gene Name
leucine rich repeat containing 37 member A3
Gene Aliases
LRRC37, LRRC37A
UniGene
Hs.551962
Gene Chromosome Location
Chr.17: 64854147-64966228 on Build GRCh38
Transcripts
NM_001303255.1 NM_199340.3 XM_011524776.1 XM_011524777.1 XM_011524778.2 XM_017024593.1 XM_017024594.1 XM_017024595.1 XM_017024596.1 XM_017024597.1 XM_017024598.1 XM_017024599.1 XM_017024601.1 XM_017024602.1 XM_017024603.1 XM_017024604.1 XM_017024605.1 XM_017024606.1 XM_017024607.1 XM_017024608.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6458639 c.4743_4744insA p.L1582fs*22 Chr.17: 64858844-64858845 on Build GRCh38
5943254 c.4705-3delT p.? Chr.17: 64858886-64858886 on Build GRCh38
5649993 c.4705-2_4705-1insT p.? Chr.17: 64858884-64858885 on Build GRCh38
982967 c.4714G>T p.E1572* Chr.17: 64858874-64858874 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs187137260 Chr.17: 64859128-64859128 on Build GRCh38
rs150333817 Chr.17: 64859036-64859036 on Build GRCh38
rs936504972 Chr.17: 64858878-64858878 on Build GRCh38
rs1028586199 Chr.17: 64858974-64858974 on Build GRCh38
rs778404242 Chr.17: 64858925-64858925 on Build GRCh38
rs540207138 Chr.17: 64858885-64858886 on Build GRCh38
rs779940606 Chr.17: 64858886-64858887 on Build GRCh38
rs747920344 Chr.17: 64858886-64858888 on Build GRCh38
rs199520780 Chr.17: 64858883-64858883 on Build GRCh38
rs989789418 Chr.17: 64859126-64859127 on Build GRCh38
rs756580917 Chr.17: 64858908-64858908 on Build GRCh38
rs531600994 Chr.17: 64859232-64859232 on Build GRCh38
rs200265536 Chr.17: 64858879-64858879 on Build GRCh38
rs971828809 Chr.17: 64859052-64859052 on Build GRCh38
rs536723957 Chr.17: 64858980-64858980 on Build GRCh38
rs576040214 Chr.17: 64859058-64859058 on Build GRCh38
rs945509183 Chr.17: 64858915-64858915 on Build GRCh38
rs1024675009 Chr.17: 64859055-64859055 on Build GRCh38
rs544592411 Chr.17: 64859074-64859074 on Build GRCh38
rs551784631 Chr.17: 64859203-64859203 on Build GRCh38
rs562427613 Chr.17: 64859222-64859222 on Build GRCh38
rs553195265 Chr.17: 64859003-64859003 on Build GRCh38
rs932640183 Chr.17: 64859182-64859182 on Build GRCh38
rs750678817 Chr.17: 64858879-64858879 on Build GRCh38
rs970375338 Chr.17: 64859066-64859066 on Build GRCh38
rs751213832 Chr.17: 64858892-64858892 on Build GRCh38
rs572173162 Chr.17: 64859231-64859231 on Build GRCh38
rs936574077 Chr.17: 64859103-64859103 on Build GRCh38
rs573265088 Chr.17: 64859022-64859022 on Build GRCh38
rs548209958 Chr.17: 64859293-64859293 on Build GRCh38
rs867357691 Chr.17: 64859227-64859227 on Build GRCh38
rs749612546 Chr.17: 64858931-64858931 on Build GRCh38
rs892556433 Chr.17: 64858891-64858891 on Build GRCh38
rs888663939 Chr.17: 64858994-64858994 on Build GRCh38
rs762861519 Chr.17: 64858886-64858886 on Build GRCh38
rs758491840 Chr.17: 64858884-64858885 on Build GRCh38
rs901222071 Chr.17: 64859167-64859167 on Build GRCh38
rs996839911 Chr.17: 64858964-64858965 on Build GRCh38
rs747118044 Chr.17: 64858886-64858886 on Build GRCh38
rs543026601 Chr.17: 64859274-64859274 on Build GRCh38
rs531981420 Chr.17: 64859202-64859202 on Build GRCh38
rs76813102 Chr.17: 64858898-64858898 on Build GRCh38
rs202232311 Chr.17: 64859149-64859149 on Build GRCh38
rs753160978 Chr.17: 64858905-64858905 on Build GRCh38
rs901213893 Chr.17: 64858942-64858942 on Build GRCh38
rs137980456 Chr.17: 64859039-64859039 on Build GRCh38
rs780701768 Chr.17: 64858899-64858899 on Build GRCh38
rs897357555 Chr.17: 64859326-64859326 on Build GRCh38
rs376498347 Chr.17: 64859024-64859024 on Build GRCh38
rs373642863 Chr.17: 64859204-64859204 on Build GRCh38
rs868839490 Chr.17: 64858884-64858884 on Build GRCh38
rs143994301 Chr.17: 64859099-64859099 on Build GRCh38
rs1005826981 Chr.17: 64859263-64859263 on Build GRCh38
rs888706870 Chr.17: 64859243-64859243 on Build GRCh38
rs62071396 Chr.17: 64858900-64858900 on Build GRCh38
rs540279386 Chr.17: 64859143-64859143 on Build GRCh38

View Full Product Details