Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71640672-71641176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARVELD3
Entrez Gene ID
91862
Gene Name
MARVEL domain containing 3
Gene Aliases
MARVD3, MRVLDC3
UniGene
Hs.513706
Gene Chromosome Location
Chr.16: 71626137-71641965 on Build GRCh38
Transcripts
NM_001017967.3 NM_001271329.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3511870 c.1044G>A p.W348* Chr.16: 71640838-71640838 on Build GRCh38
3511869 c.1010G>A p.R337K Chr.16: 71640804-71640804 on Build GRCh38
3511872 c.1126G>A p.V376M Chr.16: 71640920-71640920 on Build GRCh38
3511871 c.1115G>A p.G372D Chr.16: 71640909-71640909 on Build GRCh38
5929670 c.1220C>T p.S407F Chr.16: 71641014-71641014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748486817 Chr.16: 71640784-71640784 on Build GRCh38
rs751931733 Chr.16: 71640999-71640999 on Build GRCh38
rs749376284 Chr.16: 71640853-71640853 on Build GRCh38
rs201839766 Chr.16: 71640944-71640944 on Build GRCh38
rs953022395 Chr.16: 71641094-71641094 on Build GRCh38
rs758168166 Chr.16: 71640814-71640814 on Build GRCh38
rs756209782 Chr.16: 71640777-71640777 on Build GRCh38
rs2303225 Chr.16: 71640979-71640979 on Build GRCh38
rs74027272 Chr.16: 71641057-71641057 on Build GRCh38
rs751389133 Chr.16: 71640900-71640900 on Build GRCh38
rs763956465 Chr.16: 71640822-71640822 on Build GRCh38
rs751701787 Chr.16: 71640866-71640866 on Build GRCh38
rs776090935 Chr.16: 71640803-71640803 on Build GRCh38
rs774293472 Chr.16: 71640968-71640968 on Build GRCh38
rs1029258367 Chr.16: 71640726-71640726 on Build GRCh38
rs145897917 Chr.16: 71640707-71640707 on Build GRCh38
rs796529323 Chr.16: 71640802-71640803 on Build GRCh38
rs998282337 Chr.16: 71640791-71640791 on Build GRCh38
rs781508881 Chr.16: 71641002-71641002 on Build GRCh38
rs768191424 Chr.16: 71640958-71640958 on Build GRCh38
rs757452634 Chr.16: 71641071-71641071 on Build GRCh38
rs747561456 Chr.16: 71640868-71640868 on Build GRCh38
rs754605352 Chr.16: 71640729-71640729 on Build GRCh38
rs779139024 Chr.16: 71640861-71640861 on Build GRCh38
rs762880396 Chr.16: 71641061-71641061 on Build GRCh38
rs771412174 Chr.16: 71640870-71640870 on Build GRCh38
rs375582710 Chr.16: 71640895-71640895 on Build GRCh38
rs368492439 Chr.16: 71640896-71640896 on Build GRCh38
rs372021599 Chr.16: 71641076-71641076 on Build GRCh38
rs774098515 Chr.16: 71641035-71641035 on Build GRCh38
rs367583136 Chr.16: 71640973-71640973 on Build GRCh38
rs901751702 Chr.16: 71640700-71640700 on Build GRCh38
rs775173002 Chr.16: 71640890-71640890 on Build GRCh38
rs752535125 Chr.16: 71640736-71640736 on Build GRCh38
rs752823274 Chr.16: 71641060-71641060 on Build GRCh38
rs762292191 Chr.16: 71640696-71640696 on Build GRCh38
rs899601406 Chr.16: 71640731-71640731 on Build GRCh38
rs762218577 Chr.16: 71640802-71640803 on Build GRCh38
rs999101774 Chr.16: 71640859-71640859 on Build GRCh38
rs371616837 Chr.16: 71641011-71641011 on Build GRCh38
rs146595006 Chr.16: 71640829-71640829 on Build GRCh38
rs751823671 Chr.16: 71641068-71641068 on Build GRCh38
rs778616264 Chr.16: 71640733-71640733 on Build GRCh38
rs561280267 Chr.16: 71640799-71640799 on Build GRCh38
rs764806516 Chr.16: 71640888-71640888 on Build GRCh38
rs780256659 Chr.16: 71640778-71640778 on Build GRCh38
rs755037149 Chr.16: 71640910-71640910 on Build GRCh38
rs777901977 Chr.16: 71640992-71640992 on Build GRCh38
rs758329235 Chr.16: 71640737-71640737 on Build GRCh38
rs918195587 Chr.16: 71640821-71640821 on Build GRCh38
rs370061470 Chr.16: 71640805-71640805 on Build GRCh38
rs760497823 Chr.16: 71640974-71640974 on Build GRCh38
rs757227071 Chr.16: 71640831-71640831 on Build GRCh38
rs138854747 Chr.16: 71640941-71640941 on Build GRCh38
rs760111945 Chr.16: 71640722-71640722 on Build GRCh38
rs778166227 Chr.16: 71640915-71640915 on Build GRCh38
rs755190760 Chr.16: 71640875-71640875 on Build GRCh38
rs771673411 Chr.16: 71641093-71641093 on Build GRCh38
rs755160925 Chr.16: 71640857-71640857 on Build GRCh38
rs758525659 Chr.16: 71640914-71640914 on Build GRCh38
rs200707122 Chr.16: 71641023-71641023 on Build GRCh38
rs753002870 Chr.16: 71640913-71640913 on Build GRCh38
rs753050079 Chr.16: 71640948-71640948 on Build GRCh38
rs200552224 Chr.16: 71640804-71640804 on Build GRCh38
rs757332669 Chr.16: 71640902-71640902 on Build GRCh38
rs766050352 Chr.16: 71640970-71640979 on Build GRCh38
rs764933699 Chr.16: 71640810-71640810 on Build GRCh38
rs141358444 Chr.16: 71640908-71640908 on Build GRCh38
rs908621690 Chr.16: 71640892-71640892 on Build GRCh38
rs766052344 Chr.16: 71640724-71640724 on Build GRCh38
rs757563926 Chr.16: 71641000-71641000 on Build GRCh38
rs781437368 Chr.16: 71640920-71640920 on Build GRCh38
rs79422919 Chr.16: 71641036-71641036 on Build GRCh38
rs753441407 Chr.16: 71640727-71640727 on Build GRCh38
rs774271511 Chr.16: 71641143-71641143 on Build GRCh38
rs74425399 Chr.16: 71640863-71640863 on Build GRCh38
rs540095054 Chr.16: 71640907-71640907 on Build GRCh38
rs778408721 Chr.16: 71641013-71641013 on Build GRCh38
rs768437612 Chr.16: 71641033-71641033 on Build GRCh38
rs535348201 Chr.16: 71641136-71641136 on Build GRCh38
rs112608295 Chr.16: 71641003-71641003 on Build GRCh38
rs200159769 Chr.16: 71641040-71641040 on Build GRCh38
rs763254946 Chr.16: 71640947-71640963 on Build GRCh38
rs193188432 Chr.16: 71641062-71641062 on Build GRCh38
rs765406380 Chr.16: 71640976-71640976 on Build GRCh38
rs779667480 Chr.16: 71641108-71641108 on Build GRCh38
rs774636026 Chr.16: 71640783-71640783 on Build GRCh38
rs781617184 Chr.16: 71640933-71640933 on Build GRCh38
rs926787510 Chr.16: 71640860-71640860 on Build GRCh38
rs747366699 Chr.16: 71640931-71640931 on Build GRCh38
rs979591156 Chr.16: 71640785-71640785 on Build GRCh38
rs767496976 Chr.16: 71640903-71640903 on Build GRCh38
rs1051210772 Chr.16: 71640796-71640796 on Build GRCh38
rs780346547 Chr.16: 71640847-71640847 on Build GRCh38
rs746438834 Chr.16: 71641004-71641004 on Build GRCh38
rs777695867 Chr.16: 71640764-71640764 on Build GRCh38
rs752452674 Chr.16: 71640811-71640811 on Build GRCh38
rs768993397 Chr.16: 71640781-71640781 on Build GRCh38
rs767930440 Chr.16: 71641087-71641087 on Build GRCh38
rs200660505 Chr.16: 71640763-71640763 on Build GRCh38
rs1051200872 Chr.16: 71641056-71641056 on Build GRCh38
rs553658133 Chr.16: 71641138-71641138 on Build GRCh38
rs1041077153 Chr.16: 71641130-71641130 on Build GRCh38
rs762726448 Chr.16: 71640954-71640954 on Build GRCh38
rs941107898 Chr.16: 71640695-71640695 on Build GRCh38
rs903403755 Chr.16: 71640807-71640807 on Build GRCh38
rs767584918 Chr.16: 71640969-71640969 on Build GRCh38
rs766637805 Chr.16: 71640865-71640865 on Build GRCh38
rs760720819 Chr.16: 71641053-71641053 on Build GRCh38
rs1048497281 Chr.16: 71640879-71640879 on Build GRCh38
rs748681774 Chr.16: 71641029-71641029 on Build GRCh38
rs13337162 Chr.16: 71641052-71641052 on Build GRCh38
rs779033551 Chr.16: 71640911-71640911 on Build GRCh38
rs987790704 Chr.16: 71640816-71640816 on Build GRCh38
rs200621755 Chr.16: 71640978-71640978 on Build GRCh38
rs768054161 Chr.16: 71640704-71640704 on Build GRCh38
rs749560231 Chr.16: 71640779-71640779 on Build GRCh38
rs554767506 Chr.16: 71640788-71640788 on Build GRCh38
rs771428755 Chr.16: 71640932-71640932 on Build GRCh38
rs984236147 Chr.16: 71641097-71641097 on Build GRCh38
rs909532995 Chr.16: 71641016-71641016 on Build GRCh38
rs746662556 Chr.16: 71641150-71641150 on Build GRCh38
rs373575019 Chr.16: 71640787-71640787 on Build GRCh38
rs983135271 Chr.16: 71641005-71641005 on Build GRCh38
rs568001755 Chr.16: 71641063-71641063 on Build GRCh38
rs866358853 Chr.16: 71640826-71640826 on Build GRCh38
rs940123391 Chr.16: 71641115-71641115 on Build GRCh38
rs773538793 Chr.16: 71640921-71640922 on Build GRCh38
rs776187895 Chr.16: 71640876-71640876 on Build GRCh38

View Full Product Details