Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2071385-2071885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOB3A
Entrez Gene ID
126308
Gene Name
MOB kinase activator 3A
Gene Aliases
MOB1C, MOBKL2A, moblak
UniGene
Hs.744938
Gene Chromosome Location
Chr.19: 2071036-2096502 on Build GRCh38
Transcripts
NM_130807.2 XM_011527683.2

SNP Detail

SNP ID Chromosome Location
rs1021077639 Chr.19: 2071486-2071486 on Build GRCh38
rs143272659 Chr.19: 2071485-2071485 on Build GRCh38
rs962159202 Chr.19: 2071752-2071752 on Build GRCh38
rs372371476 Chr.19: 2071582-2071582 on Build GRCh38
rs991700202 Chr.19: 2071797-2071797 on Build GRCh38
rs1012196994 Chr.19: 2071472-2071472 on Build GRCh38
rs1047540838 Chr.19: 2071418-2071418 on Build GRCh38
rs192250704 Chr.19: 2071439-2071439 on Build GRCh38
rs931430388 Chr.19: 2071569-2071569 on Build GRCh38
rs565806804 Chr.19: 2071805-2071805 on Build GRCh38
rs991584326 Chr.19: 2071559-2071559 on Build GRCh38
rs1050137729 Chr.19: 2071422-2071422 on Build GRCh38
rs887114969 Chr.19: 2071421-2071421 on Build GRCh38
rs1015011303 Chr.19: 2071751-2071751 on Build GRCh38
rs1032095227 Chr.19: 2071478-2071478 on Build GRCh38
rs1040956874 Chr.19: 2071433-2071433 on Build GRCh38
rs935501968 Chr.19: 2071709-2071709 on Build GRCh38
rs373210187 Chr.19: 2071553-2071553 on Build GRCh38
rs115821202 Chr.19: 2071850-2071850 on Build GRCh38
rs1035880850 Chr.19: 2071503-2071503 on Build GRCh38
rs958982249 Chr.19: 2071791-2071791 on Build GRCh38
rs899709706 Chr.19: 2071640-2071640 on Build GRCh38
rs10420854 Chr.19: 2071687-2071687 on Build GRCh38
rs1009754293 Chr.19: 2071715-2071715 on Build GRCh38
rs1041587331 Chr.19: 2071444-2071444 on Build GRCh38
rs921257258 Chr.19: 2071858-2071858 on Build GRCh38
rs1040999135 Chr.19: 2071630-2071630 on Build GRCh38
rs902651253 Chr.19: 2071706-2071706 on Build GRCh38
rs1008586413 Chr.19: 2071432-2071432 on Build GRCh38
rs961880184 Chr.19: 2071522-2071522 on Build GRCh38
rs982869726 Chr.19: 2071593-2071593 on Build GRCh38
rs573024171 Chr.19: 2071790-2071790 on Build GRCh38
rs899678537 Chr.19: 2071437-2071437 on Build GRCh38
rs562564010 Chr.19: 2071854-2071854 on Build GRCh38
rs944426476 Chr.19: 2071680-2071680 on Build GRCh38
rs931920280 Chr.19: 2071581-2071581 on Build GRCh38
rs902694078 Chr.19: 2071446-2071447 on Build GRCh38
rs528737916 Chr.19: 2071645-2071645 on Build GRCh38
rs531087107 Chr.19: 2071519-2071519 on Build GRCh38
rs35053596 Chr.19: 2071427-2071427 on Build GRCh38
rs573042772 Chr.19: 2071410-2071410 on Build GRCh38
rs115172384 Chr.19: 2071798-2071798 on Build GRCh38
rs564854788 Chr.19: 2071446-2071446 on Build GRCh38
rs920086523 Chr.19: 2071568-2071568 on Build GRCh38
rs768015995 Chr.19: 2071692-2071692 on Build GRCh38
rs944496374 Chr.19: 2071442-2071443 on Build GRCh38
rs973225032 Chr.19: 2071765-2071765 on Build GRCh38
rs139078887 Chr.19: 2071629-2071629 on Build GRCh38
rs755543980 Chr.19: 2071644-2071644 on Build GRCh38
rs576881348 Chr.19: 2071847-2071847 on Build GRCh38
rs952898551 Chr.19: 2071807-2071807 on Build GRCh38
rs116204589 Chr.19: 2071736-2071736 on Build GRCh38
rs996656550 Chr.19: 2071458-2071458 on Build GRCh38
rs1012521772 Chr.19: 2071734-2071734 on Build GRCh38
rs982129541 Chr.19: 2071496-2071496 on Build GRCh38
rs557378084 Chr.19: 2071693-2071693 on Build GRCh38
rs571271517 Chr.19: 2071686-2071686 on Build GRCh38
rs906546718 Chr.19: 2071741-2071741 on Build GRCh38
rs72981474 Chr.19: 2071731-2071731 on Build GRCh38
rs553458581 Chr.19: 2071792-2071792 on Build GRCh38
rs1032559957 Chr.19: 2071470-2071470 on Build GRCh38
rs558908368 Chr.19: 2071846-2071846 on Build GRCh38
rs1801652 Chr.19: 2071822-2071822 on Build GRCh38
rs908567535 Chr.19: 2071607-2071607 on Build GRCh38
rs546923216 Chr.19: 2071670-2071670 on Build GRCh38
rs893524304 Chr.19: 2071483-2071483 on Build GRCh38
rs367874835 Chr.19: 2071732-2071732 on Build GRCh38
rs532181778 Chr.19: 2071448-2071448 on Build GRCh38
rs34418412 Chr.19: 2071481-2071482 on Build GRCh38
rs182169831 Chr.19: 2071457-2071457 on Build GRCh38
rs931339958 Chr.19: 2071412-2071412 on Build GRCh38
rs924359088 Chr.19: 2071855-2071855 on Build GRCh38
rs908599132 Chr.19: 2071849-2071849 on Build GRCh38
rs970400572 Chr.19: 2071494-2071494 on Build GRCh38
rs1004001509 Chr.19: 2071750-2071750 on Build GRCh38

View Full Product Details