Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2073095-2073588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOB3A
Entrez Gene ID
126308
Gene Name
MOB kinase activator 3A
Gene Aliases
MOB1C, MOBKL2A, moblak
UniGene
Hs.744938
Gene Chromosome Location
Chr.19: 2071036-2096502 on Build GRCh38
Transcripts
NM_130807.2 XM_011527683.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6573564 c.649C>T p.H217Y Chr.19: 2073400-2073400 on Build GRCh38
6228103 c.638C>T p.A213V Chr.19: 2073411-2073411 on Build GRCh38
6714073 c.640C>T p.R214W Chr.19: 2073409-2073409 on Build GRCh38
6714081 c.636C>T p.T212T Chr.19: 2073413-2073413 on Build GRCh38
3892071 c.647G>C p.C216S Chr.19: 2073402-2073402 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911423623 Chr.19: 2073268-2073268 on Build GRCh38
rs199578809 Chr.19: 2073385-2073385 on Build GRCh38
rs1039589620 Chr.19: 2073543-2073543 on Build GRCh38
rs1006436914 Chr.19: 2073313-2073313 on Build GRCh38
rs775684733 Chr.19: 2073387-2073387 on Build GRCh38
rs577341609 Chr.19: 2073236-2073236 on Build GRCh38
rs755605847 Chr.19: 2073372-2073372 on Build GRCh38
rs770729492 Chr.19: 2073377-2073377 on Build GRCh38
rs756895525 Chr.19: 2073427-2073427 on Build GRCh38
rs141783611 Chr.19: 2073408-2073408 on Build GRCh38
rs896505805 Chr.19: 2073314-2073314 on Build GRCh38
rs1012180410 Chr.19: 2073330-2073330 on Build GRCh38
rs373522907 Chr.19: 2073376-2073376 on Build GRCh38
rs560485012 Chr.19: 2073361-2073361 on Build GRCh38
rs900734213 Chr.19: 2073406-2073406 on Build GRCh38
rs778716035 Chr.19: 2073431-2073431 on Build GRCh38
rs771868773 Chr.19: 2073386-2073386 on Build GRCh38
rs774116448 Chr.19: 2073384-2073384 on Build GRCh38
rs373455207 Chr.19: 2073336-2073336 on Build GRCh38
rs2286365 Chr.19: 2073479-2073479 on Build GRCh38
rs748994428 Chr.19: 2073426-2073426 on Build GRCh38
rs193283677 Chr.19: 2073351-2073351 on Build GRCh38
rs927188621 Chr.19: 2073356-2073356 on Build GRCh38
rs557449339 Chr.19: 2073232-2073232 on Build GRCh38
rs779839540 Chr.19: 2073472-2073472 on Build GRCh38
rs553347897 Chr.19: 2073215-2073215 on Build GRCh38
rs767639622 Chr.19: 2073346-2073346 on Build GRCh38
rs150549861 Chr.19: 2073332-2073332 on Build GRCh38
rs993933954 Chr.19: 2073298-2073298 on Build GRCh38
rs776848250 Chr.19: 2073444-2073444 on Build GRCh38
rs764758411 Chr.19: 2073503-2073503 on Build GRCh38
rs369446517 Chr.19: 2073388-2073388 on Build GRCh38
rs201385305 Chr.19: 2073455-2073455 on Build GRCh38
rs775321341 Chr.19: 2073439-2073439 on Build GRCh38
rs777326092 Chr.19: 2073374-2073374 on Build GRCh38
rs146206720 Chr.19: 2073435-2073435 on Build GRCh38
rs962141586 Chr.19: 2073191-2073191 on Build GRCh38
rs139225109 Chr.19: 2073488-2073488 on Build GRCh38
rs762380169 Chr.19: 2073418-2073421 on Build GRCh38
rs368995913 Chr.19: 2073360-2073360 on Build GRCh38
rs2286364 Chr.19: 2073355-2073355 on Build GRCh38
rs1030092311 Chr.19: 2073497-2073497 on Build GRCh38
rs771850716 Chr.19: 2073438-2073438 on Build GRCh38
rs758145 Chr.19: 2073467-2073467 on Build GRCh38
rs1052866635 Chr.19: 2073169-2073169 on Build GRCh38
rs138583524 Chr.19: 2073350-2073350 on Build GRCh38
rs924132766 Chr.19: 2073302-2073302 on Build GRCh38
rs368335071 Chr.19: 2073369-2073369 on Build GRCh38
rs755588493 Chr.19: 2073424-2073424 on Build GRCh38
rs774906291 Chr.19: 2073349-2073349 on Build GRCh38
rs768208914 Chr.19: 2073429-2073429 on Build GRCh38
rs777282167 Chr.19: 2073425-2073425 on Build GRCh38
rs946911159 Chr.19: 2073273-2073273 on Build GRCh38
rs1056463719 Chr.19: 2073304-2073304 on Build GRCh38
rs370833640 Chr.19: 2073447-2073447 on Build GRCh38
rs942490152 Chr.19: 2073538-2073538 on Build GRCh38
rs535422301 Chr.19: 2073192-2073192 on Build GRCh38
rs1006428096 Chr.19: 2073159-2073159 on Build GRCh38
rs185503935 Chr.19: 2073496-2073496 on Build GRCh38
rs201863872 Chr.19: 2073443-2073443 on Build GRCh38
rs772649223 Chr.19: 2073528-2073528 on Build GRCh38
rs373193407 Chr.19: 2073413-2073413 on Build GRCh38
rs1056827033 Chr.19: 2073269-2073269 on Build GRCh38
rs375009334 Chr.19: 2073449-2073449 on Build GRCh38
rs200767082 Chr.19: 2073451-2073451 on Build GRCh38
rs375222150 Chr.19: 2073362-2073362 on Build GRCh38
rs568353329 Chr.19: 2073177-2073177 on Build GRCh38
rs752069035 Chr.19: 2073416-2073416 on Build GRCh38
rs377375695 Chr.19: 2073409-2073409 on Build GRCh38
rs111744616 Chr.19: 2073228-2073228 on Build GRCh38
rs368298669 Chr.19: 2073481-2073481 on Build GRCh38
rs536787976 Chr.19: 2073505-2073505 on Build GRCh38
rs141791372 Chr.19: 2073221-2073221 on Build GRCh38
rs762168032 Chr.19: 2073446-2073446 on Build GRCh38
rs142028177 Chr.19: 2073412-2073412 on Build GRCh38
rs1021734885 Chr.19: 2073519-2073519 on Build GRCh38
rs951932358 Chr.19: 2073142-2073142 on Build GRCh38
rs750135805 Chr.19: 2073518-2073518 on Build GRCh38
rs201344352 Chr.19: 2073364-2073364 on Build GRCh38
rs373445365 Chr.19: 2073392-2073392 on Build GRCh38
rs1044316030 Chr.19: 2073301-2073301 on Build GRCh38
rs749329827 Chr.19: 2073176-2073176 on Build GRCh38
rs768461107 Chr.19: 2073132-2073132 on Build GRCh38
rs750036745 Chr.19: 2073456-2073456 on Build GRCh38
rs934776418 Chr.19: 2073166-2073167 on Build GRCh38
rs965025835 Chr.19: 2073321-2073321 on Build GRCh38
rs1044707184 Chr.19: 2073214-2073214 on Build GRCh38
rs761245581 Chr.19: 2073502-2073502 on Build GRCh38
rs148051183 Chr.19: 2073220-2073220 on Build GRCh38
rs371831143 Chr.19: 2073506-2073506 on Build GRCh38
rs763444997 Chr.19: 2073448-2073448 on Build GRCh38
rs539070850 Chr.19: 2073482-2073482 on Build GRCh38
rs777973138 Chr.19: 2073136-2073136 on Build GRCh38
rs980106855 Chr.19: 2073529-2073529 on Build GRCh38
rs927166046 Chr.19: 2073463-2073463 on Build GRCh38
rs774576002 Chr.19: 2073303-2073303 on Build GRCh38
rs371364999 Chr.19: 2073410-2073410 on Build GRCh38
rs753312706 Chr.19: 2073452-2073452 on Build GRCh38
rs776336838 Chr.19: 2073229-2073229 on Build GRCh38
rs190504898 Chr.19: 2073549-2073549 on Build GRCh38
rs887656907 Chr.19: 2073333-2073333 on Build GRCh38
rs756948502 Chr.19: 2073454-2073454 on Build GRCh38
rs780795210 Chr.19: 2073370-2073370 on Build GRCh38

View Full Product Details