Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43029939-43030397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RND2
Entrez Gene ID
8153
Gene Name
Rho family GTPase 2
Gene Aliases
ARHN, RHO7, RhoN
UniGene
Hs.603111
Gene Chromosome Location
Chr.17: 43025241-43032041 on Build GRCh38
Transcripts
NM_005440.4 XM_011525316.1 XM_011525317.2

SNP Detail

SNP ID Chromosome Location
rs921648554 Chr.17: 43030105-43030105 on Build GRCh38
rs893903070 Chr.17: 43030328-43030328 on Build GRCh38
rs530709766 Chr.17: 43030229-43030229 on Build GRCh38
rs557788612 Chr.17: 43030089-43030089 on Build GRCh38
rs1046582435 Chr.17: 43030250-43030250 on Build GRCh38
rs903082114 Chr.17: 43030351-43030351 on Build GRCh38
rs967334919 Chr.17: 43030104-43030104 on Build GRCh38
rs116803226 Chr.17: 43030027-43030027 on Build GRCh38
rs544065735 Chr.17: 43030260-43030260 on Build GRCh38
rs1047800480 Chr.17: 43029968-43029968 on Build GRCh38
rs975169995 Chr.17: 43030142-43030142 on Build GRCh38
rs750915847 Chr.17: 43030322-43030322 on Build GRCh38
rs908793611 Chr.17: 43029975-43029975 on Build GRCh38
rs1008454943 Chr.17: 43030116-43030116 on Build GRCh38
rs905254321 Chr.17: 43029995-43029995 on Build GRCh38
rs866067844 Chr.17: 43030304-43030304 on Build GRCh38
rs906539255 Chr.17: 43030315-43030315 on Build GRCh38
rs982474221 Chr.17: 43030164-43030164 on Build GRCh38
rs138432344 Chr.17: 43030330-43030330 on Build GRCh38
rs560525130 Chr.17: 43030169-43030169 on Build GRCh38
rs376049563 Chr.17: 43030129-43030129 on Build GRCh38
rs1046569729 Chr.17: 43029981-43029981 on Build GRCh38
rs930517019 Chr.17: 43030112-43030112 on Build GRCh38
rs919592517 Chr.17: 43030146-43030146 on Build GRCh38
rs564876052 Chr.17: 43029974-43029974 on Build GRCh38
rs572848031 Chr.17: 43030123-43030123 on Build GRCh38
rs866794223 Chr.17: 43030312-43030312 on Build GRCh38
rs949547989 Chr.17: 43029980-43029980 on Build GRCh38
rs1053773928 Chr.17: 43030030-43030030 on Build GRCh38
rs893846133 Chr.17: 43030060-43030060 on Build GRCh38
rs938070027 Chr.17: 43030298-43030298 on Build GRCh38
rs757727349 Chr.17: 43030040-43030040 on Build GRCh38
rs1008176342 Chr.17: 43030373-43030373 on Build GRCh38
rs550778192 Chr.17: 43030297-43030297 on Build GRCh38
rs1042410797 Chr.17: 43030339-43030339 on Build GRCh38
rs974908035 Chr.17: 43030092-43030092 on Build GRCh38
rs554920016 Chr.17: 43030161-43030161 on Build GRCh38
rs989645608 Chr.17: 43029996-43029996 on Build GRCh38
rs926806211 Chr.17: 43030292-43030292 on Build GRCh38
rs1022589226 Chr.17: 43029999-43029999 on Build GRCh38
rs186971184 Chr.17: 43030072-43030072 on Build GRCh38

View Full Product Details