Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42560162-42560666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERHL2
Entrez Gene ID
253190
Gene Name
serine hydrolase-like 2
Gene Aliases
dJ222E13.1
UniGene
Hs.728878
Gene Chromosome Location
Chr.22: 42553857-42574382 on Build GRCh38
Transcripts
NM_001284334.1 NM_014509.4 NR_104300.1 NR_104301.1 XM_017028738.1 XM_017028739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6775376 c.573G>T p.G191G Chr.22: 42560225-42560225 on Build GRCh38
3061875 c.600G>A p.T200T Chr.22: 42560252-42560252 on Build GRCh38
1225258 c.590G>T p.R197I Chr.22: 42560242-42560242 on Build GRCh38
371870 c.571G>A p.G191R Chr.22: 42560223-42560223 on Build GRCh38
726940 c.580C>G p.L194V Chr.22: 42560232-42560232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371875204 Chr.22: 42560284-42560284 on Build GRCh38
rs927523739 Chr.22: 42560526-42560526 on Build GRCh38
rs755546211 Chr.22: 42560388-42560388 on Build GRCh38
rs754325484 Chr.22: 42560274-42560274 on Build GRCh38
rs200900344 Chr.22: 42560218-42560218 on Build GRCh38
rs756314453 Chr.22: 42560441-42560441 on Build GRCh38
rs1051589011 Chr.22: 42560582-42560582 on Build GRCh38
rs1014400976 Chr.22: 42560358-42560358 on Build GRCh38
rs75210421 Chr.22: 42560204-42560204 on Build GRCh38
rs768357339 Chr.22: 42560288-42560288 on Build GRCh38
rs557997523 Chr.22: 42560251-42560251 on Build GRCh38
rs943229615 Chr.22: 42560452-42560452 on Build GRCh38
rs1013717259 Chr.22: 42560461-42560461 on Build GRCh38
rs774227929 Chr.22: 42560317-42560317 on Build GRCh38
rs943175875 Chr.22: 42560605-42560605 on Build GRCh38
rs980319170 Chr.22: 42560501-42560501 on Build GRCh38
rs1051579168 Chr.22: 42560436-42560436 on Build GRCh38
rs1000524030 Chr.22: 42560434-42560434 on Build GRCh38
rs945140269 Chr.22: 42560435-42560435 on Build GRCh38
rs75727575 Chr.22: 42560183-42560183 on Build GRCh38
rs752050760 Chr.22: 42560321-42560321 on Build GRCh38
rs779184626 Chr.22: 42560285-42560285 on Build GRCh38
rs1038362898 Chr.22: 42560374-42560374 on Build GRCh38
rs771701981 Chr.22: 42560201-42560201 on Build GRCh38
rs766780876 Chr.22: 42560316-42560316 on Build GRCh38
rs13054167 Chr.22: 42560407-42560407 on Build GRCh38
rs775150115 Chr.22: 42560199-42560199 on Build GRCh38
rs778326624 Chr.22: 42560198-42560198 on Build GRCh38
rs78094491 Chr.22: 42560481-42560481 on Build GRCh38
rs912668766 Chr.22: 42560583-42560583 on Build GRCh38
rs942512970 Chr.22: 42560593-42560593 on Build GRCh38
rs143547639 Chr.22: 42560482-42560482 on Build GRCh38
rs555149447 Chr.22: 42560530-42560530 on Build GRCh38
rs573539087 Chr.22: 42560542-42560542 on Build GRCh38
rs562877702 Chr.22: 42560627-42560627 on Build GRCh38
rs148540655 Chr.22: 42560223-42560223 on Build GRCh38
rs1029944957 Chr.22: 42560185-42560185 on Build GRCh38
rs566963383 Chr.22: 42560347-42560347 on Build GRCh38
rs756517281 Chr.22: 42560197-42560197 on Build GRCh38
rs367942039 Chr.22: 42560570-42560570 on Build GRCh38
rs781378984 Chr.22: 42560184-42560184 on Build GRCh38
rs150625034 Chr.22: 42560252-42560252 on Build GRCh38
rs1001690104 Chr.22: 42560520-42560520 on Build GRCh38
rs747651908 Chr.22: 42560299-42560299 on Build GRCh38
rs773044581 Chr.22: 42560311-42560311 on Build GRCh38
rs572099700 Chr.22: 42560585-42560585 on Build GRCh38
rs929988644 Chr.22: 42560568-42560568 on Build GRCh38
rs1030710672 Chr.22: 42560444-42560444 on Build GRCh38
rs537732904 Chr.22: 42560433-42560433 on Build GRCh38
rs970249339 Chr.22: 42560368-42560368 on Build GRCh38
rs367748289 Chr.22: 42560220-42560220 on Build GRCh38
rs759762227 Chr.22: 42560225-42560225 on Build GRCh38
rs989134714 Chr.22: 42560536-42560536 on Build GRCh38
rs55732589 Chr.22: 42560451-42560451 on Build GRCh38
rs1006739404 Chr.22: 42560403-42560403 on Build GRCh38
rs758209011 Chr.22: 42560182-42560182 on Build GRCh38
rs553547515 Chr.22: 42560565-42560565 on Build GRCh38
rs748691203 Chr.22: 42560187-42560187 on Build GRCh38
rs548344776 Chr.22: 42560326-42560326 on Build GRCh38
rs535643261 Chr.22: 42560494-42560494 on Build GRCh38
rs1039571642 Chr.22: 42560617-42560617 on Build GRCh38
rs146353799 Chr.22: 42560222-42560222 on Build GRCh38
rs918549755 Chr.22: 42560549-42560549 on Build GRCh38
rs544687355 Chr.22: 42560556-42560556 on Build GRCh38
rs769325277 Chr.22: 42560301-42560301 on Build GRCh38
rs1051193658 Chr.22: 42560399-42560399 on Build GRCh38
rs988975013 Chr.22: 42560273-42560273 on Build GRCh38
rs1024437026 Chr.22: 42560497-42560497 on Build GRCh38
rs930101703 Chr.22: 42560385-42560385 on Build GRCh38
rs139417319 Chr.22: 42560380-42560380 on Build GRCh38
rs1048217123 Chr.22: 42560475-42560475 on Build GRCh38
rs559876636 Chr.22: 42560466-42560466 on Build GRCh38
rs141883409 Chr.22: 42560233-42560233 on Build GRCh38
rs899115940 Chr.22: 42560455-42560455 on Build GRCh38
rs533690443 Chr.22: 42560470-42560470 on Build GRCh38
rs745759564 Chr.22: 42560350-42560350 on Build GRCh38
rs142867325 Chr.22: 42560205-42560205 on Build GRCh38
rs890357868 Chr.22: 42560248-42560248 on Build GRCh38
rs780893518 Chr.22: 42560291-42560291 on Build GRCh38
rs957639561 Chr.22: 42560532-42560532 on Build GRCh38
rs980731830 Chr.22: 42560292-42560292 on Build GRCh38
rs768298869 Chr.22: 42560574-42560574 on Build GRCh38
rs890921839 Chr.22: 42560402-42560402 on Build GRCh38
rs147360692 Chr.22: 42560278-42560278 on Build GRCh38
rs139695066 Chr.22: 42560267-42560267 on Build GRCh38
rs781067318 Chr.22: 42560219-42560219 on Build GRCh38
rs913573251 Chr.22: 42560567-42560567 on Build GRCh38
rs987631561 Chr.22: 42560579-42560579 on Build GRCh38
rs1000743025 Chr.22: 42560628-42560628 on Build GRCh38
rs767644863 Chr.22: 42560228-42560228 on Build GRCh38
rs974043252 Chr.22: 42560546-42560546 on Build GRCh38
rs751859494 Chr.22: 42560224-42560224 on Build GRCh38
rs941347152 Chr.22: 42560359-42560359 on Build GRCh38
rs752736653 Chr.22: 42560423-42560423 on Build GRCh38
rs746680221 Chr.22: 42560287-42560287 on Build GRCh38
rs570539180 Chr.22: 42560430-42560430 on Build GRCh38

View Full Product Details