Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 13712640-13713173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC2
Entrez Gene ID
6399
Gene Name
trafficking protein particle complex 2
Gene Aliases
MIP2A, SEDL, SEDT, TRAPPC2P1, TRS20, ZNF547L, hYP38334
UniGene
Hs.592238
Gene Chromosome Location
Chr.X: 13712242-13734635 on Build GRCh38
Transcripts
NM_001011658.3 NM_001128835.2 NM_014563.5 XM_011545565.1 XM_011545566.2

SNP Detail

SNP ID Chromosome Location
rs1058892 Chr.X: 13712888-13712888 on Build GRCh38
rs879233903 Chr.X: 13713065-13713065 on Build GRCh38
rs865815058 Chr.X: 13713103-13713103 on Build GRCh38
rs771455096 Chr.X: 13712965-13712965 on Build GRCh38
rs1058897 Chr.X: 13712795-13712795 on Build GRCh38
rs1058889 Chr.X: 13712930-13712930 on Build GRCh38
rs756413153 Chr.X: 13712856-13712856 on Build GRCh38
rs187740735 Chr.X: 13712921-13712921 on Build GRCh38
rs558114277 Chr.X: 13713117-13713119 on Build GRCh38
rs146362855 Chr.X: 13712808-13712808 on Build GRCh38
rs148298471 Chr.X: 13712964-13712964 on Build GRCh38
rs774359940 Chr.X: 13712992-13712992 on Build GRCh38
rs1058894 Chr.X: 13712857-13712857 on Build GRCh38
rs192328990 Chr.X: 13712971-13712971 on Build GRCh38
rs187464655 Chr.X: 13712773-13712773 on Build GRCh38
rs184839802 Chr.X: 13712846-13712846 on Build GRCh38
rs1058891 Chr.X: 13712897-13712897 on Build GRCh38
rs1058888 Chr.X: 13712934-13712934 on Build GRCh38
rs1058893 Chr.X: 13712868-13712868 on Build GRCh38
rs914009383 Chr.X: 13712868-13712868 on Build GRCh38
rs7716 Chr.X: 13712668-13712668 on Build GRCh38
rs1136277 Chr.X: 13713054-13713054 on Build GRCh38
rs1058887 Chr.X: 13712968-13712968 on Build GRCh38
rs1136278 Chr.X: 13713049-13713049 on Build GRCh38
rs192015551 Chr.X: 13712802-13712802 on Build GRCh38
rs566484686 Chr.X: 13713021-13713021 on Build GRCh38
rs184278401 Chr.X: 13713059-13713059 on Build GRCh38
rs1046405564 Chr.X: 13713058-13713058 on Build GRCh38
rs1136276 Chr.X: 13713055-13713055 on Build GRCh38
rs1058895 Chr.X: 13712839-13712839 on Build GRCh38
rs1136280 Chr.X: 13713022-13713022 on Build GRCh38
rs1058886 Chr.X: 13712970-13712970 on Build GRCh38
rs766836189 Chr.X: 13712959-13712960 on Build GRCh38
rs35590746 Chr.X: 13713104-13713104 on Build GRCh38
rs1064528 Chr.X: 13712843-13712843 on Build GRCh38
rs1058890 Chr.X: 13712926-13712926 on Build GRCh38
rs1058896 Chr.X: 13712807-13712807 on Build GRCh38
rs878988409 Chr.X: 13713047-13713047 on Build GRCh38

View Full Product Details