Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11311847-11312258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN16
Entrez Gene ID
26526
Gene Name
tetraspanin 16
Gene Aliases
TM-8, TM4-B, TM4SF16
UniGene
Hs.579784
Gene Chromosome Location
Chr.19: 11296139-11326996 on Build GRCh38
Transcripts
NM_001282509.1 NM_001282510.1 NM_012466.3 NR_104199.1 XM_011527901.1 XM_011527902.1 XM_011527903.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3528527 c.613C>T p.H205Y Chr.19: 11312148-11312148 on Build GRCh38
3821927 c.642G>T p.Q214H Chr.19: 11312177-11312177 on Build GRCh38
6368565 c.657T>A p.S219R Chr.19: 11312192-11312192 on Build GRCh38
5854617 c.604G>A p.G202S Chr.19: 11312139-11312139 on Build GRCh38
6294731 c.608G>T p.C203F Chr.19: 11312143-11312143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764407493 Chr.19: 11312090-11312090 on Build GRCh38
rs570587158 Chr.19: 11312177-11312177 on Build GRCh38
rs939929427 Chr.19: 11311873-11311873 on Build GRCh38
rs997926451 Chr.19: 11312047-11312047 on Build GRCh38
rs963974027 Chr.19: 11311908-11311908 on Build GRCh38
rs768788950 Chr.19: 11312207-11312207 on Build GRCh38
rs916172097 Chr.19: 11311973-11311973 on Build GRCh38
rs966234500 Chr.19: 11312021-11312021 on Build GRCh38
rs917074069 Chr.19: 11311932-11311932 on Build GRCh38
rs1037283723 Chr.19: 11311905-11311905 on Build GRCh38
rs557865636 Chr.19: 11312020-11312020 on Build GRCh38
rs12983513 Chr.19: 11312141-11312141 on Build GRCh38
rs141964186 Chr.19: 11311914-11311914 on Build GRCh38
rs760850776 Chr.19: 11312206-11312206 on Build GRCh38
rs758243490 Chr.19: 11312114-11312114 on Build GRCh38
rs201270086 Chr.19: 11312131-11312131 on Build GRCh38
rs147697998 Chr.19: 11312205-11312205 on Build GRCh38
rs376613720 Chr.19: 11312219-11312219 on Build GRCh38
rs540278287 Chr.19: 11312046-11312046 on Build GRCh38
rs757122469 Chr.19: 11312098-11312098 on Build GRCh38
rs765126446 Chr.19: 11312228-11312228 on Build GRCh38
rs750151278 Chr.19: 11312233-11312233 on Build GRCh38
rs899782002 Chr.19: 11312049-11312049 on Build GRCh38
rs1052147271 Chr.19: 11312009-11312009 on Build GRCh38
rs762077644 Chr.19: 11312221-11312221 on Build GRCh38
rs1029953686 Chr.19: 11312082-11312082 on Build GRCh38
rs571009351 Chr.19: 11311993-11311993 on Build GRCh38
rs545911886 Chr.19: 11312008-11312008 on Build GRCh38
rs927382584 Chr.19: 11312231-11312231 on Build GRCh38
rs928829852 Chr.19: 11312000-11312000 on Build GRCh38
rs183907342 Chr.19: 11312117-11312117 on Build GRCh38
rs955156320 Chr.19: 11311931-11311931 on Build GRCh38
rs559590863 Chr.19: 11312080-11312080 on Build GRCh38
rs529953554 Chr.19: 11312084-11312084 on Build GRCh38
rs750130951 Chr.19: 11312198-11312201 on Build GRCh38
rs551252181 Chr.19: 11311964-11311964 on Build GRCh38
rs750395687 Chr.19: 11312105-11312105 on Build GRCh38
rs1044495125 Chr.19: 11311939-11311939 on Build GRCh38
rs950135248 Chr.19: 11311938-11311938 on Build GRCh38
rs892058856 Chr.19: 11311997-11311997 on Build GRCh38
rs572881364 Chr.19: 11312035-11312035 on Build GRCh38
rs770431286 Chr.19: 11311989-11311989 on Build GRCh38
rs192026480 Chr.19: 11312101-11312101 on Build GRCh38
rs746514757 Chr.19: 11312121-11312121 on Build GRCh38
rs962718785 Chr.19: 11311971-11311971 on Build GRCh38
rs948998559 Chr.19: 11311974-11311974 on Build GRCh38
rs754679131 Chr.19: 11312122-11312122 on Build GRCh38
rs536010140 Chr.19: 11311944-11311944 on Build GRCh38
rs777022293 Chr.19: 11312220-11312220 on Build GRCh38
rs746438605 Chr.19: 11312150-11312150 on Build GRCh38
rs974520209 Chr.19: 11312139-11312139 on Build GRCh38
rs1028035610 Chr.19: 11312067-11312067 on Build GRCh38
rs1044213996 Chr.19: 11312211-11312211 on Build GRCh38
rs1029489767 Chr.19: 11311916-11311916 on Build GRCh38
rs775707184 Chr.19: 11312183-11312183 on Build GRCh38
rs553678704 Chr.19: 11311897-11311897 on Build GRCh38
rs905913931 Chr.19: 11311946-11311946 on Build GRCh38
rs754054885 Chr.19: 11312092-11312092 on Build GRCh38
rs771318324 Chr.19: 11312140-11312140 on Build GRCh38

View Full Product Details