Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 51650213-51650757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD6
Entrez Gene ID
122786
Gene Name
FERM domain containing 6
Gene Aliases
C14orf31, EX1, Willin, c14_5320
UniGene
Hs.434914
Gene Chromosome Location
Chr.14: 51489100-51730727 on Build GRCh38
Transcripts
NM_001042481.2 XM_011536423.1 XM_011536424.1 NR_037676.1

Gene Symbol
FRMD6-AS1
Entrez Gene ID
145438
Gene Name
FRMD6 antisense RNA 1
Gene Aliases
C14orf82
UniGene
Hs.645410
Gene Chromosome Location
Chr.14: 51649516-51651744 on Build GRCh38
Transcripts
NM_001042481.2 XM_011536423.1 XM_011536424.1 NR_037676.1

SNP Detail

SNP ID Chromosome Location
rs1018421150 Chr.14: 51650624-51650625 on Build GRCh38
rs934183692 Chr.14: 51650594-51650609 on Build GRCh38
rs930758788 Chr.14: 51650570-51650570 on Build GRCh38
rs1049243385 Chr.14: 51650735-51650735 on Build GRCh38
rs572034304 Chr.14: 51650343-51650343 on Build GRCh38
rs930809577 Chr.14: 51650722-51650722 on Build GRCh38
rs911381400 Chr.14: 51650371-51650371 on Build GRCh38
rs543327663 Chr.14: 51650514-51650514 on Build GRCh38
rs1027705660 Chr.14: 51650453-51650453 on Build GRCh38
rs945677181 Chr.14: 51650401-51650401 on Build GRCh38
rs952849657 Chr.14: 51650369-51650369 on Build GRCh38
rs986469079 Chr.14: 51650675-51650675 on Build GRCh38
rs1001468827 Chr.14: 51650620-51650620 on Build GRCh38
rs944974202 Chr.14: 51650544-51650544 on Build GRCh38
rs781170714 Chr.14: 51650393-51650394 on Build GRCh38
rs886492147 Chr.14: 51650593-51650593 on Build GRCh38
rs942326156 Chr.14: 51650692-51650692 on Build GRCh38
rs866438407 Chr.14: 51650402-51650402 on Build GRCh38
rs900622909 Chr.14: 51650443-51650443 on Build GRCh38
rs918703284 Chr.14: 51650349-51650349 on Build GRCh38
rs983374173 Chr.14: 51650312-51650312 on Build GRCh38
rs906516774 Chr.14: 51650599-51650599 on Build GRCh38
rs776614565 Chr.14: 51650721-51650721 on Build GRCh38
rs540670233 Chr.14: 51650403-51650403 on Build GRCh38
rs989862311 Chr.14: 51650505-51650505 on Build GRCh38
rs957400572 Chr.14: 51650497-51650497 on Build GRCh38
rs558594638 Chr.14: 51650289-51650292 on Build GRCh38
rs771742710 Chr.14: 51650391-51650392 on Build GRCh38
rs977691168 Chr.14: 51650707-51650707 on Build GRCh38
rs2884099 Chr.14: 51650448-51650448 on Build GRCh38
rs945729897 Chr.14: 51650521-51650521 on Build GRCh38
rs989913152 Chr.14: 51650638-51650638 on Build GRCh38
rs1009508529 Chr.14: 51650435-51650435 on Build GRCh38
rs912154275 Chr.14: 51650535-51650535 on Build GRCh38
rs917129980 Chr.14: 51650327-51650327 on Build GRCh38
rs573051451 Chr.14: 51650618-51650618 on Build GRCh38
rs927061033 Chr.14: 51650341-51650341 on Build GRCh38
rs900676446 Chr.14: 51650565-51650565 on Build GRCh38
rs1024238754 Chr.14: 51650482-51650482 on Build GRCh38
rs910796117 Chr.14: 51650626-51650626 on Build GRCh38
rs933016727 Chr.14: 51650515-51650515 on Build GRCh38
rs1054085015 Chr.14: 51650397-51650397 on Build GRCh38
rs979949101 Chr.14: 51650339-51650339 on Build GRCh38
rs1049360121 Chr.14: 51650616-51650616 on Build GRCh38
rs977019088 Chr.14: 51650659-51650659 on Build GRCh38
rs562250725 Chr.14: 51650647-51650647 on Build GRCh38
rs967082239 Chr.14: 51650652-51650652 on Build GRCh38
rs185632187 Chr.14: 51650714-51650714 on Build GRCh38
rs545532408 Chr.14: 51650392-51650393 on Build GRCh38
rs984327166 Chr.14: 51650370-51650370 on Build GRCh38
rs1024309352 Chr.14: 51650615-51650615 on Build GRCh38
rs12590456 Chr.14: 51650522-51650522 on Build GRCh38
rs11318703 Chr.14: 51650392-51650392 on Build GRCh38
rs1004861747 Chr.14: 51650465-51650465 on Build GRCh38
rs1039183670 Chr.14: 51650440-51650440 on Build GRCh38
rs957246656 Chr.14: 51650637-51650637 on Build GRCh38
rs975023092 Chr.14: 51650720-51650720 on Build GRCh38
rs1012140252 Chr.14: 51650592-51650592 on Build GRCh38
rs980384745 Chr.14: 51650489-51650489 on Build GRCh38
rs971652881 Chr.14: 51650329-51650329 on Build GRCh38
rs796902005 Chr.14: 51650400-51650400 on Build GRCh38
rs866040068 Chr.14: 51650490-51650490 on Build GRCh38
rs935764111 Chr.14: 51650391-51650391 on Build GRCh38
rs527784400 Chr.14: 51650669-51650669 on Build GRCh38
rs869068811 Chr.14: 51650413-51650414 on Build GRCh38
rs540868805 Chr.14: 51650412-51650413 on Build GRCh38
rs181495471 Chr.14: 51650583-51650583 on Build GRCh38
rs1004912674 Chr.14: 51650605-51650605 on Build GRCh38
rs922943851 Chr.14: 51650502-51650503 on Build GRCh38
rs912960554 Chr.14: 51650519-51650519 on Build GRCh38
rs555446935 Chr.14: 51650270-51650270 on Build GRCh38
rs1039643976 Chr.14: 51650550-51650550 on Build GRCh38
rs35868898 Chr.14: 51650456-51650456 on Build GRCh38
rs912221340 Chr.14: 51650684-51650684 on Build GRCh38
rs1034642434 Chr.14: 51650621-51650621 on Build GRCh38
rs977428196 Chr.14: 51650389-51650389 on Build GRCh38
rs942937840 Chr.14: 51650381-51650381 on Build GRCh38
rs868136387 Chr.14: 51650545-51650545 on Build GRCh38
rs890684634 Chr.14: 51650428-51650428 on Build GRCh38
rs927143264 Chr.14: 51650494-51650494 on Build GRCh38
rs1024945503 Chr.14: 51650268-51650268 on Build GRCh38
rs72145350 Chr.14: 51650285-51650288 on Build GRCh38
rs971323474 Chr.14: 51650486-51650486 on Build GRCh38
rs922880383 Chr.14: 51650672-51650672 on Build GRCh38

View Full Product Details