Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68804929-68805478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANP32A
Entrez Gene ID
8125
Gene Name
acidic nuclear phosphoprotein 32 family member A
Gene Aliases
C15orf1, HPPCn, I1PP2A, LANP, MAPM, PHAP1, PHAPI, PP32
UniGene
Hs.458747
Gene Chromosome Location
Chr.15: 68778535-68820922 on Build GRCh38
Transcripts
NM_006305.3 NR_026808.1

Gene Symbol
ANP32A-IT1
Entrez Gene ID
80035
Gene Name
ANP32A intronic transcript 1
Gene Aliases
C15orf28, HsT18971, NCRNA00321
UniGene
Hs.662150
Gene Chromosome Location
Chr.15: 68803821-68807101 on Build GRCh38
Transcripts
NM_006305.3 NR_026808.1

SNP Detail

SNP ID Chromosome Location
rs919718335 Chr.15: 68805008-68805008 on Build GRCh38
rs386785060 Chr.15: 68805256-68805257 on Build GRCh38
rs946502571 Chr.15: 68805048-68805048 on Build GRCh38
rs990704982 Chr.15: 68805015-68805015 on Build GRCh38
rs1043825768 Chr.15: 68805068-68805068 on Build GRCh38
rs960598317 Chr.15: 68804976-68804976 on Build GRCh38
rs62008192 Chr.15: 68804973-68804973 on Build GRCh38
rs897963849 Chr.15: 68805451-68805451 on Build GRCh38
rs924073951 Chr.15: 68805338-68805338 on Build GRCh38
rs34496286 Chr.15: 68805450-68805451 on Build GRCh38
rs575503999 Chr.15: 68805080-68805080 on Build GRCh38
rs2958405 Chr.15: 68805267-68805267 on Build GRCh38
rs544375269 Chr.15: 68805202-68805202 on Build GRCh38
rs139131823 Chr.15: 68805216-68805216 on Build GRCh38
rs747356042 Chr.15: 68805442-68805442 on Build GRCh38
rs564991431 Chr.15: 68805292-68805292 on Build GRCh38
rs1470902 Chr.15: 68805453-68805453 on Build GRCh38
rs549132439 Chr.15: 68805215-68805215 on Build GRCh38
rs752453456 Chr.15: 68805212-68805212 on Build GRCh38
rs901826688 Chr.15: 68805206-68805206 on Build GRCh38
rs996398960 Chr.15: 68805192-68805192 on Build GRCh38
rs754140220 Chr.15: 68805207-68805207 on Build GRCh38
rs990737662 Chr.15: 68805275-68805275 on Build GRCh38
rs935386393 Chr.15: 68805140-68805140 on Build GRCh38
rs1040410080 Chr.15: 68805340-68805340 on Build GRCh38
rs1040336390 Chr.15: 68805142-68805142 on Build GRCh38
rs1470901 Chr.15: 68805426-68805426 on Build GRCh38
rs902134655 Chr.15: 68805159-68805159 on Build GRCh38
rs28474727 Chr.15: 68805256-68805256 on Build GRCh38
rs551122308 Chr.15: 68805366-68805366 on Build GRCh38
rs146865702 Chr.15: 68805013-68805013 on Build GRCh38
rs77247539 Chr.15: 68805448-68805448 on Build GRCh38
rs931025150 Chr.15: 68805319-68805319 on Build GRCh38
rs1000955744 Chr.15: 68805263-68805263 on Build GRCh38
rs186282680 Chr.15: 68805255-68805255 on Build GRCh38
rs778227773 Chr.15: 68805293-68805293 on Build GRCh38
rs927887470 Chr.15: 68804975-68804975 on Build GRCh38
rs772308953 Chr.15: 68805266-68805266 on Build GRCh38
rs935405627 Chr.15: 68805339-68805339 on Build GRCh38
rs1050645580 Chr.15: 68805378-68805378 on Build GRCh38
rs530923484 Chr.15: 68805301-68805301 on Build GRCh38
rs538114928 Chr.15: 68805300-68805300 on Build GRCh38
rs538719716 Chr.15: 68805046-68805046 on Build GRCh38
rs143147231 Chr.15: 68805297-68805297 on Build GRCh38
rs887956007 Chr.15: 68805205-68805205 on Build GRCh38
rs1000926085 Chr.15: 68805445-68805445 on Build GRCh38
rs370008419 Chr.15: 68805195-68805195 on Build GRCh38
rs756775254 Chr.15: 68805269-68805269 on Build GRCh38
rs916435389 Chr.15: 68805023-68805023 on Build GRCh38
rs992454960 Chr.15: 68805271-68805271 on Build GRCh38
rs886612279 Chr.15: 68805089-68805089 on Build GRCh38
rs924013723 Chr.15: 68805082-68805082 on Build GRCh38
rs28599719 Chr.15: 68805257-68805257 on Build GRCh38
rs923294894 Chr.15: 68805351-68805351 on Build GRCh38
rs748143017 Chr.15: 68805367-68805367 on Build GRCh38
rs757567203 Chr.15: 68805228-68805228 on Build GRCh38
rs558722497 Chr.15: 68805056-68805056 on Build GRCh38
rs758791037 Chr.15: 68805377-68805377 on Build GRCh38
rs554477663 Chr.15: 68805214-68805214 on Build GRCh38
rs940916603 Chr.15: 68805144-68805144 on Build GRCh38
rs1024917470 Chr.15: 68805272-68805272 on Build GRCh38

View Full Product Details