Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18470517-18471017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR3192
Entrez Gene ID
100422875
Gene Name
microRNA 3192
Gene Aliases
mir-3192
UniGene
Gene Chromosome Location
Chr.20: 18470615-18470691 on Build GRCh38
Transcripts
NR_036160.1 NM_001282526.1 NM_006466.3 NR_104209.1 XM_011529141.1 XM_017027596.1

Gene Symbol
POLR3F
Entrez Gene ID
10621
Gene Name
RNA polymerase III subunit F
Gene Aliases
RPC39, RPC6
UniGene
Hs.472227
Gene Chromosome Location
Chr.20: 18467389-18484648 on Build GRCh38
Transcripts
NR_036160.1 NM_001282526.1 NM_006466.3 NR_104209.1 XM_011529141.1 XM_017027596.1

SNP Detail

SNP ID Chromosome Location
rs924463363 Chr.20: 18470710-18470710 on Build GRCh38
rs989408279 Chr.20: 18470750-18470750 on Build GRCh38
rs11907020 Chr.20: 18470681-18470681 on Build GRCh38
rs1019026003 Chr.20: 18470670-18470670 on Build GRCh38
rs940442404 Chr.20: 18470890-18470890 on Build GRCh38
rs938589670 Chr.20: 18470745-18470745 on Build GRCh38
rs948425508 Chr.20: 18470771-18470771 on Build GRCh38
rs995052311 Chr.20: 18470645-18470645 on Build GRCh38
rs977637464 Chr.20: 18470708-18470708 on Build GRCh38
rs773140898 Chr.20: 18470629-18470629 on Build GRCh38
rs375180685 Chr.20: 18470827-18470827 on Build GRCh38
rs936492653 Chr.20: 18470706-18470706 on Build GRCh38
rs1046206524 Chr.20: 18470614-18470614 on Build GRCh38
rs766189405 Chr.20: 18470663-18470663 on Build GRCh38
rs1037380173 Chr.20: 18470787-18470787 on Build GRCh38
rs561865585 Chr.20: 18470985-18470985 on Build GRCh38
rs6045423 Chr.20: 18470545-18470545 on Build GRCh38
rs906316278 Chr.20: 18470886-18470886 on Build GRCh38
rs543684545 Chr.20: 18470944-18470944 on Build GRCh38
rs6111978 Chr.20: 18470656-18470656 on Build GRCh38
rs996045251 Chr.20: 18470587-18470587 on Build GRCh38
rs746444840 Chr.20: 18470696-18470696 on Build GRCh38
rs906254403 Chr.20: 18470626-18470626 on Build GRCh38
rs916486000 Chr.20: 18470759-18470759 on Build GRCh38
rs951585476 Chr.20: 18470653-18470653 on Build GRCh38
rs977500820 Chr.20: 18470687-18470687 on Build GRCh38
rs1019916057 Chr.20: 18470945-18470945 on Build GRCh38
rs898871159 Chr.20: 18470813-18470813 on Build GRCh38
rs899297329 Chr.20: 18470635-18470635 on Build GRCh38
rs557918308 Chr.20: 18470903-18470903 on Build GRCh38
rs775629928 Chr.20: 18470664-18470664 on Build GRCh38
rs759475994 Chr.20: 18470669-18470669 on Build GRCh38
rs929089330 Chr.20: 18470834-18470834 on Build GRCh38
rs889363863 Chr.20: 18470989-18470989 on Build GRCh38
rs898657180 Chr.20: 18470549-18470549 on Build GRCh38
rs6045424 Chr.20: 18470590-18470590 on Build GRCh38
rs915575736 Chr.20: 18470751-18470751 on Build GRCh38
rs536150870 Chr.20: 18470565-18470565 on Build GRCh38
rs1036072514 Chr.20: 18470893-18470893 on Build GRCh38
rs576192678 Chr.20: 18470906-18470906 on Build GRCh38
rs1046093961 Chr.20: 18470800-18470800 on Build GRCh38
rs376614009 Chr.20: 18470634-18470634 on Build GRCh38
rs554130490 Chr.20: 18470627-18470627 on Build GRCh38
rs947723335 Chr.20: 18470784-18470784 on Build GRCh38
rs953409564 Chr.20: 18470659-18470659 on Build GRCh38
rs899821808 Chr.20: 18470977-18470977 on Build GRCh38

View Full Product Details