Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44965449-44965998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK4-AS1
Entrez Gene ID
100505826
Gene Name
STK4 antisense RNA 1 (head to head)
Gene Aliases
-
UniGene
Hs.196065
Gene Chromosome Location
Chr.20: 44963794-44966458 on Build GRCh38
Transcripts
NR_038341.1

SNP Detail

SNP ID Chromosome Location
rs1033832538 Chr.20: 44965913-44965916 on Build GRCh38
rs376150901 Chr.20: 44965794-44965796 on Build GRCh38
rs945775367 Chr.20: 44965923-44965923 on Build GRCh38
rs891629552 Chr.20: 44965480-44965480 on Build GRCh38
rs552915422 Chr.20: 44965892-44965892 on Build GRCh38
rs1010111087 Chr.20: 44965484-44965484 on Build GRCh38
rs754409586 Chr.20: 44965528-44965528 on Build GRCh38
rs1045567145 Chr.20: 44965943-44965943 on Build GRCh38
rs1054614624 Chr.20: 44965863-44965863 on Build GRCh38
rs754897888 Chr.20: 44965575-44965575 on Build GRCh38
rs548660831 Chr.20: 44965604-44965604 on Build GRCh38
rs969804899 Chr.20: 44965530-44965530 on Build GRCh38
rs976310849 Chr.20: 44965668-44965668 on Build GRCh38
rs528591233 Chr.20: 44965589-44965589 on Build GRCh38
rs949669211 Chr.20: 44965860-44965866 on Build GRCh38
rs1014971932 Chr.20: 44965613-44965613 on Build GRCh38
rs190785111 Chr.20: 44965582-44965582 on Build GRCh38
rs995027578 Chr.20: 44965930-44965930 on Build GRCh38
rs985289458 Chr.20: 44965597-44965597 on Build GRCh38
rs561365613 Chr.20: 44965529-44965529 on Build GRCh38
rs541537669 Chr.20: 44965903-44965903 on Build GRCh38
rs371359267 Chr.20: 44965731-44965731 on Build GRCh38
rs148823318 Chr.20: 44965559-44965559 on Build GRCh38
rs140062906 Chr.20: 44965664-44965665 on Build GRCh38
rs147983639 Chr.20: 44965722-44965722 on Build GRCh38
rs1034305662 Chr.20: 44965579-44965579 on Build GRCh38
rs1026540814 Chr.20: 44965599-44965599 on Build GRCh38
rs1054180842 Chr.20: 44965478-44965478 on Build GRCh38
rs532832966 Chr.20: 44965562-44965562 on Build GRCh38
rs1045430195 Chr.20: 44965879-44965879 on Build GRCh38
rs188002420 Chr.20: 44965890-44965890 on Build GRCh38
rs530387300 Chr.20: 44965532-44965532 on Build GRCh38
rs925808691 Chr.20: 44965771-44965771 on Build GRCh38
rs923420623 Chr.20: 44965673-44965673 on Build GRCh38
rs570686958 Chr.20: 44965813-44965813 on Build GRCh38
rs943487939 Chr.20: 44965702-44965702 on Build GRCh38
rs907024508 Chr.20: 44965967-44965967 on Build GRCh38
rs543710555 Chr.20: 44965545-44965545 on Build GRCh38
rs891492356 Chr.20: 44965509-44965509 on Build GRCh38
rs778743481 Chr.20: 44965570-44965570 on Build GRCh38
rs143420519 Chr.20: 44965657-44965657 on Build GRCh38
rs1001718181 Chr.20: 44965910-44965910 on Build GRCh38
rs573774146 Chr.20: 44965593-44965593 on Build GRCh38
rs899304222 Chr.20: 44965918-44965918 on Build GRCh38
rs1026488761 Chr.20: 44965961-44965961 on Build GRCh38
rs891732368 Chr.20: 44965856-44965856 on Build GRCh38
rs751051204 Chr.20: 44965524-44965524 on Build GRCh38
rs202012683 Chr.20: 44965784-44965786 on Build GRCh38
rs182969900 Chr.20: 44965784-44965784 on Build GRCh38
rs905611092 Chr.20: 44965906-44965907 on Build GRCh38
rs572961548 Chr.20: 44965898-44965898 on Build GRCh38
rs370075341 Chr.20: 44965697-44965697 on Build GRCh38
rs935644841 Chr.20: 44965850-44965850 on Build GRCh38
rs186330253 Chr.20: 44965564-44965564 on Build GRCh38
rs891578424 Chr.20: 44965896-44965896 on Build GRCh38

View Full Product Details